Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54029797-54030291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54056030 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC5
Entrez Gene ID
3222
Gene Name
homeobox C5
Gene Aliases
CP11, HOX3, HOX3D
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54035361 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC6
Entrez Gene ID
3223
Gene Name
homeobox C6
Gene Aliases
CP25, HHO.C8, HOX3, HOX3C
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54030823 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168849 c.628G>A p.G210S Chr.12: 54029882-54029882 on Build GRCh38
6553341 c.628G>T p.G210C Chr.12: 54029882-54029882 on Build GRCh38
1933601 c.643G>A p.A215T Chr.12: 54029897-54029897 on Build GRCh38
5510608 c.627C>G p.G209G Chr.12: 54029881-54029881 on Build GRCh38
1188514 c.575G>A p.R192H Chr.12: 54029829-54029829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976125070 Chr.12: 54029828-54029828 on Build GRCh38
rs1030404980 Chr.12: 54030122-54030122 on Build GRCh38
rs916716448 Chr.12: 54029895-54029895 on Build GRCh38
rs954922927 Chr.12: 54030200-54030200 on Build GRCh38
rs200427348 Chr.12: 54029879-54029879 on Build GRCh38
rs373295522 Chr.12: 54029870-54029870 on Build GRCh38
rs369084398 Chr.12: 54029863-54029863 on Build GRCh38
rs939957363 Chr.12: 54030018-54030018 on Build GRCh38
rs1018610306 Chr.12: 54030037-54030038 on Build GRCh38
rs148638675 Chr.12: 54029865-54029865 on Build GRCh38
rs768294867 Chr.12: 54029884-54029884 on Build GRCh38
rs749663420 Chr.12: 54030260-54030260 on Build GRCh38
rs536100834 Chr.12: 54030202-54030202 on Build GRCh38
rs141463318 Chr.12: 54030156-54030156 on Build GRCh38
rs775634833 Chr.12: 54029876-54029876 on Build GRCh38
rs759954812 Chr.12: 54029900-54029900 on Build GRCh38
rs747214600 Chr.12: 54029853-54029853 on Build GRCh38
rs924130326 Chr.12: 54030248-54030248 on Build GRCh38
rs753211992 Chr.12: 54029909-54029909 on Build GRCh38
rs781534587 Chr.12: 54029941-54029941 on Build GRCh38
rs553588814 Chr.12: 54030131-54030131 on Build GRCh38
rs954142535 Chr.12: 54030243-54030243 on Build GRCh38
rs775505948 Chr.12: 54029833-54029833 on Build GRCh38
rs904782541 Chr.12: 54030142-54030142 on Build GRCh38
rs1046125618 Chr.12: 54029899-54029899 on Build GRCh38
rs771141589 Chr.12: 54029858-54029858 on Build GRCh38
rs777768728 Chr.12: 54029840-54029840 on Build GRCh38
rs746975549 Chr.12: 54030123-54030123 on Build GRCh38
rs768523141 Chr.12: 54030229-54030229 on Build GRCh38
rs777858592 Chr.12: 54029930-54029930 on Build GRCh38
rs147247423 Chr.12: 54029892-54029893 on Build GRCh38
rs1042245 Chr.12: 54030152-54030152 on Build GRCh38
rs965238815 Chr.12: 54030093-54030093 on Build GRCh38
rs746073698 Chr.12: 54029942-54029942 on Build GRCh38
rs756628790 Chr.12: 54030120-54030120 on Build GRCh38
rs768676417 Chr.12: 54029991-54029991 on Build GRCh38
rs1006773536 Chr.12: 54030017-54030017 on Build GRCh38
rs749261491 Chr.12: 54029985-54029985 on Build GRCh38
rs767242025 Chr.12: 54029894-54029894 on Build GRCh38
rs983949510 Chr.12: 54030246-54030246 on Build GRCh38
rs765056816 Chr.12: 54029874-54029875 on Build GRCh38
rs1036679240 Chr.12: 54030090-54030090 on Build GRCh38
rs758850699 Chr.12: 54029910-54029910 on Build GRCh38
rs965422041 Chr.12: 54030166-54030166 on Build GRCh38
rs201035626 Chr.12: 54029933-54029933 on Build GRCh38
rs1021782622 Chr.12: 54030139-54030139 on Build GRCh38
rs750132624 Chr.12: 54029938-54029939 on Build GRCh38
rs202121790 Chr.12: 54029885-54029885 on Build GRCh38
rs950687160 Chr.12: 54029851-54029851 on Build GRCh38
rs932521351 Chr.12: 54029918-54029918 on Build GRCh38
rs999087321 Chr.12: 54030147-54030147 on Build GRCh38
rs983478792 Chr.12: 54029854-54029854 on Build GRCh38
rs765585868 Chr.12: 54029905-54029905 on Build GRCh38
rs976893207 Chr.12: 54029874-54029874 on Build GRCh38
rs535021715 Chr.12: 54030101-54030101 on Build GRCh38
rs949544340 Chr.12: 54030208-54030208 on Build GRCh38
rs762837267 Chr.12: 54029972-54029973 on Build GRCh38
rs779937440 Chr.12: 54029983-54029983 on Build GRCh38
rs977220574 Chr.12: 54030189-54030189 on Build GRCh38
rs868498553 Chr.12: 54030002-54030002 on Build GRCh38
rs761405003 Chr.12: 54029891-54029891 on Build GRCh38
rs761693320 Chr.12: 54029842-54029843 on Build GRCh38
rs750024894 Chr.12: 54029896-54029896 on Build GRCh38
rs939068866 Chr.12: 54030118-54030118 on Build GRCh38
rs769716268 Chr.12: 54029962-54029962 on Build GRCh38
rs1249078 Chr.12: 54030207-54030207 on Build GRCh38
rs991085658 Chr.12: 54030159-54030159 on Build GRCh38
rs906227310 Chr.12: 54030116-54030116 on Build GRCh38

View Full Product Details