Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54582640-54583136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R1A
Entrez Gene ID
5502
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1A
Gene Aliases
I1, IPP1
UniGene
Hs.505662
Gene Chromosome Location
Chr.12: 54579240-54588659 on Build GRCh38
Transcripts
NM_006741.3 XM_005268995.4 XM_006719471.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871986 c.185C>T p.S62F Chr.12: 54582794-54582794 on Build GRCh38
6381041 c.238A>G p.T80A Chr.12: 54582741-54582741 on Build GRCh38
1663644 c.228G>C p.R76S Chr.12: 54582751-54582751 on Build GRCh38
3462878 c.227G>A p.R76K Chr.12: 54582752-54582752 on Build GRCh38
6862424 c.184-8C>A p.? Chr.12: 54582803-54582803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746153163 Chr.12: 54582723-54582723 on Build GRCh38
rs964068425 Chr.12: 54582854-54582854 on Build GRCh38
rs112270496 Chr.12: 54582983-54582983 on Build GRCh38
rs752066559 Chr.12: 54582833-54582833 on Build GRCh38
rs1033446042 Chr.12: 54582880-54582880 on Build GRCh38
rs1021845361 Chr.12: 54582920-54582920 on Build GRCh38
rs754153838 Chr.12: 54582830-54582866 on Build GRCh38
rs939979735 Chr.12: 54583056-54583056 on Build GRCh38
rs775079860 Chr.12: 54582698-54582698 on Build GRCh38
rs760547376 Chr.12: 54582824-54582824 on Build GRCh38
rs932668664 Chr.12: 54582818-54582818 on Build GRCh38
rs1036904101 Chr.12: 54582841-54582841 on Build GRCh38
rs771549937 Chr.12: 54582787-54582787 on Build GRCh38
rs976776806 Chr.12: 54583008-54583008 on Build GRCh38
rs962631922 Chr.12: 54583014-54583017 on Build GRCh38
rs928163661 Chr.12: 54582941-54582941 on Build GRCh38
rs1054845841 Chr.12: 54583089-54583089 on Build GRCh38
rs780189550 Chr.12: 54582761-54582761 on Build GRCh38
rs35146181 Chr.12: 54582888-54582888 on Build GRCh38
rs151136515 Chr.12: 54583057-54583057 on Build GRCh38
rs1031862860 Chr.12: 54582991-54582991 on Build GRCh38
rs140203195 Chr.12: 54583067-54583067 on Build GRCh38
rs756331474 Chr.12: 54582754-54582754 on Build GRCh38
rs1033372468 Chr.12: 54582669-54582669 on Build GRCh38
rs973742966 Chr.12: 54583018-54583021 on Build GRCh38
rs372773531 Chr.12: 54582773-54582773 on Build GRCh38
rs1052586261 Chr.12: 54583082-54583083 on Build GRCh38
rs954008938 Chr.12: 54583031-54583031 on Build GRCh38
rs992264206 Chr.12: 54582853-54582875 on Build GRCh38
rs976703205 Chr.12: 54582780-54582780 on Build GRCh38
rs915540951 Chr.12: 54583028-54583028 on Build GRCh38
rs757333307 Chr.12: 54582721-54582721 on Build GRCh38
rs200354799 Chr.12: 54582783-54582783 on Build GRCh38
rs984128001 Chr.12: 54583046-54583046 on Build GRCh38
rs928385330 Chr.12: 54583066-54583066 on Build GRCh38
rs921356942 Chr.12: 54583014-54583014 on Build GRCh38
rs71455285 Chr.12: 54582903-54582903 on Build GRCh38
rs191712993 Chr.12: 54583084-54583084 on Build GRCh38
rs781322013 Chr.12: 54582722-54582722 on Build GRCh38
rs532929450 Chr.12: 54582830-54582830 on Build GRCh38
rs969036145 Chr.12: 54582955-54582955 on Build GRCh38
rs749099536 Chr.12: 54582766-54582766 on Build GRCh38
rs915679730 Chr.12: 54582928-54582928 on Build GRCh38
rs150313461 Chr.12: 54583092-54583092 on Build GRCh38
rs1010887865 Chr.12: 54582846-54582846 on Build GRCh38
rs924687257 Chr.12: 54582767-54582767 on Build GRCh38
rs962717971 Chr.12: 54582901-54582901 on Build GRCh38
rs969256560 Chr.12: 54582756-54582756 on Build GRCh38
rs948416834 Chr.12: 54582929-54582929 on Build GRCh38
rs534606445 Chr.12: 54582859-54582859 on Build GRCh38
rs1043671606 Chr.12: 54582984-54582984 on Build GRCh38
rs569781355 Chr.12: 54582758-54582758 on Build GRCh38
rs11837630 Chr.12: 54582672-54582672 on Build GRCh38
rs1021897118 Chr.12: 54582752-54582752 on Build GRCh38
rs909784222 Chr.12: 54583047-54583047 on Build GRCh38
rs1003315632 Chr.12: 54582871-54582871 on Build GRCh38
rs772784390 Chr.12: 54582793-54582793 on Build GRCh38
rs375563735 Chr.12: 54583065-54583065 on Build GRCh38
rs1004164967 Chr.12: 54582819-54582819 on Build GRCh38
rs796775164 Chr.12: 54582971-54582971 on Build GRCh38
rs558619586 Chr.12: 54582823-54582823 on Build GRCh38
rs764208047 Chr.12: 54582828-54582829 on Build GRCh38
rs541980738 Chr.12: 54582916-54582916 on Build GRCh38
rs760531509 Chr.12: 54582829-54582829 on Build GRCh38
rs774193725 Chr.12: 54582774-54582774 on Build GRCh38
rs187207876 Chr.12: 54583054-54583054 on Build GRCh38
rs145020620 Chr.12: 54583036-54583037 on Build GRCh38
rs1045261922 Chr.12: 54582935-54582935 on Build GRCh38
rs576733948 Chr.12: 54582834-54582834 on Build GRCh38
rs1533235 Chr.12: 54582989-54582989 on Build GRCh38
rs752079087 Chr.12: 54582701-54582701 on Build GRCh38
rs753126626 Chr.12: 54582682-54582682 on Build GRCh38
rs527299071 Chr.12: 54582992-54582992 on Build GRCh38
rs760205257 Chr.12: 54582800-54582800 on Build GRCh38
rs548957076 Chr.12: 54583105-54583105 on Build GRCh38
rs764545266 Chr.12: 54582691-54582691 on Build GRCh38
rs566427290 Chr.12: 54582673-54582673 on Build GRCh38
rs536007751 Chr.12: 54582662-54582662 on Build GRCh38
rs553075188 Chr.12: 54582870-54582870 on Build GRCh38
rs758787858 Chr.12: 54582688-54582688 on Build GRCh38
rs372299483 Chr.12: 54583013-54583016 on Build GRCh38
rs368572328 Chr.12: 54583074-54583074 on Build GRCh38
rs1053139206 Chr.12: 54582970-54582970 on Build GRCh38
rs965948051 Chr.12: 54582996-54582996 on Build GRCh38
rs939179279 Chr.12: 54583083-54583083 on Build GRCh38

View Full Product Details