Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54856692-54857196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUCL1
Entrez Gene ID
118430
Gene Name
mucin like 1
Gene Aliases
SBEM
UniGene
Hs.348419
Gene Chromosome Location
Chr.12: 54854515-54858393 on Build GRCh38
Transcripts
NM_058173.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3462893 c.115G>A p.D39N Chr.12: 54856784-54856784 on Build GRCh38
468618 c.141T>A p.T47T Chr.12: 54856810-54856810 on Build GRCh38
1562016 c.118G>A p.E40K Chr.12: 54856787-54856787 on Build GRCh38
372715 c.188C>A p.T63N Chr.12: 54856857-54856857 on Build GRCh38
5051738 c.158C>T p.A53V Chr.12: 54856827-54856827 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542082932 Chr.12: 54857127-54857127 on Build GRCh38
rs367915417 Chr.12: 54856903-54856903 on Build GRCh38
rs995400940 Chr.12: 54857099-54857099 on Build GRCh38
rs540816405 Chr.12: 54856812-54856812 on Build GRCh38
rs369718217 Chr.12: 54856921-54856921 on Build GRCh38
rs762988048 Chr.12: 54856885-54856885 on Build GRCh38
rs200915552 Chr.12: 54856817-54856817 on Build GRCh38
rs768496159 Chr.12: 54856815-54856828 on Build GRCh38
rs572968352 Chr.12: 54856973-54856973 on Build GRCh38
rs971646771 Chr.12: 54857152-54857152 on Build GRCh38
rs991834316 Chr.12: 54857116-54857116 on Build GRCh38
rs1051488581 Chr.12: 54856963-54856963 on Build GRCh38
rs891813779 Chr.12: 54856948-54856948 on Build GRCh38
rs1051197169 Chr.12: 54856986-54856986 on Build GRCh38
rs147966123 Chr.12: 54856793-54856793 on Build GRCh38
rs534066612 Chr.12: 54856865-54856865 on Build GRCh38
rs745356490 Chr.12: 54856794-54856808 on Build GRCh38
rs550948813 Chr.12: 54856827-54856827 on Build GRCh38
rs748761979 Chr.12: 54856859-54856859 on Build GRCh38
rs201375025 Chr.12: 54856947-54856947 on Build GRCh38
rs149677659 Chr.12: 54856896-54856896 on Build GRCh38
rs868776896 Chr.12: 54857158-54857158 on Build GRCh38
rs867119928 Chr.12: 54856767-54856767 on Build GRCh38
rs757249683 Chr.12: 54856775-54856775 on Build GRCh38
rs1047949595 Chr.12: 54856731-54856731 on Build GRCh38
rs751395820 Chr.12: 54856841-54856841 on Build GRCh38
rs138424285 Chr.12: 54856844-54856844 on Build GRCh38
rs977751203 Chr.12: 54856779-54856793 on Build GRCh38
rs775954021 Chr.12: 54856914-54856914 on Build GRCh38
rs761062381 Chr.12: 54856842-54856865 on Build GRCh38
rs376466037 Chr.12: 54856888-54856888 on Build GRCh38
rs747013504 Chr.12: 54856813-54856813 on Build GRCh38
rs375520014 Chr.12: 54856931-54856931 on Build GRCh38
rs768458381 Chr.12: 54856861-54856861 on Build GRCh38
rs778680853 Chr.12: 54856869-54856869 on Build GRCh38
rs748956313 Chr.12: 54856785-54856785 on Build GRCh38
rs371508787 Chr.12: 54856879-54856879 on Build GRCh38
rs751988883 Chr.12: 54856836-54856852 on Build GRCh38
rs143525110 Chr.12: 54856874-54856874 on Build GRCh38
rs1031104406 Chr.12: 54856804-54856818 on Build GRCh38
rs763073933 Chr.12: 54856916-54856916 on Build GRCh38
rs758547450 Chr.12: 54856760-54856760 on Build GRCh38
rs1018073159 Chr.12: 54856997-54856997 on Build GRCh38
rs752715149 Chr.12: 54856822-54856822 on Build GRCh38
rs917531217 Chr.12: 54857138-54857138 on Build GRCh38
rs200116983 Chr.12: 54856878-54856878 on Build GRCh38
rs1004257441 Chr.12: 54857067-54857067 on Build GRCh38
rs148032746 Chr.12: 54856877-54856877 on Build GRCh38
rs761605995 Chr.12: 54856827-54856828 on Build GRCh38
rs756110708 Chr.12: 54856886-54856886 on Build GRCh38
rs556943211 Chr.12: 54856893-54856893 on Build GRCh38
rs767217981 Chr.12: 54856894-54856894 on Build GRCh38
rs761952536 Chr.12: 54856922-54856922 on Build GRCh38
rs201805259 Chr.12: 54856819-54856819 on Build GRCh38
rs141077987 Chr.12: 54857097-54857097 on Build GRCh38
rs747753345 Chr.12: 54856902-54856902 on Build GRCh38
rs776366438 Chr.12: 54856820-54856843 on Build GRCh38
rs1349910 Chr.12: 54856828-54856828 on Build GRCh38
rs745427604 Chr.12: 54856776-54856776 on Build GRCh38
rs759721973 Chr.12: 54856809-54856809 on Build GRCh38
rs1006725699 Chr.12: 54856988-54856988 on Build GRCh38
rs369923533 Chr.12: 54856943-54856943 on Build GRCh38
rs545800314 Chr.12: 54856790-54856790 on Build GRCh38
rs566608120 Chr.12: 54856858-54856858 on Build GRCh38
rs982740293 Chr.12: 54856748-54856748 on Build GRCh38
rs575091928 Chr.12: 54856722-54856722 on Build GRCh38
rs561770597 Chr.12: 54857070-54857070 on Build GRCh38
rs996841998 Chr.12: 54856870-54856870 on Build GRCh38
rs752903584 Chr.12: 54856757-54856757 on Build GRCh38
rs369648332 Chr.12: 54856751-54856751 on Build GRCh38
rs776982597 Chr.12: 54856807-54856807 on Build GRCh38
rs10783685 Chr.12: 54856765-54856765 on Build GRCh38
rs891081860 Chr.12: 54856989-54856989 on Build GRCh38
rs764260065 Chr.12: 54856832-54856832 on Build GRCh38
rs778457169 Chr.12: 54857166-54857166 on Build GRCh38
rs1010586458 Chr.12: 54856836-54856836 on Build GRCh38
rs199514917 Chr.12: 54856818-54856818 on Build GRCh38
rs767732528 Chr.12: 54856926-54856926 on Build GRCh38
rs146963141 Chr.12: 54857168-54857168 on Build GRCh38
rs745786226 Chr.12: 54856851-54856851 on Build GRCh38
rs371095531 Chr.12: 54856778-54856778 on Build GRCh38
rs917498621 Chr.12: 54856910-54856910 on Build GRCh38
rs145467450 Chr.12: 54856901-54856901 on Build GRCh38
rs746201787 Chr.12: 54856909-54856909 on Build GRCh38
rs780103508 Chr.12: 54856897-54856897 on Build GRCh38
rs10876639 Chr.12: 54856930-54856930 on Build GRCh38
rs751145875 Chr.12: 54857006-54857006 on Build GRCh38
rs766522580 Chr.12: 54856835-54856858 on Build GRCh38
rs375312325 Chr.12: 54856742-54856742 on Build GRCh38
rs753814277 Chr.12: 54856941-54856941 on Build GRCh38
rs528224202 Chr.12: 54856803-54856803 on Build GRCh38
rs895995064 Chr.12: 54856743-54856743 on Build GRCh38
rs1044583366 Chr.12: 54856929-54856929 on Build GRCh38
rs35119772 Chr.12: 54856910-54856911 on Build GRCh38
rs146901988 Chr.12: 54856792-54856792 on Build GRCh38
rs768713891 Chr.12: 54856919-54856919 on Build GRCh38
rs754982887 Chr.12: 54856744-54856744 on Build GRCh38
rs768368179 Chr.12: 54856787-54856787 on Build GRCh38
rs761731578 Chr.12: 54856889-54856889 on Build GRCh38
rs757008040 Chr.12: 54856953-54856953 on Build GRCh38
rs756777985 Chr.12: 54857033-54857033 on Build GRCh38
rs756009441 Chr.12: 54856853-54856853 on Build GRCh38
rs773483623 Chr.12: 54856828-54856829 on Build GRCh38
rs553609628 Chr.12: 54856806-54856806 on Build GRCh38
rs533144605 Chr.12: 54856843-54856843 on Build GRCh38
rs551778561 Chr.12: 54856852-54856852 on Build GRCh38
rs781431223 Chr.12: 54856905-54856905 on Build GRCh38
rs1025377118 Chr.12: 54857091-54857091 on Build GRCh38
rs149273418 Chr.12: 54856872-54856872 on Build GRCh38
rs771677113 Chr.12: 54856798-54856812 on Build GRCh38
rs568672064 Chr.12: 54856980-54856980 on Build GRCh38
rs769950148 Chr.12: 54856828-54856834 on Build GRCh38
rs775257595 Chr.12: 54856804-54856827 on Build GRCh38
rs775276102 Chr.12: 54856780-54856780 on Build GRCh38
rs1033018723 Chr.12: 54856811-54856811 on Build GRCh38
rs995237236 Chr.12: 54857073-54857073 on Build GRCh38
rs770324873 Chr.12: 54856911-54856911 on Build GRCh38

View Full Product Details