Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55820330-55820834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORMDL2
Entrez Gene ID
29095
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Gene Aliases
HSPC160, MST095, MSTP095, adoplin-2
UniGene
Hs.534450
Gene Chromosome Location
Chr.12: 55818022-55821175 on Build GRCh38
Transcripts
NM_014182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043376 c.414G>C p.K138N Chr.12: 55820347-55820347 on Build GRCh38
7337549 c.443T>C p.F148S Chr.12: 55820376-55820376 on Build GRCh38
694546 c.421C>G p.Q141E Chr.12: 55820354-55820354 on Build GRCh38
6351458 c.436C>T p.R146C Chr.12: 55820369-55820369 on Build GRCh38
4827966 c.411G>A p.P137P Chr.12: 55820344-55820344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375352682 Chr.12: 55820419-55820419 on Build GRCh38
rs185258417 Chr.12: 55820631-55820631 on Build GRCh38
rs145894501 Chr.12: 55820400-55820400 on Build GRCh38
rs1012925453 Chr.12: 55820711-55820711 on Build GRCh38
rs368255078 Chr.12: 55820396-55820396 on Build GRCh38
rs773947594 Chr.12: 55820443-55820443 on Build GRCh38
rs755516615 Chr.12: 55820397-55820397 on Build GRCh38
rs112289913 Chr.12: 55820639-55820639 on Build GRCh38
rs142915973 Chr.12: 55820807-55820807 on Build GRCh38
rs878928766 Chr.12: 55820697-55820697 on Build GRCh38
rs751772063 Chr.12: 55820482-55820482 on Build GRCh38
rs745722815 Chr.12: 55820424-55820424 on Build GRCh38
rs971380032 Chr.12: 55820745-55820745 on Build GRCh38
rs981547911 Chr.12: 55820747-55820747 on Build GRCh38
rs749783944 Chr.12: 55820571-55820571 on Build GRCh38
rs765822021 Chr.12: 55820467-55820467 on Build GRCh38
rs375046398 Chr.12: 55820490-55820490 on Build GRCh38
rs761470784 Chr.12: 55820448-55820448 on Build GRCh38
rs1018145347 Chr.12: 55820687-55820687 on Build GRCh38
rs771506381 Chr.12: 55820454-55820454 on Build GRCh38
rs763789329 Chr.12: 55820370-55820370 on Build GRCh38
rs14299 Chr.12: 55820665-55820665 on Build GRCh38
rs569990100 Chr.12: 55820489-55820489 on Build GRCh38
rs201312723 Chr.12: 55820356-55820356 on Build GRCh38
rs778180037 Chr.12: 55820415-55820415 on Build GRCh38
rs138499406 Chr.12: 55820798-55820798 on Build GRCh38
rs1050009288 Chr.12: 55820426-55820426 on Build GRCh38
rs118090673 Chr.12: 55820686-55820686 on Build GRCh38
rs977110644 Chr.12: 55820782-55820782 on Build GRCh38
rs781344544 Chr.12: 55820423-55820423 on Build GRCh38
rs1021227253 Chr.12: 55820740-55820740 on Build GRCh38
rs148412496 Chr.12: 55820394-55820394 on Build GRCh38
rs967034891 Chr.12: 55820764-55820764 on Build GRCh38
rs755423873 Chr.12: 55820644-55820644 on Build GRCh38
rs764575179 Chr.12: 55820480-55820480 on Build GRCh38
rs894722504 Chr.12: 55820511-55820511 on Build GRCh38
rs867185271 Chr.12: 55820655-55820655 on Build GRCh38
rs775355921 Chr.12: 55820359-55820359 on Build GRCh38
rs928381290 Chr.12: 55820806-55820806 on Build GRCh38
rs570613022 Chr.12: 55820758-55820758 on Build GRCh38
rs990052175 Chr.12: 55820703-55820703 on Build GRCh38
rs750572536 Chr.12: 55820493-55820493 on Build GRCh38
rs577712540 Chr.12: 55820701-55820702 on Build GRCh38
rs755979123 Chr.12: 55820499-55820499 on Build GRCh38
rs563977009 Chr.12: 55820542-55820542 on Build GRCh38
rs774644163 Chr.12: 55820372-55820372 on Build GRCh38
rs193220654 Chr.12: 55820369-55820369 on Build GRCh38
rs747265933 Chr.12: 55820417-55820417 on Build GRCh38
rs1030440710 Chr.12: 55820633-55820635 on Build GRCh38
rs942299535 Chr.12: 55820532-55820532 on Build GRCh38
rs111416072 Chr.12: 55820527-55820527 on Build GRCh38
rs779697000 Chr.12: 55820432-55820432 on Build GRCh38
rs765517220 Chr.12: 55820399-55820399 on Build GRCh38
rs992065619 Chr.12: 55820808-55820808 on Build GRCh38
rs113256470 Chr.12: 55820478-55820478 on Build GRCh38
rs769835196 Chr.12: 55820425-55820425 on Build GRCh38
rs749127859 Chr.12: 55820437-55820437 on Build GRCh38
rs768474178 Chr.12: 55820440-55820440 on Build GRCh38
rs760219483 Chr.12: 55820358-55820358 on Build GRCh38
rs757556143 Chr.12: 55820484-55820484 on Build GRCh38
rs946410068 Chr.12: 55820409-55820409 on Build GRCh38
rs1056017638 Chr.12: 55820461-55820461 on Build GRCh38
rs548581301 Chr.12: 55820462-55820462 on Build GRCh38
rs758671585 Chr.12: 55820403-55820403 on Build GRCh38
rs1023012375 Chr.12: 55820742-55820742 on Build GRCh38
rs752973841 Chr.12: 55820473-55820473 on Build GRCh38
rs761276920 Chr.12: 55820389-55820389 on Build GRCh38

View Full Product Details