Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55838516-55838953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP19
Entrez Gene ID
4327
Gene Name
matrix metallopeptidase 19
Gene Aliases
CODA, MMP18, RASI-1
UniGene
Hs.591033
Gene Chromosome Location
Chr.12: 55835430-55842983 on Build GRCh38
Transcripts
NM_001272101.1 NM_002429.5 NR_073606.1 XM_006719401.3 XM_011538359.1 XM_017019308.1 XM_017019309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391990 c.883G>A p.A295T Chr.12: 55838618-55838618 on Build GRCh38
941420 c.807G>T p.E269D Chr.12: 55838694-55838694 on Build GRCh38
3463276 c.796G>A p.E266K Chr.12: 55838705-55838705 on Build GRCh38
1935111 c.866G>A p.C289Y Chr.12: 55838635-55838635 on Build GRCh38
941421 c.776G>A p.S259N Chr.12: 55838725-55838725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752893251 Chr.12: 55838630-55838630 on Build GRCh38
rs763169803 Chr.12: 55838695-55838697 on Build GRCh38
rs747060381 Chr.12: 55838638-55838638 on Build GRCh38
rs567783555 Chr.12: 55838631-55838631 on Build GRCh38
rs777119142 Chr.12: 55838681-55838681 on Build GRCh38
rs191417431 Chr.12: 55838899-55838899 on Build GRCh38
rs751618957 Chr.12: 55838539-55838539 on Build GRCh38
rs182533371 Chr.12: 55838828-55838828 on Build GRCh38
rs768317442 Chr.12: 55838672-55838672 on Build GRCh38
rs757350617 Chr.12: 55838724-55838724 on Build GRCh38
rs760085316 Chr.12: 55838683-55838683 on Build GRCh38
rs112390036 Chr.12: 55838852-55838852 on Build GRCh38
rs372490290 Chr.12: 55838632-55838632 on Build GRCh38
rs749046078 Chr.12: 55838773-55838773 on Build GRCh38
rs772911437 Chr.12: 55838688-55838689 on Build GRCh38
rs138914662 Chr.12: 55838671-55838671 on Build GRCh38
rs754315733 Chr.12: 55838597-55838597 on Build GRCh38
rs761346983 Chr.12: 55838833-55838833 on Build GRCh38
rs556393129 Chr.12: 55838737-55838737 on Build GRCh38
rs902451177 Chr.12: 55838880-55838880 on Build GRCh38
rs769713187 Chr.12: 55838572-55838572 on Build GRCh38
rs767583975 Chr.12: 55838738-55838738 on Build GRCh38
rs11609152 Chr.12: 55838863-55838863 on Build GRCh38
rs949881595 Chr.12: 55838891-55838891 on Build GRCh38
rs775477507 Chr.12: 55838657-55838657 on Build GRCh38
rs773899473 Chr.12: 55838675-55838675 on Build GRCh38
rs751124452 Chr.12: 55838592-55838592 on Build GRCh38
rs915062819 Chr.12: 55838827-55838827 on Build GRCh38
rs753019666 Chr.12: 55838699-55838699 on Build GRCh38
rs185048424 Chr.12: 55838900-55838900 on Build GRCh38
rs376880823 Chr.12: 55838573-55838573 on Build GRCh38
rs758711137 Chr.12: 55838702-55838702 on Build GRCh38
rs368214782 Chr.12: 55838596-55838596 on Build GRCh38
rs765555025 Chr.12: 55838689-55838689 on Build GRCh38
rs745862921 Chr.12: 55838651-55838651 on Build GRCh38
rs770926355 Chr.12: 55838648-55838648 on Build GRCh38
rs549579023 Chr.12: 55838594-55838594 on Build GRCh38
rs368226088 Chr.12: 55838606-55838606 on Build GRCh38
rs776453257 Chr.12: 55838545-55838545 on Build GRCh38
rs766942080 Chr.12: 55838678-55838678 on Build GRCh38
rs777887188 Chr.12: 55838633-55838633 on Build GRCh38
rs763492965 Chr.12: 55838575-55838575 on Build GRCh38
rs780174995 Chr.12: 55838746-55838746 on Build GRCh38
rs996584586 Chr.12: 55838604-55838604 on Build GRCh38
rs1042690263 Chr.12: 55838875-55838875 on Build GRCh38
rs186948962 Chr.12: 55838890-55838890 on Build GRCh38
rs759343973 Chr.12: 55838561-55838561 on Build GRCh38
rs942777023 Chr.12: 55838814-55838814 on Build GRCh38
rs1026922948 Chr.12: 55838739-55838739 on Build GRCh38
rs769703002 Chr.12: 55838656-55838656 on Build GRCh38
rs761330779 Chr.12: 55838676-55838676 on Build GRCh38
rs768350860 Chr.12: 55838780-55838780 on Build GRCh38
rs373219794 Chr.12: 55838731-55838731 on Build GRCh38
rs751732515 Chr.12: 55838712-55838712 on Build GRCh38
rs959463989 Chr.12: 55838817-55838817 on Build GRCh38
rs902186430 Chr.12: 55838593-55838593 on Build GRCh38
rs779307393 Chr.12: 55838611-55838611 on Build GRCh38
rs764316231 Chr.12: 55838708-55838708 on Build GRCh38
rs1042017805 Chr.12: 55838576-55838576 on Build GRCh38
rs892916021 Chr.12: 55838591-55838591 on Build GRCh38
rs199781926 Chr.12: 55838685-55838685 on Build GRCh38
rs973401115 Chr.12: 55838693-55838696 on Build GRCh38
rs756164499 Chr.12: 55838745-55838745 on Build GRCh38

View Full Product Details