Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56202653-56203173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF41
Entrez Gene ID
10193
Gene Name
ring finger protein 41
Gene Aliases
FLRF, NRDP1, SBBI03
UniGene
Hs.524502
Gene Chromosome Location
Chr.12: 56202504-56221982 on Build GRCh38
Transcripts
NM_001242826.1 NM_005785.3 NM_194358.2 NM_194359.2 NR_040053.1 XM_005268561.4 XM_011537734.2 XM_011537735.2

SNP Detail

SNP ID Chromosome Location
rs529626265 Chr.12: 56203037-56203037 on Build GRCh38
rs1031053521 Chr.12: 56203123-56203123 on Build GRCh38
rs368093865 Chr.12: 56203075-56203076 on Build GRCh38
rs371588310 Chr.12: 56203061-56203062 on Build GRCh38
rs1002303682 Chr.12: 56203076-56203076 on Build GRCh38
rs1025563725 Chr.12: 56203102-56203102 on Build GRCh38
rs1008956501 Chr.12: 56203149-56203149 on Build GRCh38
rs558749751 Chr.12: 56202683-56202683 on Build GRCh38
rs1050007038 Chr.12: 56202951-56202951 on Build GRCh38
rs755628187 Chr.12: 56202871-56202874 on Build GRCh38
rs920953407 Chr.12: 56202932-56202932 on Build GRCh38
rs866656516 Chr.12: 56203040-56203040 on Build GRCh38
rs896910861 Chr.12: 56203024-56203024 on Build GRCh38
rs868845639 Chr.12: 56203097-56203097 on Build GRCh38
rs144419068 Chr.12: 56203039-56203039 on Build GRCh38
rs1012526466 Chr.12: 56203085-56203085 on Build GRCh38
rs943781395 Chr.12: 56202968-56202968 on Build GRCh38
rs989858293 Chr.12: 56203129-56203129 on Build GRCh38
rs895377001 Chr.12: 56203058-56203058 on Build GRCh38
rs199543319 Chr.12: 56203061-56203061 on Build GRCh38
rs1013617327 Chr.12: 56203065-56203066 on Build GRCh38
rs923931135 Chr.12: 56202750-56202750 on Build GRCh38
rs386376621 Chr.12: 56203068-56203069 on Build GRCh38
rs190633246 Chr.12: 56202782-56202782 on Build GRCh38
rs1036704963 Chr.12: 56203014-56203014 on Build GRCh38
rs869150544 Chr.12: 56203062-56203063 on Build GRCh38
rs568428027 Chr.12: 56202989-56202989 on Build GRCh38
rs970920949 Chr.12: 56203108-56203108 on Build GRCh38
rs978212056 Chr.12: 56202748-56202748 on Build GRCh38
rs1044647078 Chr.12: 56203111-56203111 on Build GRCh38
rs921978977 Chr.12: 56203135-56203135 on Build GRCh38
rs1025172399 Chr.12: 56202710-56202710 on Build GRCh38
rs975342941 Chr.12: 56202904-56202904 on Build GRCh38
rs989500502 Chr.12: 56202851-56202851 on Build GRCh38
rs934373328 Chr.12: 56203022-56203022 on Build GRCh38
rs386376622 Chr.12: 56203069-56203070 on Build GRCh38
rs912375248 Chr.12: 56202756-56202756 on Build GRCh38
rs771176660 Chr.12: 56203058-56203059 on Build GRCh38
rs914451234 Chr.12: 56202944-56202944 on Build GRCh38
rs148405751 Chr.12: 56203127-56203127 on Build GRCh38
rs965687145 Chr.12: 56202769-56202769 on Build GRCh38
rs779627739 Chr.12: 56203030-56203030 on Build GRCh38
rs950743664 Chr.12: 56202741-56202741 on Build GRCh38
rs978264374 Chr.12: 56203117-56203117 on Build GRCh38
rs932598601 Chr.12: 56202945-56202945 on Build GRCh38
rs947071327 Chr.12: 56202937-56202937 on Build GRCh38
rs890999337 Chr.12: 56202966-56202966 on Build GRCh38
rs528662754 Chr.12: 56202717-56202717 on Build GRCh38

View Full Product Details