Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56212655-56213155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF41
Entrez Gene ID
10193
Gene Name
ring finger protein 41
Gene Aliases
FLRF, NRDP1, SBBI03
UniGene
Hs.524502
Gene Chromosome Location
Chr.12: 56202504-56221982 on Build GRCh38
Transcripts
NM_001242826.1 NM_005785.3 NM_194358.2 NM_194359.2 NR_040053.1 XM_005268561.4 XM_011537734.2 XM_011537735.2

SNP Detail

SNP ID Chromosome Location
rs986274595 Chr.12: 56213071-56213071 on Build GRCh38
rs1005393626 Chr.12: 56212923-56212923 on Build GRCh38
rs574715937 Chr.12: 56212759-56212759 on Build GRCh38
rs926932127 Chr.12: 56212777-56212777 on Build GRCh38
rs116951219 Chr.12: 56212701-56212701 on Build GRCh38
rs760994851 Chr.12: 56212916-56212916 on Build GRCh38
rs946894256 Chr.12: 56212694-56212694 on Build GRCh38
rs116013211 Chr.12: 56212978-56212978 on Build GRCh38
rs887482367 Chr.12: 56212796-56212796 on Build GRCh38
rs775127855 Chr.12: 56212766-56212766 on Build GRCh38
rs981346588 Chr.12: 56213091-56213091 on Build GRCh38
rs956979699 Chr.12: 56213031-56213031 on Build GRCh38
rs888249572 Chr.12: 56212897-56212897 on Build GRCh38
rs926982770 Chr.12: 56213111-56213111 on Build GRCh38
rs781220038 Chr.12: 56213117-56213117 on Build GRCh38
rs910623940 Chr.12: 56213086-56213086 on Build GRCh38
rs971330973 Chr.12: 56213065-56213065 on Build GRCh38
rs572039979 Chr.12: 56212779-56212779 on Build GRCh38
rs1045449177 Chr.12: 56212770-56212770 on Build GRCh38
rs1048799816 Chr.12: 56212784-56212784 on Build GRCh38
rs556972678 Chr.12: 56212888-56212888 on Build GRCh38
rs1022505517 Chr.12: 56213059-56213059 on Build GRCh38
rs988317547 Chr.12: 56213058-56213058 on Build GRCh38
rs944676617 Chr.12: 56212811-56212811 on Build GRCh38
rs1003600738 Chr.12: 56212995-56212995 on Build GRCh38
rs755112781 Chr.12: 56213005-56213005 on Build GRCh38
rs766955876 Chr.12: 56212973-56212973 on Build GRCh38
rs748261575 Chr.12: 56212954-56212954 on Build GRCh38
rs545930565 Chr.12: 56213042-56213042 on Build GRCh38
rs757961206 Chr.12: 56212842-56212842 on Build GRCh38
rs541041359 Chr.12: 56212999-56212999 on Build GRCh38
rs367708207 Chr.12: 56212823-56212823 on Build GRCh38
rs575661609 Chr.12: 56213015-56213015 on Build GRCh38
rs889112652 Chr.12: 56212949-56212949 on Build GRCh38
rs1040405008 Chr.12: 56212816-56212816 on Build GRCh38
rs745782221 Chr.12: 56213124-56213124 on Build GRCh38
rs901067599 Chr.12: 56212841-56212841 on Build GRCh38
rs763721769 Chr.12: 56212809-56212809 on Build GRCh38
rs372605256 Chr.12: 56213112-56213112 on Build GRCh38
rs772210809 Chr.12: 56212960-56212960 on Build GRCh38
rs564297338 Chr.12: 56213061-56213061 on Build GRCh38
rs763857707 Chr.12: 56212865-56212865 on Build GRCh38
rs1023368585 Chr.12: 56212950-56212950 on Build GRCh38
rs1036908022 Chr.12: 56212822-56212822 on Build GRCh38
rs976486823 Chr.12: 56213118-56213118 on Build GRCh38

View Full Product Details