Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57268221-57268723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
UniGene
Hs.443673
Gene Chromosome Location
Chr.12: 57253764-57430999 on Build GRCh38
Transcripts
NM_014925.3 XM_005268725.3 XM_011538031.1 XM_011538033.1 XM_011538035.1 XM_011538036.1 XM_011538037.1 XM_011538038.1 XM_011538039.1 XM_011538040.1 XM_011538042.2 XM_017018998.1 XM_017018999.1 XM_017019000.1 XM_017019001.1 XM_017019002.1 XM_017019003.1 XM_017019004.1 XM_017019005.1 XM_017019006.1 XM_017019007.1 XM_017019008.1 XM_017019009.1 XM_017019010.1 XM_017019011.1 XM_017019012.1 XM_017019013.1 XM_017019014.1 XM_017019015.1 XM_017019016.1 XM_017019017.1 XM_017019018.1 XM_017019019.1 XM_017019020.1 XM_017019021.1 XM_017019022.1 XM_017019023.1 XM_017019024.1 XM_017019025.1 XM_017019026.1 XM_017019027.1 XM_017019028.1 XM_017019029.1 XM_017019030.1 XM_017019031.1 XM_017019032.1 XM_017019033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667188 c.1872G>A p.P624P Chr.12: 57268419-57268419 on Build GRCh38
4150270 c.927G>T p.V309V Chr.12: 57268449-57268449 on Build GRCh38
4150268 c.1842G>T p.V614V Chr.12: 57268449-57268449 on Build GRCh38
4863007 c.1032G>A p.V344V Chr.12: 57268344-57268344 on Build GRCh38
4922813 c.1927C>A p.P643T Chr.12: 57268364-57268364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769130638 Chr.12: 57268430-57268430 on Build GRCh38
rs751239330 Chr.12: 57268325-57268325 on Build GRCh38
rs766025432 Chr.12: 57268322-57268322 on Build GRCh38
rs756136071 Chr.12: 57268261-57268261 on Build GRCh38
rs138191775 Chr.12: 57268311-57268311 on Build GRCh38
rs775845484 Chr.12: 57268307-57268307 on Build GRCh38
rs772376473 Chr.12: 57268306-57268306 on Build GRCh38
rs200757143 Chr.12: 57268575-57268575 on Build GRCh38
rs772341293 Chr.12: 57268403-57268403 on Build GRCh38
rs750546000 Chr.12: 57268501-57268501 on Build GRCh38
rs995408062 Chr.12: 57268635-57268635 on Build GRCh38
rs761103373 Chr.12: 57268310-57268310 on Build GRCh38
rs200887467 Chr.12: 57268318-57268318 on Build GRCh38
rs746236523 Chr.12: 57268305-57268305 on Build GRCh38
rs567791198 Chr.12: 57268578-57268578 on Build GRCh38
rs977306174 Chr.12: 57268387-57268387 on Build GRCh38
rs777902951 Chr.12: 57268357-57268357 on Build GRCh38
rs201329941 Chr.12: 57268444-57268444 on Build GRCh38
rs757345533 Chr.12: 57268366-57268366 on Build GRCh38
rs767313292 Chr.12: 57268463-57268463 on Build GRCh38
rs746018461 Chr.12: 57268398-57268398 on Build GRCh38
rs369575037 Chr.12: 57268437-57268437 on Build GRCh38
rs369946630 Chr.12: 57268494-57268494 on Build GRCh38
rs765162678 Chr.12: 57268497-57268497 on Build GRCh38
rs899372723 Chr.12: 57268591-57268591 on Build GRCh38
rs752495194 Chr.12: 57268464-57268464 on Build GRCh38
rs764119250 Chr.12: 57268481-57268481 on Build GRCh38
rs759046209 Chr.12: 57268460-57268460 on Build GRCh38
rs756082640 Chr.12: 57268356-57268356 on Build GRCh38
rs747344000 Chr.12: 57268505-57268505 on Build GRCh38
rs752548419 Chr.12: 57268355-57268355 on Build GRCh38
rs767366036 Chr.12: 57268347-57268347 on Build GRCh38
rs748374044 Chr.12: 57268482-57268482 on Build GRCh38
rs968737593 Chr.12: 57268647-57268647 on Build GRCh38
rs745883639 Chr.12: 57268472-57268473 on Build GRCh38
rs796858570 Chr.12: 57268517-57268517 on Build GRCh38
rs924620709 Chr.12: 57268323-57268323 on Build GRCh38
rs890428511 Chr.12: 57268504-57268504 on Build GRCh38
rs749413459 Chr.12: 57268273-57268273 on Build GRCh38
rs747395490 Chr.12: 57268426-57268426 on Build GRCh38
rs777937633 Chr.12: 57268264-57268264 on Build GRCh38
rs753774791 Chr.12: 57268484-57268484 on Build GRCh38
rs898955743 Chr.12: 57268472-57268472 on Build GRCh38
rs552336784 Chr.12: 57268594-57268594 on Build GRCh38
rs773928496 Chr.12: 57268445-57268445 on Build GRCh38
rs935815164 Chr.12: 57268420-57268420 on Build GRCh38
rs149171486 Chr.12: 57268419-57268419 on Build GRCh38
rs967305745 Chr.12: 57268363-57268363 on Build GRCh38
rs759365046 Chr.12: 57268341-57268341 on Build GRCh38
rs192241548 Chr.12: 57268248-57268248 on Build GRCh38
rs375131695 Chr.12: 57268379-57268379 on Build GRCh38
rs772479192 Chr.12: 57268696-57268696 on Build GRCh38
rs978827316 Chr.12: 57268694-57268694 on Build GRCh38
rs372109139 Chr.12: 57268297-57268297 on Build GRCh38
rs1019308134 Chr.12: 57268532-57268532 on Build GRCh38
rs940976129 Chr.12: 57268539-57268539 on Build GRCh38
rs748157233 Chr.12: 57268253-57268253 on Build GRCh38
rs1022085290 Chr.12: 57268606-57268606 on Build GRCh38
rs1057509622 Chr.12: 57268358-57268358 on Build GRCh38
rs373399007 Chr.12: 57268503-57268503 on Build GRCh38
rs771159605 Chr.12: 57268284-57268284 on Build GRCh38
rs1047396155 Chr.12: 57268486-57268486 on Build GRCh38
rs375121494 Chr.12: 57268470-57268470 on Build GRCh38
rs894219806 Chr.12: 57268647-57268647 on Build GRCh38
rs964568115 Chr.12: 57268586-57268586 on Build GRCh38
rs1034491319 Chr.12: 57268642-57268642 on Build GRCh38
rs996448573 Chr.12: 57268600-57268600 on Build GRCh38

View Full Product Details