Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 65453587-65453994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSRB3
Entrez Gene ID
253827
Gene Name
methionine sulfoxide reductase B3
Gene Aliases
DFNB74
UniGene
Hs.339024
Gene Chromosome Location
Chr.12: 65278643-65466907 on Build GRCh38
Transcripts
NM_001031679.2 NM_001193460.1 NM_001193461.1 NM_198080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6715969 c.310G>A p.D104N Chr.12: 65453745-65453745 on Build GRCh38
3872269 c.307C>T p.H103Y Chr.12: 65453742-65453742 on Build GRCh38
3384565 c.411+1G>A p.? Chr.12: 65453826-65453826 on Build GRCh38
4895865 c.372G>A p.V124V Chr.12: 65453807-65453807 on Build GRCh38
4895864 c.393G>A p.V131V Chr.12: 65453807-65453807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748329626 Chr.12: 65453693-65453693 on Build GRCh38
rs749763900 Chr.12: 65453814-65453814 on Build GRCh38
rs752925444 Chr.12: 65453900-65453900 on Build GRCh38
rs142397042 Chr.12: 65453619-65453619 on Build GRCh38
rs369942269 Chr.12: 65453791-65453791 on Build GRCh38
rs781656489 Chr.12: 65453916-65453918 on Build GRCh38
rs764211921 Chr.12: 65453905-65453905 on Build GRCh38
rs759271823 Chr.12: 65453885-65453885 on Build GRCh38
rs867671100 Chr.12: 65453788-65453788 on Build GRCh38
rs768179073 Chr.12: 65453840-65453840 on Build GRCh38
rs954314187 Chr.12: 65453899-65453899 on Build GRCh38
rs762906000 Chr.12: 65453764-65453764 on Build GRCh38
rs727503162 Chr.12: 65453745-65453745 on Build GRCh38
rs1003026750 Chr.12: 65453912-65453912 on Build GRCh38
rs75990461 Chr.12: 65453722-65453722 on Build GRCh38
rs372265552 Chr.12: 65453689-65453689 on Build GRCh38
rs989612418 Chr.12: 65453621-65453621 on Build GRCh38
rs968421928 Chr.12: 65453924-65453924 on Build GRCh38
rs1035887519 Chr.12: 65453915-65453915 on Build GRCh38
rs747526542 Chr.12: 65453819-65453820 on Build GRCh38
rs774879831 Chr.12: 65453726-65453726 on Build GRCh38
rs140600679 Chr.12: 65453744-65453744 on Build GRCh38
rs752879693 Chr.12: 65453797-65453797 on Build GRCh38
rs758721155 Chr.12: 65453921-65453921 on Build GRCh38
rs34513088 Chr.12: 65453792-65453792 on Build GRCh38
rs748702188 Chr.12: 65453968-65453968 on Build GRCh38
rs770192332 Chr.12: 65453698-65453698 on Build GRCh38
rs769304971 Chr.12: 65453851-65453851 on Build GRCh38
rs781105466 Chr.12: 65453691-65453691 on Build GRCh38
rs769220592 Chr.12: 65453912-65453912 on Build GRCh38
rs760382678 Chr.12: 65453738-65453738 on Build GRCh38
rs772615672 Chr.12: 65453855-65453855 on Build GRCh38
rs549371383 Chr.12: 65453754-65453754 on Build GRCh38
rs934295841 Chr.12: 65453839-65453839 on Build GRCh38
rs756314771 Chr.12: 65453903-65453903 on Build GRCh38
rs970680024 Chr.12: 65453624-65453624 on Build GRCh38
rs574394183 Chr.12: 65453965-65453965 on Build GRCh38
rs34177392 Chr.12: 65453748-65453748 on Build GRCh38
rs201206970 Chr.12: 65453828-65453828 on Build GRCh38
rs767224078 Chr.12: 65453891-65453891 on Build GRCh38
rs535883095 Chr.12: 65453723-65453723 on Build GRCh38
rs750305947 Chr.12: 65453762-65453762 on Build GRCh38
rs372753747 Chr.12: 65453927-65453927 on Build GRCh38
rs981660402 Chr.12: 65453657-65453657 on Build GRCh38
rs983441102 Chr.12: 65453714-65453714 on Build GRCh38
rs767841214 Chr.12: 65453793-65453793 on Build GRCh38
rs372222625 Chr.12: 65453777-65453777 on Build GRCh38
rs914301678 Chr.12: 65453643-65453643 on Build GRCh38
rs267603629 Chr.12: 65453750-65453750 on Build GRCh38
rs762850720 Chr.12: 65453865-65453865 on Build GRCh38
rs774122886 Chr.12: 65453884-65453884 on Build GRCh38
rs1026833082 Chr.12: 65453807-65453807 on Build GRCh38
rs558604792 Chr.12: 65453878-65453878 on Build GRCh38
rs778287765 Chr.12: 65453711-65453711 on Build GRCh38
rs535395278 Chr.12: 65453906-65453906 on Build GRCh38
rs779389792 Chr.12: 65453833-65453833 on Build GRCh38
rs578131252 Chr.12: 65453918-65453918 on Build GRCh38
rs374937043 Chr.12: 65453703-65453703 on Build GRCh38
rs1015590576 Chr.12: 65453852-65453852 on Build GRCh38
rs757834054 Chr.12: 65453916-65453916 on Build GRCh38
rs755211448 Chr.12: 65453677-65453677 on Build GRCh38
rs538374082 Chr.12: 65453843-65453843 on Build GRCh38
rs965792888 Chr.12: 65453856-65453856 on Build GRCh38
rs748859293 Chr.12: 65453938-65453938 on Build GRCh38
rs746301312 Chr.12: 65453834-65453834 on Build GRCh38
rs770443598 Chr.12: 65453947-65453947 on Build GRCh38
rs766373899 Chr.12: 65453875-65453875 on Build GRCh38
rs888517603 Chr.12: 65453837-65453837 on Build GRCh38
rs974874091 Chr.12: 65453964-65453964 on Build GRCh38
rs777748623 Chr.12: 65453804-65453804 on Build GRCh38
rs769377110 Chr.12: 65453937-65453937 on Build GRCh38
rs371970813 Chr.12: 65453803-65453803 on Build GRCh38
rs909286910 Chr.12: 65453719-65453719 on Build GRCh38
rs747757508 Chr.12: 65453848-65453848 on Build GRCh38
rs1016276177 Chr.12: 65453955-65453955 on Build GRCh38
rs775999995 Chr.12: 65453752-65453752 on Build GRCh38
rs745714349 Chr.12: 65453760-65453760 on Build GRCh38
rs554547010 Chr.12: 65453956-65453956 on Build GRCh38
rs80349215 Chr.12: 65453926-65453926 on Build GRCh38
rs1028956043 Chr.12: 65453894-65453894 on Build GRCh38

View Full Product Details