Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68201607-68202097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL26
Entrez Gene ID
55801
Gene Name
interleukin 26
Gene Aliases
AK155, IL-26
UniGene
Hs.272350
Gene Chromosome Location
Chr.12: 68201349-68225791 on Build GRCh38
Transcripts
NM_018402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942851 c.388G>T p.E130* Chr.12: 68202059-68202059 on Build GRCh38
1363843 c.502G>A p.E168K Chr.12: 68201859-68201859 on Build GRCh38
7247092 c.430-8A>G p.? Chr.12: 68201939-68201939 on Build GRCh38
1323100 c.432T>G p.I144M Chr.12: 68201929-68201929 on Build GRCh38
4044411 c.429+2T>C p.? Chr.12: 68202016-68202016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761354103 Chr.12: 68201904-68201904 on Build GRCh38
rs779937576 Chr.12: 68201930-68201930 on Build GRCh38
rs1046885756 Chr.12: 68201837-68201837 on Build GRCh38
rs774811574 Chr.12: 68201981-68201981 on Build GRCh38
rs868267024 Chr.12: 68201902-68201902 on Build GRCh38
rs10748101 Chr.12: 68202007-68202007 on Build GRCh38
rs766176256 Chr.12: 68202001-68202001 on Build GRCh38
rs745674864 Chr.12: 68201809-68201809 on Build GRCh38
rs749867315 Chr.12: 68201998-68201998 on Build GRCh38
rs369057704 Chr.12: 68201934-68201934 on Build GRCh38
rs749289218 Chr.12: 68202056-68202056 on Build GRCh38
rs1028425740 Chr.12: 68201958-68201958 on Build GRCh38
rs773640932 Chr.12: 68201854-68201854 on Build GRCh38
rs199844626 Chr.12: 68201943-68201943 on Build GRCh38
rs925243680 Chr.12: 68201768-68201768 on Build GRCh38
rs755646348 Chr.12: 68201881-68201883 on Build GRCh38
rs779696892 Chr.12: 68201814-68201814 on Build GRCh38
rs747301877 Chr.12: 68201817-68201817 on Build GRCh38
rs755213718 Chr.12: 68201938-68201938 on Build GRCh38
rs769928311 Chr.12: 68201944-68201944 on Build GRCh38
rs541483348 Chr.12: 68201700-68201700 on Build GRCh38
rs763803766 Chr.12: 68201875-68201875 on Build GRCh38
rs79293173 Chr.12: 68201686-68201686 on Build GRCh38
rs770298339 Chr.12: 68201853-68201853 on Build GRCh38
rs765274126 Chr.12: 68201906-68201906 on Build GRCh38
rs79050996 Chr.12: 68201656-68201656 on Build GRCh38
rs761540272 Chr.12: 68201993-68201993 on Build GRCh38
rs768439665 Chr.12: 68201988-68201988 on Build GRCh38
rs904685132 Chr.12: 68201730-68201730 on Build GRCh38
rs753369298 Chr.12: 68201794-68201794 on Build GRCh38
rs76585150 Chr.12: 68201658-68201658 on Build GRCh38
rs367724685 Chr.12: 68202043-68202043 on Build GRCh38
rs913750936 Chr.12: 68201729-68201729 on Build GRCh38
rs751270003 Chr.12: 68201935-68201935 on Build GRCh38
rs759773515 Chr.12: 68201982-68201982 on Build GRCh38
rs750428765 Chr.12: 68201907-68201907 on Build GRCh38
rs774558724 Chr.12: 68201863-68201863 on Build GRCh38
rs777817616 Chr.12: 68201952-68201952 on Build GRCh38
rs372744599 Chr.12: 68202058-68202058 on Build GRCh38
rs760417109 Chr.12: 68201864-68201864 on Build GRCh38
rs764885768 Chr.12: 68201996-68201996 on Build GRCh38
rs543708508 Chr.12: 68201862-68201862 on Build GRCh38
rs79462800 Chr.12: 68201659-68201659 on Build GRCh38
rs780961280 Chr.12: 68202027-68202027 on Build GRCh38
rs1043606216 Chr.12: 68201759-68201759 on Build GRCh38
rs1020685616 Chr.12: 68202051-68202051 on Build GRCh38
rs771312655 Chr.12: 68201968-68201968 on Build GRCh38
rs776721978 Chr.12: 68201848-68201848 on Build GRCh38
rs762976376 Chr.12: 68201999-68201999 on Build GRCh38
rs576296427 Chr.12: 68201769-68201769 on Build GRCh38
rs376101183 Chr.12: 68201811-68201811 on Build GRCh38
rs757294514 Chr.12: 68201799-68201799 on Build GRCh38
rs1016521402 Chr.12: 68201861-68201861 on Build GRCh38
rs376659677 Chr.12: 68202064-68202064 on Build GRCh38
rs748174615 Chr.12: 68201849-68201849 on Build GRCh38
rs768726824 Chr.12: 68201838-68201838 on Build GRCh38
rs565600537 Chr.12: 68201685-68201685 on Build GRCh38
rs1007300478 Chr.12: 68201925-68201925 on Build GRCh38
rs368646100 Chr.12: 68202050-68202050 on Build GRCh38
rs778948435 Chr.12: 68201801-68201801 on Build GRCh38
rs1055545283 Chr.12: 68201792-68201792 on Build GRCh38
rs777553129 Chr.12: 68201852-68201852 on Build GRCh38
rs758241511 Chr.12: 68201912-68201912 on Build GRCh38
rs34231221 Chr.12: 68201774-68201775 on Build GRCh38
rs1047859848 Chr.12: 68201842-68201842 on Build GRCh38
rs754701417 Chr.12: 68202020-68202020 on Build GRCh38
rs963181918 Chr.12: 68201987-68201988 on Build GRCh38
rs945240543 Chr.12: 68201734-68201734 on Build GRCh38
rs753424343 Chr.12: 68201893-68201893 on Build GRCh38
rs867606937 Chr.12: 68201843-68201843 on Build GRCh38
rs948812333 Chr.12: 68201708-68201708 on Build GRCh38
rs777950508 Chr.12: 68202052-68202052 on Build GRCh38
rs1009662510 Chr.12: 68201818-68201818 on Build GRCh38
rs10748100 Chr.12: 68201939-68201939 on Build GRCh38

View Full Product Details