Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69585592-69586085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
UniGene
Hs.189772
Gene Chromosome Location
Chr.12: 69585428-69601577 on Build GRCh38
Transcripts
NM_001198842.1 NM_006431.2

SNP Detail

SNP ID Chromosome Location
rs980229496 Chr.12: 69585712-69585712 on Build GRCh38
rs564805236 Chr.12: 69585708-69585708 on Build GRCh38
rs976937475 Chr.12: 69586018-69586018 on Build GRCh38
rs1019044087 Chr.12: 69585872-69585872 on Build GRCh38
rs368802453 Chr.12: 69585701-69585701 on Build GRCh38
rs1011441615 Chr.12: 69585691-69585691 on Build GRCh38
rs1001844365 Chr.12: 69585816-69585816 on Build GRCh38
rs1045536403 Chr.12: 69586023-69586023 on Build GRCh38
rs531195601 Chr.12: 69585687-69585687 on Build GRCh38
rs1021034368 Chr.12: 69585625-69585625 on Build GRCh38
rs1052421471 Chr.12: 69585823-69585823 on Build GRCh38
rs966843971 Chr.12: 69585675-69585675 on Build GRCh38
rs905707679 Chr.12: 69585795-69585795 on Build GRCh38
rs1035443800 Chr.12: 69585853-69585853 on Build GRCh38
rs149925318 Chr.12: 69586052-69586052 on Build GRCh38
rs140997100 Chr.12: 69585745-69585745 on Build GRCh38
rs999558723 Chr.12: 69585821-69585821 on Build GRCh38
rs1031631632 Chr.12: 69585680-69585680 on Build GRCh38
rs949783535 Chr.12: 69585787-69585787 on Build GRCh38
rs111702592 Chr.12: 69585740-69585740 on Build GRCh38
rs931555069 Chr.12: 69585743-69585743 on Build GRCh38
rs865844206 Chr.12: 69585820-69585820 on Build GRCh38
rs889909101 Chr.12: 69585640-69585640 on Build GRCh38
rs1011885097 Chr.12: 69585835-69585835 on Build GRCh38
rs374499372 Chr.12: 69585778-69585778 on Build GRCh38
rs747677153 Chr.12: 69586013-69586013 on Build GRCh38
rs556425259 Chr.12: 69585789-69585789 on Build GRCh38
rs959774536 Chr.12: 69585863-69585863 on Build GRCh38
rs975037823 Chr.12: 69585731-69585731 on Build GRCh38
rs779045190 Chr.12: 69586026-69586026 on Build GRCh38
rs200272078 Chr.12: 69586051-69586051 on Build GRCh38
rs780631202 Chr.12: 69585994-69585994 on Build GRCh38
rs574602706 Chr.12: 69585832-69585832 on Build GRCh38
rs959661046 Chr.12: 69585735-69585735 on Build GRCh38
rs901146926 Chr.12: 69585651-69585651 on Build GRCh38
rs868117636 Chr.12: 69586004-69586004 on Build GRCh38
rs983075169 Chr.12: 69585952-69585952 on Build GRCh38
rs911404449 Chr.12: 69585999-69585999 on Build GRCh38
rs547602714 Chr.12: 69585783-69585783 on Build GRCh38
rs987535935 Chr.12: 69585870-69585870 on Build GRCh38
rs74749076 Chr.12: 69585721-69585721 on Build GRCh38
rs1045565306 Chr.12: 69585792-69585792 on Build GRCh38
rs975395856 Chr.12: 69585922-69585922 on Build GRCh38
rs1001295115 Chr.12: 69585843-69585843 on Build GRCh38
rs1054773430 Chr.12: 69586044-69586044 on Build GRCh38
rs937229788 Chr.12: 69586043-69586043 on Build GRCh38
rs970260674 Chr.12: 69585842-69585842 on Build GRCh38
rs542025145 Chr.12: 69585893-69585893 on Build GRCh38
rs527461952 Chr.12: 69585931-69585931 on Build GRCh38
rs758219946 Chr.12: 69585700-69585700 on Build GRCh38
rs558264488 Chr.12: 69585747-69585747 on Build GRCh38
rs955930696 Chr.12: 69585685-69585685 on Build GRCh38
rs907557910 Chr.12: 69585988-69585988 on Build GRCh38
rs2601006 Chr.12: 69585737-69585737 on Build GRCh38
rs184089227 Chr.12: 69586033-69586033 on Build GRCh38
rs920886891 Chr.12: 69585930-69585930 on Build GRCh38
rs997435319 Chr.12: 69585676-69585676 on Build GRCh38
rs576546053 Chr.12: 69585764-69585764 on Build GRCh38
rs952986422 Chr.12: 69585975-69585975 on Build GRCh38
rs1042348340 Chr.12: 69586061-69586061 on Build GRCh38
rs972402130 Chr.12: 69585894-69585894 on Build GRCh38
rs1049031617 Chr.12: 69585623-69585623 on Build GRCh38
rs755176707 Chr.12: 69585793-69585793 on Build GRCh38
rs1021470664 Chr.12: 69585837-69585837 on Build GRCh38
rs367719211 Chr.12: 69586024-69586024 on Build GRCh38
rs377616113 Chr.12: 69585612-69585612 on Build GRCh38
rs369443707 Chr.12: 69585759-69585759 on Build GRCh38
rs901253424 Chr.12: 69585791-69585791 on Build GRCh38
rs1007369629 Chr.12: 69585641-69585641 on Build GRCh38
rs550104627 Chr.12: 69585698-69585698 on Build GRCh38
rs769293794 Chr.12: 69586056-69586056 on Build GRCh38
rs949753560 Chr.12: 69586021-69586021 on Build GRCh38
rs1041071226 Chr.12: 69585648-69585648 on Build GRCh38
rs560062903 Chr.12: 69585909-69585909 on Build GRCh38
rs911358212 Chr.12: 69585761-69585761 on Build GRCh38
rs1010573214 Chr.12: 69586054-69586054 on Build GRCh38

View Full Product Details