Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88505152-88505690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KITLG
Entrez Gene ID
4254
Gene Name
KIT ligand
Gene Aliases
DCUA, DFNA69, FPH2, FPHH, KL-1, Kitl, MGF, SCF, SF, SHEP7
UniGene
Hs.1048
Gene Chromosome Location
Chr.12: 88492793-88580473 on Build GRCh38
Transcripts
NM_000899.4 NM_003994.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5376170 c.819G>A p.V273V Chr.12: 88505199-88505199 on Build GRCh38
4404005 c.797A>C p.K266T Chr.12: 88505221-88505221 on Build GRCh38
4450726 c.806_807delAG p.E269fs*>5 Chr.12: 88505211-88505212 on Build GRCh38
1706008 c.803G>A p.R268K Chr.12: 88505215-88505215 on Build GRCh38
3813157 c.793G>C p.E265Q Chr.12: 88505225-88505225 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781400280 Chr.12: 88505186-88505186 on Build GRCh38
rs41306946 Chr.12: 88505368-88505368 on Build GRCh38
rs983223642 Chr.12: 88505424-88505424 on Build GRCh38
rs573103721 Chr.12: 88505371-88505371 on Build GRCh38
rs1039799333 Chr.12: 88505440-88505440 on Build GRCh38
rs745761824 Chr.12: 88505211-88505211 on Build GRCh38
rs1047469343 Chr.12: 88505356-88505356 on Build GRCh38
rs916774522 Chr.12: 88505293-88505293 on Build GRCh38
rs900361824 Chr.12: 88505455-88505455 on Build GRCh38
rs759455178 Chr.12: 88505467-88505467 on Build GRCh38
rs757048774 Chr.12: 88505626-88505626 on Build GRCh38
rs960388439 Chr.12: 88505547-88505547 on Build GRCh38
rs765765692 Chr.12: 88505498-88505498 on Build GRCh38
rs41311947 Chr.12: 88505505-88505505 on Build GRCh38
rs541713443 Chr.12: 88505384-88505384 on Build GRCh38
rs146721596 Chr.12: 88505450-88505450 on Build GRCh38
rs570144676 Chr.12: 88505343-88505343 on Build GRCh38
rs1008419757 Chr.12: 88505519-88505519 on Build GRCh38
rs372927175 Chr.12: 88505250-88505250 on Build GRCh38
rs751697412 Chr.12: 88505322-88505322 on Build GRCh38
rs764155170 Chr.12: 88505250-88505251 on Build GRCh38
rs978126507 Chr.12: 88505259-88505259 on Build GRCh38
rs996008371 Chr.12: 88505466-88505466 on Build GRCh38
rs537168910 Chr.12: 88505296-88505296 on Build GRCh38
rs891231394 Chr.12: 88505405-88505405 on Build GRCh38
rs187212128 Chr.12: 88505655-88505655 on Build GRCh38
rs182136643 Chr.12: 88505525-88505525 on Build GRCh38
rs144290808 Chr.12: 88505535-88505535 on Build GRCh38
rs769753985 Chr.12: 88505195-88505195 on Build GRCh38
rs140356627 Chr.12: 88505503-88505503 on Build GRCh38
rs778763249 Chr.12: 88505633-88505633 on Build GRCh38
rs143503044 Chr.12: 88505417-88505417 on Build GRCh38
rs1049845821 Chr.12: 88505507-88505507 on Build GRCh38
rs552892091 Chr.12: 88505362-88505362 on Build GRCh38
rs544751210 Chr.12: 88505469-88505469 on Build GRCh38
rs1012284847 Chr.12: 88505653-88505653 on Build GRCh38
rs755130353 Chr.12: 88505409-88505409 on Build GRCh38
rs368648104 Chr.12: 88505248-88505248 on Build GRCh38
rs776990784 Chr.12: 88505273-88505273 on Build GRCh38
rs970535611 Chr.12: 88505406-88505406 on Build GRCh38
rs41306948 Chr.12: 88505302-88505302 on Build GRCh38
rs375388664 Chr.12: 88505246-88505246 on Build GRCh38
rs748472602 Chr.12: 88505187-88505187 on Build GRCh38
rs917621486 Chr.12: 88505372-88505372 on Build GRCh38
rs763325447 Chr.12: 88505285-88505285 on Build GRCh38
rs41311949 Chr.12: 88505395-88505395 on Build GRCh38
rs572328763 Chr.12: 88505274-88505274 on Build GRCh38
rs907807897 Chr.12: 88505427-88505427 on Build GRCh38
rs891164988 Chr.12: 88505514-88505514 on Build GRCh38
rs549562382 Chr.12: 88505267-88505267 on Build GRCh38
rs991892357 Chr.12: 88505607-88505607 on Build GRCh38
rs777958525 Chr.12: 88505199-88505199 on Build GRCh38
rs941980246 Chr.12: 88505446-88505446 on Build GRCh38
rs902671730 Chr.12: 88505548-88505548 on Build GRCh38
rs760961616 Chr.12: 88505211-88505214 on Build GRCh38
rs41306431 Chr.12: 88505478-88505478 on Build GRCh38
rs574549183 Chr.12: 88505577-88505578 on Build GRCh38
rs377048625 Chr.12: 88505276-88505276 on Build GRCh38
rs974334028 Chr.12: 88505320-88505320 on Build GRCh38
rs959281672 Chr.12: 88505353-88505353 on Build GRCh38
rs539046189 Chr.12: 88505360-88505360 on Build GRCh38
rs192267590 Chr.12: 88505303-88505303 on Build GRCh38
rs754001431 Chr.12: 88505287-88505287 on Build GRCh38

View Full Product Details