Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89472137-89472683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POC1B
Entrez Gene ID
282809
Gene Name
POC1 centriolar protein B
Gene Aliases
CORD20, PIX1, TUWD12, WDR51B
UniGene
Hs.25130
Gene Chromosome Location
Chr.12: 89419718-89526262 on Build GRCh38
Transcripts
NM_001199777.1 NM_172240.2 NR_037659.1 NR_037660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4614440 c.527_528insA p.Q177fs*4 Chr.12: 89472200-89472201 on Build GRCh38
348787 c.513G>T p.W171C Chr.12: 89472215-89472215 on Build GRCh38
6751611 c.517A>G p.T173A Chr.12: 89472211-89472211 on Build GRCh38
1606735 c.453-12_453-3delTCTTCTTTCT p.? Chr.12: 89472278-89472287 on Build GRCh38
3738585 c.546C>G p.F182L Chr.12: 89472182-89472182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006661003 Chr.12: 89472425-89472425 on Build GRCh38
rs147832668 Chr.12: 89472286-89472286 on Build GRCh38
rs71082492 Chr.12: 89472289-89472297 on Build GRCh38
rs1018297721 Chr.12: 89472598-89472598 on Build GRCh38
rs993586249 Chr.12: 89472458-89472458 on Build GRCh38
rs767117053 Chr.12: 89472284-89472290 on Build GRCh38
rs867057656 Chr.12: 89472241-89472241 on Build GRCh38
rs151210595 Chr.12: 89472266-89472266 on Build GRCh38
rs528009352 Chr.12: 89472446-89472446 on Build GRCh38
rs186489432 Chr.12: 89472646-89472646 on Build GRCh38
rs74364013 Chr.12: 89472288-89472288 on Build GRCh38
rs760262938 Chr.12: 89472172-89472172 on Build GRCh38
rs1016262543 Chr.12: 89472444-89472444 on Build GRCh38
rs981838850 Chr.12: 89472654-89472654 on Build GRCh38
rs1040134078 Chr.12: 89472433-89472433 on Build GRCh38
rs868139654 Chr.12: 89472474-89472474 on Build GRCh38
rs759071227 Chr.12: 89472280-89472282 on Build GRCh38
rs750943166 Chr.12: 89472257-89472257 on Build GRCh38
rs775164968 Chr.12: 89472169-89472169 on Build GRCh38
rs59139895 Chr.12: 89472278-89472287 on Build GRCh38
rs937829957 Chr.12: 89472380-89472380 on Build GRCh38
rs1046092649 Chr.12: 89472341-89472341 on Build GRCh38
rs763347139 Chr.12: 89472264-89472264 on Build GRCh38
rs781186534 Chr.12: 89472305-89472305 on Build GRCh38
rs1030504560 Chr.12: 89472168-89472168 on Build GRCh38
rs765823697 Chr.12: 89472253-89472253 on Build GRCh38
rs368821348 Chr.12: 89472234-89472234 on Build GRCh38
rs986798731 Chr.12: 89472175-89472175 on Build GRCh38
rs1009392040 Chr.12: 89472594-89472594 on Build GRCh38
rs888350430 Chr.12: 89472415-89472415 on Build GRCh38
rs768223224 Chr.12: 89472173-89472173 on Build GRCh38
rs147021755 Chr.12: 89472287-89472287 on Build GRCh38
rs565445962 Chr.12: 89472361-89472361 on Build GRCh38
rs998522621 Chr.12: 89472456-89472456 on Build GRCh38
rs986293044 Chr.12: 89472500-89472500 on Build GRCh38
rs199977729 Chr.12: 89472268-89472268 on Build GRCh38
rs910777225 Chr.12: 89472205-89472205 on Build GRCh38
rs548047679 Chr.12: 89472483-89472483 on Build GRCh38
rs141027674 Chr.12: 89472652-89472652 on Build GRCh38
rs913572652 Chr.12: 89472649-89472649 on Build GRCh38
rs771799275 Chr.12: 89472163-89472163 on Build GRCh38
rs930007995 Chr.12: 89472291-89472293 on Build GRCh38
rs755173263 Chr.12: 89472275-89472275 on Build GRCh38
rs766685723 Chr.12: 89472265-89472265 on Build GRCh38
rs776285096 Chr.12: 89472198-89472198 on Build GRCh38
rs1030388417 Chr.12: 89472481-89472481 on Build GRCh38
rs765990029 Chr.12: 89472212-89472213 on Build GRCh38
rs772508012 Chr.12: 89472605-89472605 on Build GRCh38
rs398055805 Chr.12: 89472288-89472297 on Build GRCh38
rs1055468623 Chr.12: 89472414-89472414 on Build GRCh38
rs779026566 Chr.12: 89472411-89472412 on Build GRCh38
rs191378952 Chr.12: 89472283-89472283 on Build GRCh38

View Full Product Details