Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91056078-91056623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
UniGene
Hs.125750
Gene Chromosome Location
Chr.12: 91050491-91058354 on Build GRCh38
Transcripts
NM_007035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940282 c.91G>A p.D31N Chr.12: 91056191-91056191 on Build GRCh38
1606748 c.199C>T p.P67S Chr.12: 91056083-91056083 on Build GRCh38
6029706 c.42A>C p.I14I Chr.12: 91056240-91056240 on Build GRCh38
944365 c.185G>A p.G62D Chr.12: 91056097-91056097 on Build GRCh38
3466362 c.193G>A p.E65K Chr.12: 91056089-91056089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779808415 Chr.12: 91056397-91056397 on Build GRCh38
rs921025653 Chr.12: 91056507-91056507 on Build GRCh38
rs780006810 Chr.12: 91056275-91056275 on Build GRCh38
rs752745106 Chr.12: 91056257-91056257 on Build GRCh38
rs200335389 Chr.12: 91056532-91056532 on Build GRCh38
rs1026475863 Chr.12: 91056216-91056216 on Build GRCh38
rs776187520 Chr.12: 91056492-91056492 on Build GRCh38
rs370775164 Chr.12: 91056180-91056180 on Build GRCh38
rs1057060712 Chr.12: 91056327-91056327 on Build GRCh38
rs1009131291 Chr.12: 91056233-91056233 on Build GRCh38
rs755201083 Chr.12: 91056251-91056251 on Build GRCh38
rs757391405 Chr.12: 91056226-91056227 on Build GRCh38
rs1051227881 Chr.12: 91056527-91056527 on Build GRCh38
rs971329343 Chr.12: 91056402-91056402 on Build GRCh38
rs968432306 Chr.12: 91056281-91056281 on Build GRCh38
rs768219259 Chr.12: 91056482-91056482 on Build GRCh38
rs2735333 Chr.12: 91056213-91056213 on Build GRCh38
rs745496599 Chr.12: 91056102-91056102 on Build GRCh38
rs941032310 Chr.12: 91056463-91056463 on Build GRCh38
rs778721242 Chr.12: 91056186-91056186 on Build GRCh38
rs1017861123 Chr.12: 91056493-91056493 on Build GRCh38
rs556676522 Chr.12: 91056372-91056372 on Build GRCh38
rs762340226 Chr.12: 91056221-91056221 on Build GRCh38
rs965298134 Chr.12: 91056505-91056505 on Build GRCh38
rs765681497 Chr.12: 91056223-91056223 on Build GRCh38
rs968463385 Chr.12: 91056531-91056531 on Build GRCh38
rs569894609 Chr.12: 91056193-91056193 on Build GRCh38
rs773584773 Chr.12: 91056326-91056326 on Build GRCh38
rs746552555 Chr.12: 91056457-91056457 on Build GRCh38
rs769382533 Chr.12: 91056325-91056325 on Build GRCh38
rs993029345 Chr.12: 91056569-91056569 on Build GRCh38
rs529733507 Chr.12: 91056571-91056571 on Build GRCh38
rs903143563 Chr.12: 91056110-91056110 on Build GRCh38
rs767549485 Chr.12: 91056256-91056256 on Build GRCh38
rs756267298 Chr.12: 91056261-91056261 on Build GRCh38
rs1037670378 Chr.12: 91056423-91056423 on Build GRCh38
rs959918407 Chr.12: 91056567-91056567 on Build GRCh38
rs763193954 Chr.12: 91056227-91056227 on Build GRCh38
rs113770346 Chr.12: 91056564-91056564 on Build GRCh38
rs199813838 Chr.12: 91056162-91056162 on Build GRCh38
rs377616201 Chr.12: 91056172-91056172 on Build GRCh38
rs778947165 Chr.12: 91056296-91056296 on Build GRCh38
rs1021245039 Chr.12: 91056526-91056526 on Build GRCh38
rs746385866 Chr.12: 91056313-91056314 on Build GRCh38
rs749186513 Chr.12: 91056176-91056176 on Build GRCh38
rs78304229 Chr.12: 91056441-91056441 on Build GRCh38
rs749435376 Chr.12: 91056198-91056198 on Build GRCh38
rs201524750 Chr.12: 91056196-91056196 on Build GRCh38
rs775332060 Chr.12: 91056111-91056111 on Build GRCh38
rs540605650 Chr.12: 91056551-91056551 on Build GRCh38
rs760233605 Chr.12: 91056113-91056113 on Build GRCh38
rs746591463 Chr.12: 91056277-91056277 on Build GRCh38
rs751725649 Chr.12: 91056242-91056242 on Build GRCh38
rs750443030 Chr.12: 91056273-91056273 on Build GRCh38
rs773814068 Chr.12: 91056226-91056226 on Build GRCh38
rs113570801 Chr.12: 91056536-91056536 on Build GRCh38
rs777263244 Chr.12: 91056121-91056121 on Build GRCh38
rs747586192 Chr.12: 91056316-91056316 on Build GRCh38
rs765570180 Chr.12: 91056122-91056122 on Build GRCh38
rs12320366 Chr.12: 91056207-91056207 on Build GRCh38
rs543283787 Chr.12: 91056349-91056349 on Build GRCh38
rs771645894 Chr.12: 91056106-91056106 on Build GRCh38
rs996560503 Chr.12: 91056510-91056510 on Build GRCh38
rs775278959 Chr.12: 91056200-91056200 on Build GRCh38
rs758255217 Chr.12: 91056274-91056274 on Build GRCh38
rs202094304 Chr.12: 91056154-91056154 on Build GRCh38
rs751817390 Chr.12: 91056161-91056161 on Build GRCh38
rs915837653 Chr.12: 91056588-91056588 on Build GRCh38
rs908241916 Chr.12: 91056455-91056455 on Build GRCh38
rs141853861 Chr.12: 91056591-91056591 on Build GRCh38
rs116592190 Chr.12: 91056534-91056534 on Build GRCh38
rs147494238 Chr.12: 91056264-91056264 on Build GRCh38
rs867739680 Chr.12: 91056136-91056136 on Build GRCh38
rs750783253 Chr.12: 91056129-91056129 on Build GRCh38
rs938609998 Chr.12: 91056144-91056144 on Build GRCh38
rs374273251 Chr.12: 91056528-91056528 on Build GRCh38
rs866472061 Chr.12: 91056259-91056259 on Build GRCh38
rs564743909 Chr.12: 91056477-91056477 on Build GRCh38
rs758594193 Chr.12: 91056151-91056151 on Build GRCh38
rs763505751 Chr.12: 91056332-91056332 on Build GRCh38
rs141931395 Chr.12: 91056283-91056283 on Build GRCh38
rs28776777 Chr.12: 91056305-91056305 on Build GRCh38
rs865792604 Chr.12: 91056432-91056432 on Build GRCh38
rs766793375 Chr.12: 91056231-91056231 on Build GRCh38
rs185814382 Chr.12: 91056436-91056436 on Build GRCh38
rs763765245 Chr.12: 91056115-91056115 on Build GRCh38

View Full Product Details