Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95475720-95476229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP2
Entrez Gene ID
10988
Gene Name
methionyl aminopeptidase 2
Gene Aliases
MAP2, MNPEP, p67eIF2
UniGene
Hs.444986
Gene Chromosome Location
Chr.12: 95474046-95515839 on Build GRCh38
Transcripts
NM_001317182.1 NM_001317183.1 NM_006838.3 NR_133673.1 XM_005268583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2046291 c.189A>G p.S63S Chr.12: 95476108-95476108 on Build GRCh38
5922981 c.173A>C p.K58T Chr.12: 95476092-95476092 on Build GRCh38
3466589 c.193G>A p.D65N Chr.12: 95476112-95476112 on Build GRCh38
1364967 c.223G>A p.A75T Chr.12: 95476142-95476142 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1017986823 Chr.12: 95476032-95476032 on Build GRCh38
rs769705278 Chr.12: 95476123-95476123 on Build GRCh38
rs761105644 Chr.12: 95476192-95476192 on Build GRCh38
rs991551146 Chr.12: 95475906-95475906 on Build GRCh38
rs754237518 Chr.12: 95476071-95476071 on Build GRCh38
rs1005275712 Chr.12: 95475756-95475756 on Build GRCh38
rs149754522 Chr.12: 95476115-95476115 on Build GRCh38
rs906473475 Chr.12: 95475976-95475976 on Build GRCh38
rs770452136 Chr.12: 95476159-95476159 on Build GRCh38
rs201661818 Chr.12: 95476169-95476169 on Build GRCh38
rs748322002 Chr.12: 95476028-95476028 on Build GRCh38
rs973399178 Chr.12: 95475830-95475830 on Build GRCh38
rs777643357 Chr.12: 95476144-95476144 on Build GRCh38
rs977589737 Chr.12: 95475864-95475864 on Build GRCh38
rs926097026 Chr.12: 95475880-95475880 on Build GRCh38
rs748153343 Chr.12: 95476113-95476113 on Build GRCh38
rs113478948 Chr.12: 95475949-95475954 on Build GRCh38
rs1010998533 Chr.12: 95475957-95475957 on Build GRCh38
rs781252542 Chr.12: 95476108-95476108 on Build GRCh38
rs986164696 Chr.12: 95475820-95475820 on Build GRCh38
rs573814841 Chr.12: 95475846-95475846 on Build GRCh38
rs754098979 Chr.12: 95476199-95476199 on Build GRCh38
rs145704880 Chr.12: 95476171-95476171 on Build GRCh38
rs200315154 Chr.12: 95476132-95476132 on Build GRCh38
rs773686915 Chr.12: 95476022-95476022 on Build GRCh38
rs200564504 Chr.12: 95476189-95476189 on Build GRCh38
rs937769004 Chr.12: 95475907-95475907 on Build GRCh38
rs753255083 Chr.12: 95476033-95476033 on Build GRCh38
rs1046635034 Chr.12: 95475886-95475886 on Build GRCh38
rs1046142984 Chr.12: 95475967-95475967 on Build GRCh38
rs936107277 Chr.12: 95475888-95475888 on Build GRCh38
rs118187064 Chr.12: 95475807-95475807 on Build GRCh38
rs767282204 Chr.12: 95475937-95475937 on Build GRCh38
rs757359924 Chr.12: 95476081-95476081 on Build GRCh38
rs758542541 Chr.12: 95476100-95476100 on Build GRCh38
rs368730523 Chr.12: 95476099-95476099 on Build GRCh38
rs915905367 Chr.12: 95475942-95475942 on Build GRCh38
rs541143586 Chr.12: 95475970-95475970 on Build GRCh38
rs201989616 Chr.12: 95476055-95476055 on Build GRCh38
rs112690176 Chr.12: 95475788-95475788 on Build GRCh38
rs950114391 Chr.12: 95475965-95475965 on Build GRCh38
rs79116716 Chr.12: 95475826-95475826 on Build GRCh38
rs202160340 Chr.12: 95476174-95476174 on Build GRCh38
rs200823558 Chr.12: 95476083-95476083 on Build GRCh38
rs761017724 Chr.12: 95476034-95476034 on Build GRCh38
rs767981481 Chr.12: 95476029-95476029 on Build GRCh38
rs763836204 Chr.12: 95476160-95476161 on Build GRCh38
rs762264576 Chr.12: 95476201-95476201 on Build GRCh38
rs184726541 Chr.12: 95476164-95476164 on Build GRCh38
rs756360751 Chr.12: 95476163-95476164 on Build GRCh38
rs370409255 Chr.12: 95475952-95475952 on Build GRCh38
rs929863985 Chr.12: 95475980-95475980 on Build GRCh38
rs977415012 Chr.12: 95475824-95475824 on Build GRCh38
rs888225109 Chr.12: 95475744-95475744 on Build GRCh38
rs771632818 Chr.12: 95476185-95476185 on Build GRCh38
rs759297604 Chr.12: 95475849-95475849 on Build GRCh38
rs1018118007 Chr.12: 95476152-95476152 on Build GRCh38
rs77402056 Chr.12: 95475828-95475829 on Build GRCh38
rs1029446375 Chr.12: 95475804-95475804 on Build GRCh38
rs754193797 Chr.12: 95476012-95476012 on Build GRCh38
rs377490393 Chr.12: 95475845-95475845 on Build GRCh38
rs181755135 Chr.12: 95475827-95475827 on Build GRCh38
rs375569750 Chr.12: 95476196-95476196 on Build GRCh38
rs749131729 Chr.12: 95476157-95476157 on Build GRCh38
rs1046943591 Chr.12: 95476003-95476003 on Build GRCh38

View Full Product Details