Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95493881-95494401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP2
Entrez Gene ID
10988
Gene Name
methionyl aminopeptidase 2
Gene Aliases
MAP2, MNPEP, p67eIF2
UniGene
Hs.444986
Gene Chromosome Location
Chr.12: 95474046-95515839 on Build GRCh38
Transcripts
NM_001317182.1 NM_001317183.1 NM_006838.3 NR_133673.1 XM_005268583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423385 c.431G>T p.R144L Chr.12: 95494058-95494058 on Build GRCh38
1364970 c.473T>C p.L158S Chr.12: 95494100-95494100 on Build GRCh38
3466592 c.434C>G p.T145R Chr.12: 95494061-95494061 on Build GRCh38
4428409 c.430C>T p.R144* Chr.12: 95494057-95494057 on Build GRCh38
7376945 c.579G>C p.M193I Chr.12: 95494206-95494206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201196524 Chr.12: 95494066-95494066 on Build GRCh38
rs932992297 Chr.12: 95494191-95494191 on Build GRCh38
rs1025031661 Chr.12: 95494268-95494268 on Build GRCh38
rs755142238 Chr.12: 95494194-95494194 on Build GRCh38
rs774341974 Chr.12: 95494227-95494227 on Build GRCh38
rs199543655 Chr.12: 95494236-95494236 on Build GRCh38
rs557021179 Chr.12: 95493933-95493933 on Build GRCh38
rs868060906 Chr.12: 95494136-95494136 on Build GRCh38
rs910499146 Chr.12: 95494322-95494322 on Build GRCh38
rs200000248 Chr.12: 95494249-95494249 on Build GRCh38
rs749624333 Chr.12: 95494341-95494341 on Build GRCh38
rs750947100 Chr.12: 95494158-95494158 on Build GRCh38
rs952819592 Chr.12: 95494318-95494318 on Build GRCh38
rs377595585 Chr.12: 95494005-95494005 on Build GRCh38
rs200486702 Chr.12: 95494131-95494131 on Build GRCh38
rs188122925 Chr.12: 95494348-95494348 on Build GRCh38
rs374477849 Chr.12: 95493910-95493910 on Build GRCh38
rs964554243 Chr.12: 95494367-95494367 on Build GRCh38
rs201323246 Chr.12: 95494058-95494058 on Build GRCh38
rs1025677953 Chr.12: 95494298-95494298 on Build GRCh38
rs531454405 Chr.12: 95494284-95494284 on Build GRCh38
rs778196232 Chr.12: 95494230-95494232 on Build GRCh38
rs1023494073 Chr.12: 95494122-95494122 on Build GRCh38
rs759362738 Chr.12: 95494079-95494079 on Build GRCh38
rs113519867 Chr.12: 95494006-95494006 on Build GRCh38
rs763632330 Chr.12: 95494029-95494029 on Build GRCh38
rs1001160910 Chr.12: 95494201-95494201 on Build GRCh38
rs986486884 Chr.12: 95493983-95493983 on Build GRCh38
rs201703803 Chr.12: 95494238-95494238 on Build GRCh38
rs776476167 Chr.12: 95494252-95494252 on Build GRCh38
rs564385859 Chr.12: 95494269-95494269 on Build GRCh38
rs769920214 Chr.12: 95494226-95494226 on Build GRCh38
rs932970283 Chr.12: 95493969-95493969 on Build GRCh38
rs111286983 Chr.12: 95494266-95494266 on Build GRCh38
rs147032981 Chr.12: 95494290-95494290 on Build GRCh38
rs142508741 Chr.12: 95494076-95494076 on Build GRCh38
rs146718454 Chr.12: 95494098-95494098 on Build GRCh38
rs141578207 Chr.12: 95494173-95494173 on Build GRCh38
rs748195097 Chr.12: 95494203-95494203 on Build GRCh38
rs753363029 Chr.12: 95494112-95494112 on Build GRCh38
rs1032480720 Chr.12: 95493972-95493972 on Build GRCh38
rs200947777 Chr.12: 95494014-95494014 on Build GRCh38
rs139276469 Chr.12: 95494114-95494114 on Build GRCh38
rs147951626 Chr.12: 95494119-95494119 on Build GRCh38
rs200594182 Chr.12: 95494110-95494110 on Build GRCh38
rs780214322 Chr.12: 95494176-95494176 on Build GRCh38
rs774195553 Chr.12: 95494077-95494077 on Build GRCh38
rs759279776 Chr.12: 95494022-95494022 on Build GRCh38
rs776632195 Chr.12: 95494060-95494060 on Build GRCh38
rs746546994 Chr.12: 95493939-95493939 on Build GRCh38
rs781422875 Chr.12: 95494197-95494197 on Build GRCh38
rs767176764 Chr.12: 95494023-95494023 on Build GRCh38
rs755647039 Chr.12: 95494027-95494027 on Build GRCh38
rs950626889 Chr.12: 95494050-95494050 on Build GRCh38
rs529784453 Chr.12: 95494052-95494052 on Build GRCh38
rs902775036 Chr.12: 95494200-95494200 on Build GRCh38
rs201929594 Chr.12: 95494174-95494174 on Build GRCh38
rs200273310 Chr.12: 95494062-95494062 on Build GRCh38
rs200617355 Chr.12: 95494237-95494237 on Build GRCh38
rs780306578 Chr.12: 95494057-95494057 on Build GRCh38
rs747412133 Chr.12: 95494182-95494182 on Build GRCh38
rs757793010 Chr.12: 95494033-95494033 on Build GRCh38
rs753414576 Chr.12: 95494031-95494031 on Build GRCh38
rs941965312 Chr.12: 95494364-95494364 on Build GRCh38
rs543973521 Chr.12: 95493923-95493923 on Build GRCh38
rs200428360 Chr.12: 95494049-95494049 on Build GRCh38
rs760581075 Chr.12: 95494245-95494245 on Build GRCh38
rs112688379 Chr.12: 95494187-95494187 on Build GRCh38
rs760189259 Chr.12: 95494105-95494105 on Build GRCh38
rs984747652 Chr.12: 95494333-95494333 on Build GRCh38
rs368706865 Chr.12: 95494096-95494096 on Build GRCh38
rs202192351 Chr.12: 95494041-95494041 on Build GRCh38
rs761389224 Chr.12: 95494265-95494265 on Build GRCh38
rs568123900 Chr.12: 95494294-95494294 on Build GRCh38
rs922934324 Chr.12: 95493937-95493937 on Build GRCh38
rs201713329 Chr.12: 95494059-95494059 on Build GRCh38

View Full Product Details