Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6592943-6593437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL21
Entrez Gene ID
9903
Gene Name
kelch like family member 21
Gene Aliases
-
UniGene
Hs.7764
Gene Chromosome Location
Chr.1: 6590724-6602898 on Build GRCh38
Transcripts
NM_001324309.1 NM_014851.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6696608 c.1742A>G p.D581G Chr.1: 6593417-6593417 on Build GRCh38
2239675 c.1767G>A p.P589P Chr.1: 6593392-6593392 on Build GRCh38
5053653 c.1757G>A p.R586Q Chr.1: 6593402-6593402 on Build GRCh38
7174912 c.1779C>T p.P593P Chr.1: 6593380-6593380 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs983867019 Chr.1: 6593140-6593140 on Build GRCh38
rs908293146 Chr.1: 6593325-6593325 on Build GRCh38
rs952570594 Chr.1: 6593305-6593305 on Build GRCh38
rs770370942 Chr.1: 6593326-6593326 on Build GRCh38
rs894569747 Chr.1: 6593039-6593039 on Build GRCh38
rs770497387 Chr.1: 6593409-6593409 on Build GRCh38
rs367791738 Chr.1: 6593387-6593387 on Build GRCh38
rs113764508 Chr.1: 6593068-6593068 on Build GRCh38
rs376195825 Chr.1: 6593350-6593350 on Build GRCh38
rs377276646 Chr.1: 6593364-6593364 on Build GRCh38
rs374292340 Chr.1: 6593360-6593360 on Build GRCh38
rs749999473 Chr.1: 6593339-6593339 on Build GRCh38
rs781097466 Chr.1: 6593354-6593354 on Build GRCh38
rs780899154 Chr.1: 6593179-6593179 on Build GRCh38
rs755966938 Chr.1: 6593242-6593242 on Build GRCh38
rs987899797 Chr.1: 6593043-6593043 on Build GRCh38
rs749497823 Chr.1: 6593329-6593330 on Build GRCh38
rs768243229 Chr.1: 6593335-6593335 on Build GRCh38
rs954478408 Chr.1: 6593291-6593291 on Build GRCh38
rs11811607 Chr.1: 6593279-6593279 on Build GRCh38
rs537646904 Chr.1: 6593393-6593393 on Build GRCh38
rs969745759 Chr.1: 6592997-6592997 on Build GRCh38
rs980598690 Chr.1: 6593013-6593013 on Build GRCh38
rs573788361 Chr.1: 6593075-6593075 on Build GRCh38
rs750408643 Chr.1: 6593390-6593390 on Build GRCh38
rs755318508 Chr.1: 6593005-6593005 on Build GRCh38
rs368737518 Chr.1: 6593314-6593314 on Build GRCh38
rs185426059 Chr.1: 6592996-6592996 on Build GRCh38
rs769044711 Chr.1: 6593368-6593368 on Build GRCh38
rs753562914 Chr.1: 6593346-6593346 on Build GRCh38
rs189473278 Chr.1: 6593059-6593059 on Build GRCh38
rs371149254 Chr.1: 6593370-6593370 on Build GRCh38
rs751851649 Chr.1: 6593351-6593351 on Build GRCh38
rs1019498817 Chr.1: 6593073-6593073 on Build GRCh38
rs907402124 Chr.1: 6593008-6593008 on Build GRCh38
rs939657277 Chr.1: 6593160-6593160 on Build GRCh38
rs987098835 Chr.1: 6593300-6593300 on Build GRCh38
rs1005386623 Chr.1: 6593403-6593403 on Build GRCh38
rs1046062679 Chr.1: 6593168-6593168 on Build GRCh38
rs578162244 Chr.1: 6593119-6593119 on Build GRCh38
rs755115961 Chr.1: 6593378-6593379 on Build GRCh38
rs750117523 Chr.1: 6593401-6593401 on Build GRCh38
rs796580371 Chr.1: 6593074-6593075 on Build GRCh38
rs895946683 Chr.1: 6593109-6593109 on Build GRCh38
rs899343653 Chr.1: 6593093-6593093 on Build GRCh38
rs560743615 Chr.1: 6593127-6593128 on Build GRCh38
rs771799484 Chr.1: 6593371-6593371 on Build GRCh38
rs371099946 Chr.1: 6593352-6593352 on Build GRCh38
rs759354176 Chr.1: 6593381-6593381 on Build GRCh38
rs200232322 Chr.1: 6593397-6593397 on Build GRCh38
rs555223365 Chr.1: 6593074-6593074 on Build GRCh38
rs141252990 Chr.1: 6593380-6593380 on Build GRCh38
rs766962294 Chr.1: 6593338-6593338 on Build GRCh38
rs74049690 Chr.1: 6593171-6593171 on Build GRCh38
rs1028595929 Chr.1: 6593298-6593298 on Build GRCh38
rs886762991 Chr.1: 6592980-6592980 on Build GRCh38
rs142755720 Chr.1: 6592992-6592992 on Build GRCh38
rs756226341 Chr.1: 6593392-6593392 on Build GRCh38
rs143513148 Chr.1: 6593396-6593396 on Build GRCh38
rs757545813 Chr.1: 6593388-6593388 on Build GRCh38
rs549662495 Chr.1: 6593243-6593243 on Build GRCh38
rs41278024 Chr.1: 6593062-6593062 on Build GRCh38
rs371853612 Chr.1: 6593177-6593177 on Build GRCh38
rs748581256 Chr.1: 6593400-6593400 on Build GRCh38
rs562989987 Chr.1: 6593095-6593095 on Build GRCh38
rs981790321 Chr.1: 6593333-6593333 on Build GRCh38
rs754825029 Chr.1: 6593349-6593349 on Build GRCh38
rs377537456 Chr.1: 6593345-6593345 on Build GRCh38
rs533804098 Chr.1: 6593389-6593389 on Build GRCh38
rs375443749 Chr.1: 6593379-6593379 on Build GRCh38
rs1019550864 Chr.1: 6593257-6593257 on Build GRCh38
rs772593214 Chr.1: 6593372-6593372 on Build GRCh38
rs957787044 Chr.1: 6593016-6593016 on Build GRCh38
rs772932433 Chr.1: 6593402-6593402 on Build GRCh38
rs1034477133 Chr.1: 6593014-6593014 on Build GRCh38
rs376561822 Chr.1: 6593412-6593413 on Build GRCh38
rs1014203825 Chr.1: 6593153-6593153 on Build GRCh38
rs550278332 Chr.1: 6592987-6592987 on Build GRCh38
rs774001863 Chr.1: 6593337-6593337 on Build GRCh38
rs1047786798 Chr.1: 6593102-6593102 on Build GRCh38
rs1012114671 Chr.1: 6593207-6593207 on Build GRCh38
rs931082736 Chr.1: 6593116-6593116 on Build GRCh38
rs575651447 Chr.1: 6593274-6593275 on Build GRCh38
rs1005159469 Chr.1: 6592994-6592994 on Build GRCh38
rs545412937 Chr.1: 6593167-6593167 on Build GRCh38
rs760500033 Chr.1: 6593341-6593341 on Build GRCh38
rs550957678 Chr.1: 6593386-6593386 on Build GRCh38
rs1021705041 Chr.1: 6593237-6593237 on Build GRCh38
rs969960084 Chr.1: 6593066-6593066 on Build GRCh38
rs1003049182 Chr.1: 6593012-6593012 on Build GRCh38
rs894625143 Chr.1: 6593187-6593187 on Build GRCh38
rs777790336 Chr.1: 6593248-6593248 on Build GRCh38
rs754754784 Chr.1: 6593027-6593027 on Build GRCh38
rs531955822 Chr.1: 6593282-6593282 on Build GRCh38
rs530864532 Chr.1: 6593054-6593054 on Build GRCh38
rs527934091 Chr.1: 6593185-6593185 on Build GRCh38
rs908243025 Chr.1: 6593158-6593158 on Build GRCh38
rs374109878 Chr.1: 6593397-6593398 on Build GRCh38
rs548994144 Chr.1: 6593180-6593180 on Build GRCh38
rs201663664 Chr.1: 6593327-6593327 on Build GRCh38
rs910796660 Chr.1: 6593071-6593071 on Build GRCh38
rs763362965 Chr.1: 6593328-6593328 on Build GRCh38
rs544343328 Chr.1: 6593090-6593090 on Build GRCh38
rs373921277 Chr.1: 6593315-6593315 on Build GRCh38
rs770184255 Chr.1: 6593363-6593363 on Build GRCh38

View Full Product Details