Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6638153-6638670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
UniGene
Hs.462640
Gene Chromosome Location
Chr.1: 6634168-6701906 on Build GRCh38
Transcripts
NM_018198.3 XM_017001745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291672 c.1313C>T p.A438V Chr.1: 6638305-6638305 on Build GRCh38
4009527 c.1254-2A>G p.? Chr.1: 6638366-6638366 on Build GRCh38
7268259 c.1254-6C>T p.? Chr.1: 6638370-6638370 on Build GRCh38
299182 c.1254-8C>T p.? Chr.1: 6638372-6638372 on Build GRCh38
3491933 c.1272G>A p.R424R Chr.1: 6638346-6638346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891620499 Chr.1: 6638511-6638511 on Build GRCh38
rs377476862 Chr.1: 6638382-6638382 on Build GRCh38
rs1044699857 Chr.1: 6638494-6638494 on Build GRCh38
rs903860827 Chr.1: 6638460-6638460 on Build GRCh38
rs554152969 Chr.1: 6638527-6638527 on Build GRCh38
rs200987537 Chr.1: 6638340-6638340 on Build GRCh38
rs919455389 Chr.1: 6638183-6638183 on Build GRCh38
rs1016249394 Chr.1: 6638643-6638643 on Build GRCh38
rs777159794 Chr.1: 6638355-6638355 on Build GRCh38
rs765314517 Chr.1: 6638524-6638524 on Build GRCh38
rs949005458 Chr.1: 6638484-6638484 on Build GRCh38
rs764909374 Chr.1: 6638327-6638327 on Build GRCh38
rs139633039 Chr.1: 6638297-6638297 on Build GRCh38
rs370489757 Chr.1: 6638391-6638391 on Build GRCh38
rs146854619 Chr.1: 6638242-6638242 on Build GRCh38
rs751135995 Chr.1: 6638370-6638370 on Build GRCh38
rs773296928 Chr.1: 6638341-6638341 on Build GRCh38
rs140594822 Chr.1: 6638339-6638339 on Build GRCh38
rs765576477 Chr.1: 6638357-6638357 on Build GRCh38
rs763172106 Chr.1: 6638581-6638581 on Build GRCh38
rs577611781 Chr.1: 6638435-6638435 on Build GRCh38
rs1056567520 Chr.1: 6638309-6638309 on Build GRCh38
rs756867738 Chr.1: 6638371-6638371 on Build GRCh38
rs916159700 Chr.1: 6638474-6638474 on Build GRCh38
rs750444549 Chr.1: 6638376-6638376 on Build GRCh38
rs773708089 Chr.1: 6638315-6638315 on Build GRCh38
rs992998585 Chr.1: 6638436-6638436 on Build GRCh38
rs556242190 Chr.1: 6638426-6638426 on Build GRCh38
rs759920176 Chr.1: 6638356-6638356 on Build GRCh38
rs955078457 Chr.1: 6638238-6638238 on Build GRCh38
rs778233892 Chr.1: 6638256-6638256 on Build GRCh38
rs754642448 Chr.1: 6638249-6638249 on Build GRCh38
rs536424394 Chr.1: 6638406-6638410 on Build GRCh38
rs889023146 Chr.1: 6638606-6638606 on Build GRCh38
rs759145267 Chr.1: 6638325-6638325 on Build GRCh38
rs745627956 Chr.1: 6638336-6638336 on Build GRCh38
rs1052204438 Chr.1: 6638563-6638563 on Build GRCh38
rs764944103 Chr.1: 6638476-6638478 on Build GRCh38
rs972628592 Chr.1: 6638179-6638179 on Build GRCh38
rs1051637641 Chr.1: 6638582-6638582 on Build GRCh38
rs748237613 Chr.1: 6638304-6638304 on Build GRCh38
rs767540150 Chr.1: 6638372-6638372 on Build GRCh38
rs537909510 Chr.1: 6638425-6638425 on Build GRCh38
rs752910813 Chr.1: 6638385-6638385 on Build GRCh38
rs988527752 Chr.1: 6638196-6638196 on Build GRCh38
rs997354681 Chr.1: 6638575-6638575 on Build GRCh38
rs41278032 Chr.1: 6638324-6638324 on Build GRCh38
rs375687606 Chr.1: 6638367-6638367 on Build GRCh38
rs779310757 Chr.1: 6638611-6638611 on Build GRCh38
rs755935506 Chr.1: 6638378-6638378 on Build GRCh38
rs755913541 Chr.1: 6638337-6638337 on Build GRCh38
rs775730223 Chr.1: 6638406-6638406 on Build GRCh38
rs371867795 Chr.1: 6638403-6638403 on Build GRCh38
rs757910644 Chr.1: 6638329-6638329 on Build GRCh38
rs1000651434 Chr.1: 6638322-6638322 on Build GRCh38
rs376085938 Chr.1: 6638607-6638607 on Build GRCh38
rs955759086 Chr.1: 6638178-6638178 on Build GRCh38
rs201717591 Chr.1: 6638277-6638277 on Build GRCh38
rs545423666 Chr.1: 6638446-6638446 on Build GRCh38
rs374928580 Chr.1: 6638397-6638397 on Build GRCh38
rs747469514 Chr.1: 6638343-6638343 on Build GRCh38
rs752279412 Chr.1: 6638260-6638260 on Build GRCh38
rs866621019 Chr.1: 6638215-6638215 on Build GRCh38
rs892261194 Chr.1: 6638568-6638568 on Build GRCh38
rs771455111 Chr.1: 6638347-6638347 on Build GRCh38
rs1018325398 Chr.1: 6638628-6638628 on Build GRCh38
rs367644840 Chr.1: 6638400-6638400 on Build GRCh38
rs781137767 Chr.1: 6638278-6638278 on Build GRCh38
rs150490156 Chr.1: 6638369-6638369 on Build GRCh38
rs771044478 Chr.1: 6638399-6638399 on Build GRCh38
rs775201877 Chr.1: 6638360-6638360 on Build GRCh38
rs768953446 Chr.1: 6638295-6638295 on Build GRCh38
rs757599456 Chr.1: 6638549-6638549 on Build GRCh38
rs757273974 Chr.1: 6638331-6638331 on Build GRCh38
rs948018291 Chr.1: 6638447-6638447 on Build GRCh38
rs766676805 Chr.1: 6638320-6638320 on Build GRCh38
rs200095851 Chr.1: 6638398-6638398 on Build GRCh38
rs373360478 Chr.1: 6638266-6638266 on Build GRCh38
rs749462472 Chr.1: 6638279-6638279 on Build GRCh38
rs752340028 Chr.1: 6638328-6638328 on Build GRCh38
rs941519109 Chr.1: 6638263-6638263 on Build GRCh38
rs960241166 Chr.1: 6638428-6638428 on Build GRCh38
rs377744080 Chr.1: 6638267-6638267 on Build GRCh38
rs761778663 Chr.1: 6638402-6638402 on Build GRCh38
rs1045450898 Chr.1: 6638453-6638453 on Build GRCh38
rs182172432 Chr.1: 6638268-6638268 on Build GRCh38
rs762617759 Chr.1: 6638412-6638412 on Build GRCh38
rs143752788 Chr.1: 6638223-6638223 on Build GRCh38
rs924997638 Chr.1: 6638542-6638542 on Build GRCh38
rs41278034 Chr.1: 6638330-6638330 on Build GRCh38
rs772103836 Chr.1: 6638305-6638305 on Build GRCh38
rs201664213 Chr.1: 6638319-6638319 on Build GRCh38
rs781094762 Chr.1: 6638332-6638332 on Build GRCh38

View Full Product Details