Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10430023-10430530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1
Entrez Gene ID
378708
Gene Name
apoptosis-inducing, TAF9-like domain 1
Gene Aliases
CENP-S, CENPS, FAAP16, MHF1
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10442815 on Build GRCh38
Transcripts
NM_199294.2 NM_001270517.1 NM_198544.3 NM_199006.2

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_199294.2 NM_001270517.1 NM_198544.3 NM_199006.2

SNP Detail

SNP ID Chromosome Location
rs763077885 Chr.1: 10430112-10430114 on Build GRCh38
rs748065363 Chr.1: 10430480-10430480 on Build GRCh38
rs775531844 Chr.1: 10430502-10430502 on Build GRCh38
rs1056204903 Chr.1: 10430470-10430470 on Build GRCh38
rs200229439 Chr.1: 10430326-10430327 on Build GRCh38
rs956760687 Chr.1: 10430322-10430322 on Build GRCh38
rs144662034 Chr.1: 10430154-10430154 on Build GRCh38
rs946382972 Chr.1: 10430287-10430287 on Build GRCh38
rs1022697274 Chr.1: 10430336-10430336 on Build GRCh38
rs914873946 Chr.1: 10430273-10430273 on Build GRCh38
rs552013045 Chr.1: 10430330-10430330 on Build GRCh38
rs545745253 Chr.1: 10430379-10430379 on Build GRCh38
rs649101 Chr.1: 10430054-10430054 on Build GRCh38
rs1009542705 Chr.1: 10430326-10430326 on Build GRCh38
rs1046757246 Chr.1: 10430412-10430412 on Build GRCh38
rs772057575 Chr.1: 10430481-10430481 on Build GRCh38
rs966202767 Chr.1: 10430371-10430371 on Build GRCh38
rs914987060 Chr.1: 10430500-10430500 on Build GRCh38
rs754914445 Chr.1: 10430408-10430408 on Build GRCh38
rs928163130 Chr.1: 10430145-10430145 on Build GRCh38
rs1035077922 Chr.1: 10430305-10430305 on Build GRCh38
rs1045392202 Chr.1: 10430445-10430445 on Build GRCh38
rs556332764 Chr.1: 10430328-10430328 on Build GRCh38
rs566706183 Chr.1: 10430073-10430073 on Build GRCh38
rs1009591798 Chr.1: 10430486-10430486 on Build GRCh38
rs553739349 Chr.1: 10430461-10430461 on Build GRCh38
rs905086890 Chr.1: 10430455-10430455 on Build GRCh38
rs746721788 Chr.1: 10430491-10430491 on Build GRCh38
rs982226049 Chr.1: 10430111-10430111 on Build GRCh38
rs1040813131 Chr.1: 10430094-10430094 on Build GRCh38
rs994262445 Chr.1: 10430374-10430374 on Build GRCh38
rs910757741 Chr.1: 10430349-10430349 on Build GRCh38
rs544729355 Chr.1: 10430418-10430418 on Build GRCh38
rs779042321 Chr.1: 10430475-10430475 on Build GRCh38
rs661256 Chr.1: 10430482-10430482 on Build GRCh38
rs916839985 Chr.1: 10430068-10430068 on Build GRCh38
rs968355945 Chr.1: 10430345-10430345 on Build GRCh38
rs760314861 Chr.1: 10430210-10430210 on Build GRCh38
rs922358127 Chr.1: 10430393-10430393 on Build GRCh38
rs929634957 Chr.1: 10430195-10430195 on Build GRCh38
rs148511711 Chr.1: 10430293-10430293 on Build GRCh38
rs905022360 Chr.1: 10430272-10430272 on Build GRCh38
rs556063628 Chr.1: 10430213-10430213 on Build GRCh38
rs929693914 Chr.1: 10430405-10430405 on Build GRCh38
rs116395144 Chr.1: 10430465-10430465 on Build GRCh38
rs868153548 Chr.1: 10430090-10430090 on Build GRCh38
rs941327180 Chr.1: 10430433-10430433 on Build GRCh38
rs1046734884 Chr.1: 10430208-10430208 on Build GRCh38
rs649639 Chr.1: 10430228-10430228 on Build GRCh38
rs942458406 Chr.1: 10430059-10430059 on Build GRCh38
rs986777663 Chr.1: 10430348-10430348 on Build GRCh38
rs888394589 Chr.1: 10430210-10430210 on Build GRCh38
rs948196816 Chr.1: 10430101-10430101 on Build GRCh38
rs554024579 Chr.1: 10430188-10430188 on Build GRCh38
rs952711770 Chr.1: 10430423-10430423 on Build GRCh38
rs1007519312 Chr.1: 10430506-10430506 on Build GRCh38
rs34266391 Chr.1: 10430225-10430225 on Build GRCh38
rs661272 Chr.1: 10430493-10430493 on Build GRCh38
rs1005981346 Chr.1: 10430430-10430430 on Build GRCh38

View Full Product Details