Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96934890-96935388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEDD1
Entrez Gene ID
121441
Gene Name
neural precursor cell expressed, developmentally down-regulated 1
Gene Aliases
GCP-WD, TUBGCP7
UniGene
Hs.270084
Gene Chromosome Location
Chr.12: 96907222-96953691 on Build GRCh38
Transcripts
NM_001135175.1 NM_001135176.1 NM_001135177.1 NM_152905.3 XM_005268644.2 XM_006719237.3 XM_011537903.2 XM_011537904.2 XM_011537905.2 XM_011537906.2 XM_017018801.1 XM_017018802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6553610 c.499C>T p.H167Y Chr.12: 96934985-96934985 on Build GRCh38
6306181 c.733A>C p.S245R Chr.12: 96935198-96935198 on Build GRCh38
944794 c.619G>T p.A207S Chr.12: 96935105-96935105 on Build GRCh38
6357592 c.581G>A p.S194N Chr.12: 96935067-96935067 on Build GRCh38
469112 c.663C>G p.L221L Chr.12: 96935149-96935149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750416970 Chr.12: 96935245-96935245 on Build GRCh38
rs938203713 Chr.12: 96935323-96935323 on Build GRCh38
rs201968415 Chr.12: 96934937-96934937 on Build GRCh38
rs908047855 Chr.12: 96935070-96935070 on Build GRCh38
rs753638609 Chr.12: 96935252-96935253 on Build GRCh38
rs1030811743 Chr.12: 96934949-96934949 on Build GRCh38
rs144154513 Chr.12: 96934977-96934977 on Build GRCh38
rs746909964 Chr.12: 96934996-96934996 on Build GRCh38
rs769132307 Chr.12: 96935220-96935220 on Build GRCh38
rs754885884 Chr.12: 96935309-96935309 on Build GRCh38
rs778219585 Chr.12: 96935175-96935175 on Build GRCh38
rs772667382 Chr.12: 96935190-96935190 on Build GRCh38
rs767480249 Chr.12: 96935255-96935255 on Build GRCh38
rs1030619609 Chr.12: 96935135-96935135 on Build GRCh38
rs780983875 Chr.12: 96935112-96935112 on Build GRCh38
rs180934258 Chr.12: 96934972-96934972 on Build GRCh38
rs548810330 Chr.12: 96935258-96935258 on Build GRCh38
rs770163663 Chr.12: 96935298-96935298 on Build GRCh38
rs751523284 Chr.12: 96935103-96935103 on Build GRCh38
rs772737741 Chr.12: 96935030-96935030 on Build GRCh38
rs764452619 Chr.12: 96935092-96935092 on Build GRCh38
rs748866951 Chr.12: 96935031-96935031 on Build GRCh38
rs748878028 Chr.12: 96935171-96935171 on Build GRCh38
rs140680808 Chr.12: 96934982-96934982 on Build GRCh38
rs780902090 Chr.12: 96935017-96935017 on Build GRCh38
rs767998123 Chr.12: 96935059-96935059 on Build GRCh38
rs747798095 Chr.12: 96934927-96934927 on Build GRCh38
rs545296201 Chr.12: 96935113-96935113 on Build GRCh38
rs1052223260 Chr.12: 96935149-96935149 on Build GRCh38
rs772647839 Chr.12: 96934947-96934947 on Build GRCh38
rs530477785 Chr.12: 96935314-96935314 on Build GRCh38
rs78013157 Chr.12: 96934990-96934990 on Build GRCh38
rs780092017 Chr.12: 96934987-96934987 on Build GRCh38
rs199889686 Chr.12: 96935023-96935023 on Build GRCh38
rs772518586 Chr.12: 96935329-96935329 on Build GRCh38
rs919477866 Chr.12: 96935287-96935287 on Build GRCh38
rs150834909 Chr.12: 96935333-96935333 on Build GRCh38
rs982220269 Chr.12: 96935002-96935002 on Build GRCh38
rs776981627 Chr.12: 96935233-96935233 on Build GRCh38
rs763390496 Chr.12: 96934951-96934951 on Build GRCh38
rs769302162 Chr.12: 96934931-96934931 on Build GRCh38
rs754872911 Chr.12: 96934998-96934998 on Build GRCh38
rs746410089 Chr.12: 96935179-96935179 on Build GRCh38
rs775949319 Chr.12: 96935064-96935064 on Build GRCh38
rs200076088 Chr.12: 96934944-96934944 on Build GRCh38
rs777449805 Chr.12: 96935118-96935118 on Build GRCh38
rs947000427 Chr.12: 96935340-96935340 on Build GRCh38
rs942168588 Chr.12: 96935361-96935361 on Build GRCh38
rs143026492 Chr.12: 96935120-96935120 on Build GRCh38
rs957870449 Chr.12: 96935249-96935249 on Build GRCh38
rs761878864 Chr.12: 96935237-96935237 on Build GRCh38
rs757254484 Chr.12: 96934986-96934986 on Build GRCh38
rs373391483 Chr.12: 96935187-96935187 on Build GRCh38
rs900616568 Chr.12: 96935117-96935117 on Build GRCh38
rs942053179 Chr.12: 96935095-96935095 on Build GRCh38
rs74829058 Chr.12: 96935321-96935321 on Build GRCh38
rs754855019 Chr.12: 96935105-96935105 on Build GRCh38
rs185161765 Chr.12: 96935246-96935246 on Build GRCh38
rs747808314 Chr.12: 96935021-96935021 on Build GRCh38
rs753931076 Chr.12: 96934985-96934985 on Build GRCh38
rs770540675 Chr.12: 96935172-96935172 on Build GRCh38
rs779763541 Chr.12: 96935203-96935203 on Build GRCh38
rs759496769 Chr.12: 96935250-96935250 on Build GRCh38
rs996178057 Chr.12: 96935146-96935146 on Build GRCh38
rs755869609 Chr.12: 96935115-96935115 on Build GRCh38
rs546226677 Chr.12: 96935359-96935359 on Build GRCh38
rs770323459 Chr.12: 96935032-96935032 on Build GRCh38
rs765230177 Chr.12: 96935099-96935099 on Build GRCh38
rs761009009 Chr.12: 96935067-96935067 on Build GRCh38
rs774978952 Chr.12: 96935036-96935036 on Build GRCh38
rs963327175 Chr.12: 96935247-96935247 on Build GRCh38
rs1610921 Chr.12: 96934937-96934944 on Build GRCh38
rs953470877 Chr.12: 96935164-96935164 on Build GRCh38
rs200584281 Chr.12: 96935241-96935241 on Build GRCh38
rs377708251 Chr.12: 96935214-96935214 on Build GRCh38
rs969573911 Chr.12: 96935271-96935271 on Build GRCh38
rs200832974 Chr.12: 96935093-96935093 on Build GRCh38
rs774689097 Chr.12: 96934967-96934967 on Build GRCh38
rs1010736443 Chr.12: 96935257-96935257 on Build GRCh38
rs1023376713 Chr.12: 96935267-96935267 on Build GRCh38
rs756348400 Chr.12: 96934983-96934983 on Build GRCh38
rs759956083 Chr.12: 96935038-96935038 on Build GRCh38
rs761450553 Chr.12: 96934936-96934937 on Build GRCh38

View Full Product Details