Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98751263-98751709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
NM_001204065.1 NM_001204066.1 NM_001204067.1 NM_001204068.1 NM_001204069.1 NM_001204070.1 NM_001204079.1 NM_001204080.1 NM_001204081.1 NM_020140.3 NM_152788.4 NM_181670.3 XM_005269028.4 XM_005269029.4 XM_005269032.3 XM_006719504.3 XM_006719505.3 XM_006719506.3 XM_006719507.3 XM_006719508.3 XM_006719509.3 XM_006719510.3 XM_006719512.3 XM_006719513.3 XM_006719514.3 XM_011538571.2 XM_017019651.1 XM_017019652.1 XM_017019653.1 XM_017019654.1 XM_017019655.1 XM_017019656.1 XM_017019657.1 XM_017019658.1 XM_017019659.1 XM_017019660.1 XM_017019661.1 XM_017019662.1 XM_017019663.1 XM_017019664.1 XM_017019665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4780179 c.3661C>T p.R1221C Chr.12: 98751366-98751366 on Build GRCh38
1245086 c.1367A>C p.E456A Chr.12: 98751410-98751410 on Build GRCh38
4544497 c.3587G>A p.G1196E Chr.12: 98751440-98751440 on Build GRCh38
944911 c.3661C>T p.R1221C Chr.12: 98751366-98751366 on Build GRCh38
4544492 c.1335G>A p.G445G Chr.12: 98751442-98751442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758876045 Chr.12: 98751442-98751442 on Build GRCh38
rs986906074 Chr.12: 98751358-98751358 on Build GRCh38
rs372400178 Chr.12: 98751318-98751318 on Build GRCh38
rs558580368 Chr.12: 98751577-98751577 on Build GRCh38
rs765902934 Chr.12: 98751382-98751382 on Build GRCh38
rs898137921 Chr.12: 98751642-98751642 on Build GRCh38
rs745771542 Chr.12: 98751534-98751534 on Build GRCh38
rs761350003 Chr.12: 98751378-98751378 on Build GRCh38
rs778885837 Chr.12: 98751514-98751514 on Build GRCh38
rs1015212069 Chr.12: 98751315-98751315 on Build GRCh38
rs912602945 Chr.12: 98751365-98751365 on Build GRCh38
rs368912180 Chr.12: 98751475-98751475 on Build GRCh38
rs533781845 Chr.12: 98751558-98751558 on Build GRCh38
rs372408713 Chr.12: 98751480-98751480 on Build GRCh38
rs979313143 Chr.12: 98751536-98751536 on Build GRCh38
rs775169608 Chr.12: 98751541-98751541 on Build GRCh38
rs377650383 Chr.12: 98751427-98751427 on Build GRCh38
rs149038689 Chr.12: 98751682-98751682 on Build GRCh38
rs954330491 Chr.12: 98751348-98751348 on Build GRCh38
rs775146299 Chr.12: 98751652-98751652 on Build GRCh38
rs896265602 Chr.12: 98751582-98751582 on Build GRCh38
rs369534902 Chr.12: 98751376-98751376 on Build GRCh38
rs147552668 Chr.12: 98751653-98751653 on Build GRCh38
rs369136720 Chr.12: 98751547-98751547 on Build GRCh38
rs371105634 Chr.12: 98751551-98751551 on Build GRCh38
rs867103953 Chr.12: 98751366-98751366 on Build GRCh38
rs555911932 Chr.12: 98751647-98751647 on Build GRCh38
rs780263079 Chr.12: 98751445-98751445 on Build GRCh38
rs760292764 Chr.12: 98751322-98751337 on Build GRCh38
rs536378717 Chr.12: 98751357-98751357 on Build GRCh38
rs375968874 Chr.12: 98751400-98751400 on Build GRCh38
rs115108330 Chr.12: 98751556-98751556 on Build GRCh38
rs577028505 Chr.12: 98751578-98751578 on Build GRCh38
rs763029388 Chr.12: 98751648-98751648 on Build GRCh38
rs748190855 Chr.12: 98751320-98751320 on Build GRCh38
rs995633089 Chr.12: 98751663-98751663 on Build GRCh38
rs145009099 Chr.12: 98751375-98751375 on Build GRCh38
rs770856085 Chr.12: 98751329-98751329 on Build GRCh38
rs139586088 Chr.12: 98751347-98751347 on Build GRCh38
rs1025878159 Chr.12: 98751664-98751664 on Build GRCh38
rs762536492 Chr.12: 98751675-98751675 on Build GRCh38
rs1036685567 Chr.12: 98751639-98751639 on Build GRCh38
rs760221682 Chr.12: 98751367-98751367 on Build GRCh38
rs771949076 Chr.12: 98751535-98751535 on Build GRCh38
rs77210491 Chr.12: 98751579-98751579 on Build GRCh38
rs939769035 Chr.12: 98751620-98751620 on Build GRCh38
rs375969961 Chr.12: 98751495-98751495 on Build GRCh38
rs751905985 Chr.12: 98751459-98751459 on Build GRCh38
rs896162177 Chr.12: 98751288-98751288 on Build GRCh38
rs763736713 Chr.12: 98751322-98751323 on Build GRCh38
rs375471416 Chr.12: 98751342-98751342 on Build GRCh38
rs193139656 Chr.12: 98751343-98751343 on Build GRCh38
rs946642714 Chr.12: 98751481-98751481 on Build GRCh38
rs1004321712 Chr.12: 98751310-98751310 on Build GRCh38

View Full Product Details