Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100314360-100314903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCYL2
Entrez Gene ID
55681
Gene Name
SCY1 like pseudokinase 2
Gene Aliases
CVAK104
UniGene
Hs.506481
Gene Chromosome Location
Chr.12: 100267137-100341724 on Build GRCh38
Transcripts
NM_001317784.1 NM_017988.5 XM_017019618.1 XM_017019619.1 XM_017019620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6773596 c.970-10delT p.? Chr.12: 100314479-100314479 on Build GRCh38
3870519 c.1090C>T p.P364S Chr.12: 100314609-100314609 on Build GRCh38
1562403 c.984T>G p.D328E Chr.12: 100314503-100314503 on Build GRCh38
6582225 c.1044G>C p.Q348H Chr.12: 100314563-100314563 on Build GRCh38
76357 c.1063G>A p.G355R Chr.12: 100314582-100314582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113151055 Chr.12: 100314749-100314749 on Build GRCh38
rs556342343 Chr.12: 100314545-100314545 on Build GRCh38
rs889432064 Chr.12: 100314714-100314714 on Build GRCh38
rs772756195 Chr.12: 100314506-100314506 on Build GRCh38
rs767379211 Chr.12: 100314644-100314644 on Build GRCh38
rs1009420469 Chr.12: 100314612-100314612 on Build GRCh38
rs1025725268 Chr.12: 100314433-100314433 on Build GRCh38
rs146266840 Chr.12: 100314802-100314802 on Build GRCh38
rs936384005 Chr.12: 100314696-100314696 on Build GRCh38
rs1042573164 Chr.12: 100314649-100314650 on Build GRCh38
rs1019477983 Chr.12: 100314401-100314401 on Build GRCh38
rs747828737 Chr.12: 100314470-100314470 on Build GRCh38
rs1053451915 Chr.12: 100314708-100314708 on Build GRCh38
rs113531208 Chr.12: 100314414-100314414 on Build GRCh38
rs561962708 Chr.12: 100314854-100314854 on Build GRCh38
rs930604133 Chr.12: 100314419-100314419 on Build GRCh38
rs1007503892 Chr.12: 100314471-100314471 on Build GRCh38
rs1025864816 Chr.12: 100314783-100314783 on Build GRCh38
rs754426628 Chr.12: 100314860-100314862 on Build GRCh38
rs774395237 Chr.12: 100314591-100314591 on Build GRCh38
rs774986901 Chr.12: 100314411-100314411 on Build GRCh38
rs769406364 Chr.12: 100314494-100314494 on Build GRCh38
rs1049069626 Chr.12: 100314880-100314880 on Build GRCh38
rs958964632 Chr.12: 100314480-100314480 on Build GRCh38
rs764392966 Chr.12: 100314599-100314599 on Build GRCh38
rs762434775 Chr.12: 100314520-100314520 on Build GRCh38
rs749377219 Chr.12: 100314478-100314479 on Build GRCh38
rs529118962 Chr.12: 100314857-100314857 on Build GRCh38
rs776354053 Chr.12: 100314463-100314463 on Build GRCh38
rs560415574 Chr.12: 100314627-100314627 on Build GRCh38
rs750197098 Chr.12: 100314707-100314707 on Build GRCh38
rs768340799 Chr.12: 100314461-100314461 on Build GRCh38
rs958929314 Chr.12: 100314725-100314725 on Build GRCh38
rs199838820 Chr.12: 100314442-100314442 on Build GRCh38
rs917515298 Chr.12: 100314748-100314748 on Build GRCh38
rs762518263 Chr.12: 100314556-100314556 on Build GRCh38
rs773234646 Chr.12: 100314479-100314479 on Build GRCh38
rs371388513 Chr.12: 100314650-100314650 on Build GRCh38
rs930782885 Chr.12: 100314865-100314865 on Build GRCh38
rs368534253 Chr.12: 100314636-100314636 on Build GRCh38
rs570771816 Chr.12: 100314751-100314751 on Build GRCh38
rs538115541 Chr.12: 100314402-100314402 on Build GRCh38
rs754030231 Chr.12: 100314606-100314606 on Build GRCh38
rs779844297 Chr.12: 100314449-100314449 on Build GRCh38
rs757281280 Chr.12: 100314611-100314611 on Build GRCh38
rs965194555 Chr.12: 100314409-100314409 on Build GRCh38
rs942608269 Chr.12: 100314732-100314732 on Build GRCh38
rs754903429 Chr.12: 100314633-100314633 on Build GRCh38
rs370957100 Chr.12: 100314455-100314455 on Build GRCh38
rs550055357 Chr.12: 100314826-100314826 on Build GRCh38
rs994683054 Chr.12: 100314412-100314412 on Build GRCh38
rs33968174 Chr.12: 100314589-100314589 on Build GRCh38
rs868827597 Chr.12: 100314872-100314872 on Build GRCh38
rs116057384 Chr.12: 100314667-100314667 on Build GRCh38
rs760026700 Chr.12: 100314558-100314558 on Build GRCh38
rs753075473 Chr.12: 100314587-100314587 on Build GRCh38
rs542398374 Chr.12: 100314593-100314593 on Build GRCh38
rs545898466 Chr.12: 100314685-100314685 on Build GRCh38
rs919326151 Chr.12: 100314807-100314807 on Build GRCh38
rs771531963 Chr.12: 100314538-100314538 on Build GRCh38
rs751466285 Chr.12: 100314629-100314629 on Build GRCh38
rs769997005 Chr.12: 100314470-100314472 on Build GRCh38
rs991732876 Chr.12: 100314735-100314735 on Build GRCh38
rs767792274 Chr.12: 100314585-100314585 on Build GRCh38
rs994007730 Chr.12: 100314782-100314782 on Build GRCh38

View Full Product Details