Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101394108-101394605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL1
Entrez Gene ID
400
Gene Name
ADP ribosylation factor like GTPase 1
Gene Aliases
ARFL1
UniGene
Hs.372616
Gene Chromosome Location
Chr.12: 101393120-101407820 on Build GRCh38
Transcripts
NM_001177.5 NM_001301068.1

SNP Detail

SNP ID Chromosome Location
rs913484616 Chr.12: 101394535-101394535 on Build GRCh38
rs989115012 Chr.12: 101394514-101394514 on Build GRCh38
rs980085108 Chr.12: 101394178-101394178 on Build GRCh38
rs934151905 Chr.12: 101394268-101394268 on Build GRCh38
rs1048104967 Chr.12: 101394361-101394361 on Build GRCh38
rs970134777 Chr.12: 101394440-101394440 on Build GRCh38
rs913432960 Chr.12: 101394226-101394226 on Build GRCh38
rs992911605 Chr.12: 101394134-101394134 on Build GRCh38
rs957666407 Chr.12: 101394189-101394189 on Build GRCh38
rs989453630 Chr.12: 101394216-101394216 on Build GRCh38
rs774220241 Chr.12: 101394463-101394463 on Build GRCh38
rs375974026 Chr.12: 101394179-101394179 on Build GRCh38
rs970081115 Chr.12: 101394141-101394141 on Build GRCh38
rs529528303 Chr.12: 101394287-101394287 on Build GRCh38
rs993851523 Chr.12: 101394248-101394248 on Build GRCh38
rs549292217 Chr.12: 101394338-101394338 on Build GRCh38
rs563618659 Chr.12: 101394241-101394241 on Build GRCh38
rs1038720242 Chr.12: 101394351-101394351 on Build GRCh38
rs749083485 Chr.12: 101394139-101394139 on Build GRCh38
rs1033041447 Chr.12: 101394471-101394471 on Build GRCh38
rs994799873 Chr.12: 101394359-101394359 on Build GRCh38
rs1014050091 Chr.12: 101394384-101394384 on Build GRCh38
rs1040667425 Chr.12: 101394201-101394201 on Build GRCh38
rs1005476932 Chr.12: 101394302-101394302 on Build GRCh38
rs911659073 Chr.12: 101394328-101394328 on Build GRCh38
rs950599147 Chr.12: 101394131-101394131 on Build GRCh38
rs565909292 Chr.12: 101394378-101394378 on Build GRCh38
rs1034714547 Chr.12: 101394308-101394308 on Build GRCh38
rs543939192 Chr.12: 101394238-101394238 on Build GRCh38
rs533316035 Chr.12: 101394148-101394148 on Build GRCh38
rs1024063241 Chr.12: 101394400-101394400 on Build GRCh38
rs978131559 Chr.12: 101394539-101394539 on Build GRCh38
rs957591439 Chr.12: 101394477-101394477 on Build GRCh38
rs936940492 Chr.12: 101394155-101394155 on Build GRCh38
rs181662068 Chr.12: 101394405-101394405 on Build GRCh38
rs898813799 Chr.12: 101394358-101394358 on Build GRCh38

View Full Product Details