Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107001161-107001686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRY1
Entrez Gene ID
1407
Gene Name
cryptochrome circadian clock 1
Gene Aliases
PHLL1
UniGene
Hs.151573
Gene Chromosome Location
Chr.12: 106991364-107093872 on Build GRCh38
Transcripts
NM_004075.4 XM_017018832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
213524 c.634G>T p.G212C Chr.12: 107001330-107001330 on Build GRCh38
5644626 c.639A>G p.G213G Chr.12: 107001325-107001325 on Build GRCh38
1743095 c.658C>A p.R220S Chr.12: 107001306-107001306 on Build GRCh38
70306 c.684A>T p.K228N Chr.12: 107001280-107001280 on Build GRCh38
5374050 c.618C>T p.S206S Chr.12: 107001346-107001346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779426662 Chr.12: 107001404-107001404 on Build GRCh38
rs751759108 Chr.12: 107001329-107001329 on Build GRCh38
rs760534685 Chr.12: 107001314-107001314 on Build GRCh38
rs781693580 Chr.12: 107001260-107001260 on Build GRCh38
rs1030436338 Chr.12: 107001424-107001424 on Build GRCh38
rs778491036 Chr.12: 107001374-107001374 on Build GRCh38
rs540975106 Chr.12: 107001215-107001215 on Build GRCh38
rs563918214 Chr.12: 107001237-107001237 on Build GRCh38
rs745345562 Chr.12: 107001306-107001306 on Build GRCh38
rs974245809 Chr.12: 107001558-107001558 on Build GRCh38
rs778111010 Chr.12: 107001286-107001286 on Build GRCh38
rs754173809 Chr.12: 107001357-107001357 on Build GRCh38
rs778579222 Chr.12: 107001305-107001305 on Build GRCh38
rs780190807 Chr.12: 107001384-107001385 on Build GRCh38
rs953580503 Chr.12: 107001641-107001641 on Build GRCh38
rs937213847 Chr.12: 107001526-107001526 on Build GRCh38
rs12814531 Chr.12: 107001371-107001371 on Build GRCh38
rs1001200138 Chr.12: 107001209-107001209 on Build GRCh38
rs753176949 Chr.12: 107001347-107001347 on Build GRCh38
rs925230788 Chr.12: 107001619-107001619 on Build GRCh38
rs776553940 Chr.12: 107001327-107001327 on Build GRCh38
rs997264286 Chr.12: 107001422-107001422 on Build GRCh38
rs770551812 Chr.12: 107001299-107001299 on Build GRCh38
rs8192438 Chr.12: 107001561-107001561 on Build GRCh38
rs887442160 Chr.12: 107001256-107001256 on Build GRCh38
rs1029537508 Chr.12: 107001638-107001639 on Build GRCh38
rs756570257 Chr.12: 107001356-107001356 on Build GRCh38
rs528988834 Chr.12: 107001452-107001452 on Build GRCh38
rs775441530 Chr.12: 107001313-107001313 on Build GRCh38
rs976686389 Chr.12: 107001321-107001321 on Build GRCh38
rs756439148 Chr.12: 107001273-107001273 on Build GRCh38
rs749714361 Chr.12: 107001298-107001298 on Build GRCh38
rs1000662935 Chr.12: 107001338-107001338 on Build GRCh38
rs746587793 Chr.12: 107001417-107001417 on Build GRCh38
rs918024572 Chr.12: 107001486-107001486 on Build GRCh38
rs773978446 Chr.12: 107001318-107001318 on Build GRCh38
rs1046010124 Chr.12: 107001504-107001504 on Build GRCh38
rs1045006814 Chr.12: 107001616-107001616 on Build GRCh38
rs563211758 Chr.12: 107001427-107001427 on Build GRCh38
rs759608414 Chr.12: 107001331-107001331 on Build GRCh38
rs373277139 Chr.12: 107001370-107001370 on Build GRCh38
rs8192439 Chr.12: 107001512-107001512 on Build GRCh38
rs766510536 Chr.12: 107001239-107001239 on Build GRCh38
rs1052298845 Chr.12: 107001650-107001650 on Build GRCh38
rs962927668 Chr.12: 107001543-107001543 on Build GRCh38
rs773879688 Chr.12: 107001315-107001315 on Build GRCh38
rs368388382 Chr.12: 107001308-107001308 on Build GRCh38
rs947984839 Chr.12: 107001277-107001277 on Build GRCh38
rs767504299 Chr.12: 107001343-107001343 on Build GRCh38
rs548664248 Chr.12: 107001481-107001481 on Build GRCh38
rs759963847 Chr.12: 107001576-107001576 on Build GRCh38
rs935348279 Chr.12: 107001425-107001425 on Build GRCh38
rs7306763 Chr.12: 107001659-107001659 on Build GRCh38
rs918742720 Chr.12: 107001568-107001568 on Build GRCh38
rs755050739 Chr.12: 107001252-107001252 on Build GRCh38
rs758182561 Chr.12: 107001260-107001271 on Build GRCh38
rs949900751 Chr.12: 107001501-107001501 on Build GRCh38
rs376932420 Chr.12: 107001517-107001517 on Build GRCh38
rs527984698 Chr.12: 107001532-107001532 on Build GRCh38
rs948046904 Chr.12: 107001611-107001611 on Build GRCh38
rs372005501 Chr.12: 107001394-107001394 on Build GRCh38
rs11836777 Chr.12: 107001638-107001638 on Build GRCh38
rs772009011 Chr.12: 107001475-107001475 on Build GRCh38
rs146142950 Chr.12: 107001570-107001570 on Build GRCh38
rs765760031 Chr.12: 107001584-107001584 on Build GRCh38
rs917925506 Chr.12: 107001598-107001598 on Build GRCh38
rs771544337 Chr.12: 107001403-107001403 on Build GRCh38
rs1027771852 Chr.12: 107001213-107001213 on Build GRCh38
rs8192440 Chr.12: 107001328-107001328 on Build GRCh38
rs953044009 Chr.12: 107001428-107001428 on Build GRCh38
rs753163872 Chr.12: 107001265-107001265 on Build GRCh38
rs757651822 Chr.12: 107001364-107001364 on Build GRCh38

View Full Product Details