Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109409592-109410114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1H
Entrez Gene ID
283446
Gene Name
myosin IH
Gene Aliases
-
UniGene
Hs.512008
Gene Chromosome Location
Chr.12: 109388277-109448371 on Build GRCh38
Transcripts
NM_001101421.3 XM_011538223.2 XM_017019207.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7332865 c.1231G>C p.E411Q Chr.12: 109410018-109410018 on Build GRCh38
3416448 c.1191C>A p.V397V Chr.12: 109409610-109409610 on Build GRCh38
5781066 c.1277C>T p.A426V Chr.12: 109410034-109410034 on Build GRCh38
5796485 c.1201A>C p.N401H Chr.12: 109409620-109409620 on Build GRCh38
4522610 c.1170G>A p.K390K Chr.12: 109409619-109409619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147351824 Chr.12: 109409705-109409705 on Build GRCh38
rs957803715 Chr.12: 109409734-109409734 on Build GRCh38
rs949047709 Chr.12: 109409915-109409915 on Build GRCh38
rs772787662 Chr.12: 109409629-109409629 on Build GRCh38
rs759463407 Chr.12: 109410039-109410039 on Build GRCh38
rs774598989 Chr.12: 109409692-109409692 on Build GRCh38
rs976869978 Chr.12: 109409829-109409829 on Build GRCh38
rs948045016 Chr.12: 109409977-109409977 on Build GRCh38
rs999632729 Chr.12: 109409696-109409696 on Build GRCh38
rs199676779 Chr.12: 109410076-109410076 on Build GRCh38
rs760609005 Chr.12: 109409652-109409652 on Build GRCh38
rs911793118 Chr.12: 109409816-109409816 on Build GRCh38
rs895388482 Chr.12: 109409667-109409667 on Build GRCh38
rs918672783 Chr.12: 109409887-109409887 on Build GRCh38
rs762067988 Chr.12: 109409697-109409697 on Build GRCh38
rs1031095079 Chr.12: 109410065-109410065 on Build GRCh38
rs757405609 Chr.12: 109409650-109409650 on Build GRCh38
rs561198272 Chr.12: 109409668-109409668 on Build GRCh38
rs370874642 Chr.12: 109409636-109409636 on Build GRCh38
rs748533190 Chr.12: 109409970-109409970 on Build GRCh38
rs368224660 Chr.12: 109410018-109410018 on Build GRCh38
rs1015974265 Chr.12: 109409790-109409790 on Build GRCh38
rs767277416 Chr.12: 109409648-109409648 on Build GRCh38
rs954215474 Chr.12: 109409860-109409860 on Build GRCh38
rs376133892 Chr.12: 109410017-109410017 on Build GRCh38
rs1037140623 Chr.12: 109409628-109409628 on Build GRCh38
rs578088715 Chr.12: 109410028-109410028 on Build GRCh38
rs987412913 Chr.12: 109409811-109409811 on Build GRCh38
rs895117434 Chr.12: 109409926-109409926 on Build GRCh38
rs144350451 Chr.12: 109409681-109409681 on Build GRCh38
rs999685207 Chr.12: 109410060-109410060 on Build GRCh38
rs776792338 Chr.12: 109409951-109409951 on Build GRCh38
rs775284070 Chr.12: 109409907-109409908 on Build GRCh38
rs764985746 Chr.12: 109409625-109409625 on Build GRCh38
rs367858797 Chr.12: 109409925-109409925 on Build GRCh38
rs185560862 Chr.12: 109409786-109409786 on Build GRCh38
rs970900679 Chr.12: 109409695-109409695 on Build GRCh38
rs572430432 Chr.12: 109409633-109409633 on Build GRCh38
rs562696389 Chr.12: 109409740-109409740 on Build GRCh38
rs1469944 Chr.12: 109409782-109409782 on Build GRCh38
rs566443390 Chr.12: 109409787-109409787 on Build GRCh38
rs145037961 Chr.12: 109409900-109409900 on Build GRCh38
rs926237275 Chr.12: 109409885-109409885 on Build GRCh38
rs1001978249 Chr.12: 109409725-109409725 on Build GRCh38
rs1036188640 Chr.12: 109409867-109409867 on Build GRCh38
rs754782723 Chr.12: 109409638-109409638 on Build GRCh38
rs139405649 Chr.12: 109409859-109409859 on Build GRCh38
rs766504817 Chr.12: 109409983-109409983 on Build GRCh38
rs745901899 Chr.12: 109409662-109409662 on Build GRCh38
rs537935903 Chr.12: 109409967-109409967 on Build GRCh38
rs989559652 Chr.12: 109409757-109409757 on Build GRCh38
rs55913842 Chr.12: 109409982-109409982 on Build GRCh38
rs1012475526 Chr.12: 109409680-109409680 on Build GRCh38
rs754090304 Chr.12: 109409649-109409649 on Build GRCh38
rs181664035 Chr.12: 109409637-109409637 on Build GRCh38
rs774335585 Chr.12: 109410032-109410032 on Build GRCh38
rs758506327 Chr.12: 109409663-109409663 on Build GRCh38
rs979313797 Chr.12: 109409882-109409882 on Build GRCh38
rs1048669275 Chr.12: 109410011-109410011 on Build GRCh38
rs1004579640 Chr.12: 109410044-109410044 on Build GRCh38
rs747525287 Chr.12: 109409671-109409671 on Build GRCh38
rs755454821 Chr.12: 109409837-109409837 on Build GRCh38
rs965370857 Chr.12: 109409819-109409819 on Build GRCh38
rs779120914 Chr.12: 109409660-109409660 on Build GRCh38

View Full Product Details