Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109483389-109483903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE3B
Entrez Gene ID
89910
Gene Name
ubiquitin protein ligase E3B
Gene Aliases
BPIDS, KOS
UniGene
Hs.374067
Gene Chromosome Location
Chr.12: 109477410-109543628 on Build GRCh38
Transcripts
NM_001270449.1 NM_001270450.1 NM_001270451.1 NM_130466.3 NM_183415.2 XM_005253987.2 XM_006719681.3 XM_006719682.2 XM_011538959.2 XM_011538961.1 XM_017020195.1 XM_017020196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415632 c.50G>A p.R17H Chr.12: 109483601-109483601 on Build GRCh38
7118283 c.16C>G p.Q6E Chr.12: 109483567-109483567 on Build GRCh38
7118284 c.16C>G p.Q6E Chr.12: 109483567-109483567 on Build GRCh38
7153062 c.24G>A p.S8S Chr.12: 109483575-109483575 on Build GRCh38
5516629 c.50G>A p.R17H Chr.12: 109483601-109483601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372604916 Chr.12: 109483637-109483637 on Build GRCh38
rs868259069 Chr.12: 109483775-109483775 on Build GRCh38
rs1003724603 Chr.12: 109483771-109483771 on Build GRCh38
rs766274696 Chr.12: 109483642-109483642 on Build GRCh38
rs777400615 Chr.12: 109483859-109483859 on Build GRCh38
rs770693481 Chr.12: 109483443-109483443 on Build GRCh38
rs773989312 Chr.12: 109483720-109483720 on Build GRCh38
rs764278999 Chr.12: 109483666-109483666 on Build GRCh38
rs199720082 Chr.12: 109483789-109483790 on Build GRCh38
rs758877442 Chr.12: 109483745-109483745 on Build GRCh38
rs779284561 Chr.12: 109483582-109483582 on Build GRCh38
rs908966964 Chr.12: 109483819-109483819 on Build GRCh38
rs770955936 Chr.12: 109483873-109483873 on Build GRCh38
rs761850046 Chr.12: 109483614-109483614 on Build GRCh38
rs928669809 Chr.12: 109483490-109483490 on Build GRCh38
rs780649712 Chr.12: 109483853-109483853 on Build GRCh38
rs753969069 Chr.12: 109483727-109483727 on Build GRCh38
rs762970554 Chr.12: 109483706-109483706 on Build GRCh38
rs747561690 Chr.12: 109483698-109483698 on Build GRCh38
rs147402736 Chr.12: 109483610-109483610 on Build GRCh38
rs774229226 Chr.12: 109483875-109483875 on Build GRCh38
rs879071788 Chr.12: 109483493-109483493 on Build GRCh38
rs144106525 Chr.12: 109483701-109483701 on Build GRCh38
rs149953168 Chr.12: 109483687-109483687 on Build GRCh38
rs148696226 Chr.12: 109483877-109483877 on Build GRCh38
rs746722966 Chr.12: 109483693-109483693 on Build GRCh38
rs370576457 Chr.12: 109483827-109483827 on Build GRCh38
rs868830826 Chr.12: 109483412-109483412 on Build GRCh38
rs376752676 Chr.12: 109483683-109483683 on Build GRCh38
rs768162705 Chr.12: 109483694-109483694 on Build GRCh38
rs765294910 Chr.12: 109483730-109483730 on Build GRCh38
rs200114053 Chr.12: 109483663-109483663 on Build GRCh38
rs201620860 Chr.12: 109483673-109483673 on Build GRCh38
rs780507038 Chr.12: 109483812-109483812 on Build GRCh38
rs753826150 Chr.12: 109483667-109483667 on Build GRCh38
rs368120913 Chr.12: 109483738-109483738 on Build GRCh38
rs574539443 Chr.12: 109483572-109483572 on Build GRCh38
rs561861822 Chr.12: 109483790-109483790 on Build GRCh38
rs776234508 Chr.12: 109483696-109483696 on Build GRCh38
rs757679020 Chr.12: 109483575-109483575 on Build GRCh38
rs747834266 Chr.12: 109483855-109483855 on Build GRCh38
rs745562218 Chr.12: 109483501-109483501 on Build GRCh38
rs778170508 Chr.12: 109483507-109483507 on Build GRCh38
rs947630776 Chr.12: 109483565-109483565 on Build GRCh38
rs539907300 Chr.12: 109483769-109483769 on Build GRCh38
rs749669361 Chr.12: 109483542-109483553 on Build GRCh38
rs991276875 Chr.12: 109483635-109483635 on Build GRCh38
rs139172208 Chr.12: 109483643-109483643 on Build GRCh38
rs775770252 Chr.12: 109483734-109483734 on Build GRCh38
rs141528351 Chr.12: 109483590-109483590 on Build GRCh38
rs913478208 Chr.12: 109483550-109483550 on Build GRCh38
rs555841802 Chr.12: 109483492-109483492 on Build GRCh38
rs556951575 Chr.12: 109483600-109483600 on Build GRCh38
rs771722819 Chr.12: 109483591-109483591 on Build GRCh38
rs2302702 Chr.12: 109483453-109483453 on Build GRCh38
rs375632180 Chr.12: 109483638-109483638 on Build GRCh38
rs528095531 Chr.12: 109483725-109483725 on Build GRCh38
rs1023414854 Chr.12: 109483428-109483428 on Build GRCh38
rs749860136 Chr.12: 109483574-109483574 on Build GRCh38
rs1043652436 Chr.12: 109483581-109483581 on Build GRCh38
rs775981553 Chr.12: 109483612-109483612 on Build GRCh38
rs672601304 Chr.12: 109483552-109483552 on Build GRCh38
rs142757108 Chr.12: 109483559-109483559 on Build GRCh38
rs942863566 Chr.12: 109483862-109483862 on Build GRCh38
rs960045350 Chr.12: 109483497-109483497 on Build GRCh38
rs1035758486 Chr.12: 109483719-109483719 on Build GRCh38
rs752830444 Chr.12: 109483507-109483507 on Build GRCh38
rs981792778 Chr.12: 109483532-109483532 on Build GRCh38
rs200272943 Chr.12: 109483869-109483869 on Build GRCh38
rs148340310 Chr.12: 109483433-109483433 on Build GRCh38
rs756283868 Chr.12: 109483558-109483558 on Build GRCh38
rs980135888 Chr.12: 109483462-109483462 on Build GRCh38
rs757786448 Chr.12: 109483672-109483672 on Build GRCh38
rs529443721 Chr.12: 109483840-109483840 on Build GRCh38
rs746633699 Chr.12: 109483601-109483601 on Build GRCh38
rs769489667 Chr.12: 109483856-109483856 on Build GRCh38
rs773351206 Chr.12: 109483705-109483705 on Build GRCh38
rs765224973 Chr.12: 109483636-109483636 on Build GRCh38
rs769829361 Chr.12: 109483504-109483504 on Build GRCh38
rs760993283 Chr.12: 109483724-109483724 on Build GRCh38
rs979019682 Chr.12: 109483830-109483830 on Build GRCh38
rs911446500 Chr.12: 109483839-109483839 on Build GRCh38
rs780380636 Chr.12: 109483688-109483688 on Build GRCh38
rs770898948 Chr.12: 109483709-109483709 on Build GRCh38

View Full Product Details