Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110649006-110649518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HVCN1
Entrez Gene ID
84329
Gene Name
hydrogen voltage gated channel 1
Gene Aliases
HV1, VSOP
UniGene
Hs.211511
Gene Chromosome Location
Chr.12: 110648686-110704945 on Build GRCh38
Transcripts
NM_001040107.1 NM_001256413.1 NM_032369.3 XM_005253948.2 XM_011538838.1 XM_011538839.2 XM_011538840.2 XM_011538841.2 XM_011538842.2 XM_011538844.2 XM_011538845.1 XM_011538846.2 XM_011538847.1 XM_017020026.1 XM_017020027.1 NM_001082537.2 NM_001082538.2 NM_001173975.2 NM_001173976.1 NM_001319680.1 NM_001319681.1 NM_024549.5 NR_135088.1 XM_005253934.3 XM_005253935.3 XM_006719594.2 XM_006719595.2 XM_006719596.2 XM_006719597.3 XM_006719598.2 XM_006719599.2 XM_006719600.2 XM_011538733.2 XM_011538734.2 XM_011538735.1 XM_011538737.2 XM_011538738.2 XM_017019964.1 XM_017019965.1 XM_017019966.1 XM_017019967.1 XM_017019968.1 XM_017019969.1

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
UniGene
Hs.211511
Gene Chromosome Location
Chr.12: 110614027-110649130 on Build GRCh38
Transcripts
NM_001040107.1 NM_001256413.1 NM_032369.3 XM_005253948.2 XM_011538838.1 XM_011538839.2 XM_011538840.2 XM_011538841.2 XM_011538842.2 XM_011538844.2 XM_011538845.1 XM_011538846.2 XM_011538847.1 XM_017020026.1 XM_017020027.1 NM_001082537.2 NM_001082538.2 NM_001173975.2 NM_001173976.1 NM_001319680.1 NM_001319681.1 NM_024549.5 NR_135088.1 XM_005253934.3 XM_005253935.3 XM_006719594.2 XM_006719595.2 XM_006719596.2 XM_006719597.3 XM_006719598.2 XM_006719599.2 XM_006719600.2 XM_011538733.2 XM_011538734.2 XM_011538735.1 XM_011538737.2 XM_011538738.2 XM_017019964.1 XM_017019965.1 XM_017019966.1 XM_017019967.1 XM_017019968.1 XM_017019969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4969978 c.791G>A p.R264Q Chr.12: 110649441-110649441 on Build GRCh38
935004 c.790C>T p.R264* Chr.12: 110649442-110649442 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959194524 Chr.12: 110649075-110649075 on Build GRCh38
rs952907210 Chr.12: 110649179-110649179 on Build GRCh38
rs758320930 Chr.12: 110649489-110649489 on Build GRCh38
rs917811869 Chr.12: 110649099-110649099 on Build GRCh38
rs146639508 Chr.12: 110649081-110649081 on Build GRCh38
rs372842031 Chr.12: 110649310-110649310 on Build GRCh38
rs770046019 Chr.12: 110649370-110649370 on Build GRCh38
rs560887298 Chr.12: 110649326-110649326 on Build GRCh38
rs1025211445 Chr.12: 110649482-110649482 on Build GRCh38
rs749423923 Chr.12: 110649405-110649405 on Build GRCh38
rs747513404 Chr.12: 110649450-110649452 on Build GRCh38
rs945272130 Chr.12: 110649100-110649100 on Build GRCh38
rs1044532 Chr.12: 110649127-110649127 on Build GRCh38
rs759990996 Chr.12: 110649408-110649408 on Build GRCh38
rs780747069 Chr.12: 110649447-110649447 on Build GRCh38
rs983453436 Chr.12: 110649253-110649253 on Build GRCh38
rs1030803292 Chr.12: 110649031-110649032 on Build GRCh38
rs779738070 Chr.12: 110649361-110649361 on Build GRCh38
rs762965470 Chr.12: 110649118-110649118 on Build GRCh38
rs749901454 Chr.12: 110649451-110649451 on Build GRCh38
rs769510420 Chr.12: 110649338-110649338 on Build GRCh38
rs1042394807 Chr.12: 110649139-110649139 on Build GRCh38
rs193070681 Chr.12: 110649044-110649044 on Build GRCh38
rs775765211 Chr.12: 110649421-110649421 on Build GRCh38
rs1015301752 Chr.12: 110649336-110649336 on Build GRCh38
rs764873661 Chr.12: 110649442-110649442 on Build GRCh38
rs992502177 Chr.12: 110649094-110649094 on Build GRCh38
rs141396411 Chr.12: 110649327-110649327 on Build GRCh38
rs936794175 Chr.12: 110649261-110649261 on Build GRCh38
rs746773382 Chr.12: 110649406-110649406 on Build GRCh38
rs767677920 Chr.12: 110649426-110649426 on Build GRCh38
rs529752221 Chr.12: 110649489-110649489 on Build GRCh38
rs1052460927 Chr.12: 110649281-110649281 on Build GRCh38
rs778961622 Chr.12: 110649169-110649170 on Build GRCh38
rs746802313 Chr.12: 110649365-110649365 on Build GRCh38
rs759719780 Chr.12: 110649345-110649345 on Build GRCh38
rs757517445 Chr.12: 110649167-110649168 on Build GRCh38
rs754118625 Chr.12: 110649119-110649120 on Build GRCh38
rs753559656 Chr.12: 110649439-110649439 on Build GRCh38
rs758384970 Chr.12: 110649377-110649378 on Build GRCh38
rs969034689 Chr.12: 110649122-110649122 on Build GRCh38
rs10462 Chr.12: 110649334-110649334 on Build GRCh38
rs371442260 Chr.12: 110649401-110649401 on Build GRCh38
rs925286074 Chr.12: 110649238-110649238 on Build GRCh38
rs753467272 Chr.12: 110649043-110649043 on Build GRCh38
rs745980562 Chr.12: 110649171-110649172 on Build GRCh38
rs997672041 Chr.12: 110649280-110649280 on Build GRCh38
rs758319414 Chr.12: 110649178-110649178 on Build GRCh38
rs763898292 Chr.12: 110649427-110649427 on Build GRCh38
rs980473501 Chr.12: 110649147-110649147 on Build GRCh38
rs772581863 Chr.12: 110649409-110649409 on Build GRCh38
rs959478757 Chr.12: 110649435-110649435 on Build GRCh38
rs962491605 Chr.12: 110649032-110649032 on Build GRCh38
rs769265033 Chr.12: 110649073-110649073 on Build GRCh38
rs900671871 Chr.12: 110649276-110649276 on Build GRCh38
rs919595182 Chr.12: 110649160-110649160 on Build GRCh38
rs763873061 Chr.12: 110649091-110649092 on Build GRCh38
rs567316596 Chr.12: 110649256-110649256 on Build GRCh38
rs112985685 Chr.12: 110649201-110649201 on Build GRCh38
rs537432372 Chr.12: 110649269-110649269 on Build GRCh38
rs202004657 Chr.12: 110649441-110649441 on Build GRCh38
rs962337150 Chr.12: 110649354-110649354 on Build GRCh38
rs1025173695 Chr.12: 110649097-110649097 on Build GRCh38
rs926457064 Chr.12: 110649051-110649051 on Build GRCh38
rs185774837 Chr.12: 110649076-110649076 on Build GRCh38
rs778511008 Chr.12: 110649173-110649173 on Build GRCh38
rs750334834 Chr.12: 110649176-110649176 on Build GRCh38
rs774514420 Chr.12: 110649344-110649344 on Build GRCh38
rs1062830 Chr.12: 110649128-110649128 on Build GRCh38

View Full Product Details