Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111729860-111730400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
-
UniGene
Hs.331141
Gene Chromosome Location
Chr.12: 111686053-111757107 on Build GRCh38
Transcripts
NM_001136538.1 NM_025247.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259282 c.1332C>T p.I444I Chr.12: 111729894-111729894 on Build GRCh38
4466459 c.1343C>T p.P448L Chr.12: 111729905-111729905 on Build GRCh38
7353862 c.1480G>A p.D494N Chr.12: 111729949-111729949 on Build GRCh38
1241957 c.1349A>G p.H450R Chr.12: 111729911-111729911 on Build GRCh38
6560763 c.1401C>T p.T467T Chr.12: 111729870-111729870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs640783 Chr.12: 111730246-111730246 on Build GRCh38
rs540444011 Chr.12: 111729894-111729894 on Build GRCh38
rs531670349 Chr.12: 111730233-111730233 on Build GRCh38
rs1046621817 Chr.12: 111730137-111730137 on Build GRCh38
rs952428586 Chr.12: 111730381-111730381 on Build GRCh38
rs763424702 Chr.12: 111730107-111730107 on Build GRCh38
rs756511585 Chr.12: 111729956-111729956 on Build GRCh38
rs1029989645 Chr.12: 111729930-111729930 on Build GRCh38
rs990478969 Chr.12: 111730071-111730071 on Build GRCh38
rs772605037 Chr.12: 111729903-111729904 on Build GRCh38
rs1026855855 Chr.12: 111730366-111730366 on Build GRCh38
rs11066015 Chr.12: 111730205-111730205 on Build GRCh38
rs893890871 Chr.12: 111730204-111730204 on Build GRCh38
rs370592166 Chr.12: 111729905-111729905 on Build GRCh38
rs765183120 Chr.12: 111730336-111730336 on Build GRCh38
rs151106801 Chr.12: 111729911-111729911 on Build GRCh38
rs1042587090 Chr.12: 111730099-111730099 on Build GRCh38
rs116960333 Chr.12: 111730267-111730267 on Build GRCh38
rs916280288 Chr.12: 111730079-111730079 on Build GRCh38
rs906768074 Chr.12: 111730113-111730113 on Build GRCh38
rs890484805 Chr.12: 111730227-111730227 on Build GRCh38
rs527338880 Chr.12: 111730124-111730124 on Build GRCh38
rs939635239 Chr.12: 111730167-111730167 on Build GRCh38
rs142933086 Chr.12: 111729901-111729901 on Build GRCh38
rs758685679 Chr.12: 111730005-111730005 on Build GRCh38
rs369143377 Chr.12: 111730003-111730003 on Build GRCh38
rs574837934 Chr.12: 111730159-111730159 on Build GRCh38
rs145027535 Chr.12: 111729933-111729933 on Build GRCh38
rs779062320 Chr.12: 111729998-111729998 on Build GRCh38
rs141115291 Chr.12: 111729928-111729928 on Build GRCh38
rs557698280 Chr.12: 111730355-111730355 on Build GRCh38
rs994387149 Chr.12: 111730243-111730243 on Build GRCh38
rs760755071 Chr.12: 111730131-111730131 on Build GRCh38
rs996095774 Chr.12: 111729946-111729946 on Build GRCh38
rs371183790 Chr.12: 111729903-111729903 on Build GRCh38
rs753148624 Chr.12: 111729900-111729900 on Build GRCh38
rs1055104684 Chr.12: 111730178-111730178 on Build GRCh38
rs745894413 Chr.12: 111729885-111729890 on Build GRCh38
rs955340956 Chr.12: 111729937-111729937 on Build GRCh38
rs568937194 Chr.12: 111730264-111730264 on Build GRCh38
rs1005604136 Chr.12: 111730205-111730207 on Build GRCh38
rs780460902 Chr.12: 111730007-111730007 on Build GRCh38
rs754393019 Chr.12: 111729972-111729972 on Build GRCh38
rs75063348 Chr.12: 111729973-111729973 on Build GRCh38
rs952442378 Chr.12: 111730070-111730070 on Build GRCh38
rs935479636 Chr.12: 111730168-111730168 on Build GRCh38
rs549163998 Chr.12: 111730148-111730148 on Build GRCh38
rs370489200 Chr.12: 111729945-111729945 on Build GRCh38
rs753056489 Chr.12: 111729952-111729952 on Build GRCh38
rs199645118 Chr.12: 111729959-111729959 on Build GRCh38
rs1050372537 Chr.12: 111730208-111730208 on Build GRCh38
rs746104743 Chr.12: 111729914-111729914 on Build GRCh38
rs772676900 Chr.12: 111729938-111729938 on Build GRCh38
rs924201728 Chr.12: 111730144-111730144 on Build GRCh38
rs368642732 Chr.12: 111729920-111729920 on Build GRCh38
rs761176157 Chr.12: 111729932-111729932 on Build GRCh38
rs868336030 Chr.12: 111730152-111730152 on Build GRCh38
rs1019848781 Chr.12: 111729988-111729988 on Build GRCh38
rs771935027 Chr.12: 111729916-111729916 on Build GRCh38
rs532943669 Chr.12: 111729927-111729927 on Build GRCh38
rs148547215 Chr.12: 111729895-111729895 on Build GRCh38
rs375729084 Chr.12: 111729919-111729919 on Build GRCh38
rs1053884779 Chr.12: 111730193-111730193 on Build GRCh38
rs202003128 Chr.12: 111729906-111729906 on Build GRCh38
rs972989299 Chr.12: 111730050-111730050 on Build GRCh38
rs919639553 Chr.12: 111730057-111730057 on Build GRCh38
rs36046440 Chr.12: 111729949-111729949 on Build GRCh38

View Full Product Details