Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112140740-112141246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
UniGene
Hs.5148
Gene Chromosome Location
Chr.12: 112125545-112153604 on Build GRCh38
Transcripts
NM_001143906.1 NM_006700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
935160 c.370C>T p.L124F Chr.12: 112140951-112140951 on Build GRCh38
935159 c.362G>A p.R121H Chr.12: 112140943-112140943 on Build GRCh38
935161 c.399A>T p.E133D Chr.12: 112140980-112140980 on Build GRCh38
223598 c.596C>T p.T199I Chr.12: 112141177-112141177 on Build GRCh38
6135812 c.267C>T p.V89V Chr.12: 112140848-112140848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745723924 Chr.12: 112140785-112140785 on Build GRCh38
rs776240723 Chr.12: 112140861-112140861 on Build GRCh38
rs764727940 Chr.12: 112140996-112140996 on Build GRCh38
rs201271111 Chr.12: 112140837-112140837 on Build GRCh38
rs748908383 Chr.12: 112141113-112141113 on Build GRCh38
rs772866825 Chr.12: 112140985-112140985 on Build GRCh38
rs775821002 Chr.12: 112141126-112141126 on Build GRCh38
rs757415144 Chr.12: 112140927-112140927 on Build GRCh38
rs148272299 Chr.12: 112141122-112141122 on Build GRCh38
rs141135769 Chr.12: 112140915-112140915 on Build GRCh38
rs772458417 Chr.12: 112140989-112140989 on Build GRCh38
rs377133781 Chr.12: 112141020-112141020 on Build GRCh38
rs770479612 Chr.12: 112141200-112141200 on Build GRCh38
rs759901354 Chr.12: 112140914-112140914 on Build GRCh38
rs748857029 Chr.12: 112141169-112141169 on Build GRCh38
rs769372382 Chr.12: 112141086-112141086 on Build GRCh38
rs201656998 Chr.12: 112141016-112141016 on Build GRCh38
rs754869905 Chr.12: 112141045-112141045 on Build GRCh38
rs780097177 Chr.12: 112140943-112140943 on Build GRCh38
rs935865205 Chr.12: 112140983-112140983 on Build GRCh38
rs543344272 Chr.12: 112140808-112140808 on Build GRCh38
rs748662598 Chr.12: 112141196-112141196 on Build GRCh38
rs940198378 Chr.12: 112140810-112140810 on Build GRCh38
rs751336035 Chr.12: 112141144-112141144 on Build GRCh38
rs746910254 Chr.12: 112140963-112140963 on Build GRCh38
rs781196455 Chr.12: 112141071-112141071 on Build GRCh38
rs750585254 Chr.12: 112141018-112141018 on Build GRCh38
rs747403771 Chr.12: 112141127-112141127 on Build GRCh38
rs770621242 Chr.12: 112141118-112141118 on Build GRCh38
rs370443542 Chr.12: 112141078-112141078 on Build GRCh38
rs778548385 Chr.12: 112141177-112141177 on Build GRCh38
rs772065947 Chr.12: 112140815-112140815 on Build GRCh38
rs764050238 Chr.12: 112140926-112140926 on Build GRCh38
rs374469864 Chr.12: 112141137-112141137 on Build GRCh38
rs773469802 Chr.12: 112141134-112141134 on Build GRCh38
rs780551409 Chr.12: 112140978-112140978 on Build GRCh38
rs754817701 Chr.12: 112141155-112141155 on Build GRCh38
rs767312226 Chr.12: 112141157-112141157 on Build GRCh38
rs201843124 Chr.12: 112141141-112141141 on Build GRCh38
rs199998043 Chr.12: 112140934-112140934 on Build GRCh38
rs114172135 Chr.12: 112141017-112141017 on Build GRCh38
rs768079263 Chr.12: 112140838-112140838 on Build GRCh38
rs769268573 Chr.12: 112140982-112140982 on Build GRCh38
rs765372475 Chr.12: 112141130-112141130 on Build GRCh38
rs147594088 Chr.12: 112140981-112140981 on Build GRCh38
rs769552037 Chr.12: 112140895-112140895 on Build GRCh38
rs774578898 Chr.12: 112140905-112140905 on Build GRCh38
rs751454794 Chr.12: 112141023-112141023 on Build GRCh38
rs777245645 Chr.12: 112141168-112141168 on Build GRCh38
rs1033939855 Chr.12: 112141031-112141031 on Build GRCh38
rs778558952 Chr.12: 112140773-112140773 on Build GRCh38
rs934934115 Chr.12: 112141007-112141007 on Build GRCh38
rs758289708 Chr.12: 112140942-112140942 on Build GRCh38
rs777364668 Chr.12: 112141104-112141104 on Build GRCh38
rs1019494120 Chr.12: 112141171-112141171 on Build GRCh38
rs753793501 Chr.12: 112141009-112141009 on Build GRCh38
rs756835810 Chr.12: 112141176-112141176 on Build GRCh38
rs761345734 Chr.12: 112140873-112140873 on Build GRCh38
rs768683678 Chr.12: 112140974-112140974 on Build GRCh38
rs753226462 Chr.12: 112140919-112140919 on Build GRCh38
rs113339178 Chr.12: 112141129-112141129 on Build GRCh38
rs776896827 Chr.12: 112141190-112141190 on Build GRCh38
rs777216818 Chr.12: 112141128-112141128 on Build GRCh38
rs752714882 Chr.12: 112141165-112141165 on Build GRCh38
rs372425567 Chr.12: 112140830-112140830 on Build GRCh38
rs759846308 Chr.12: 112140987-112140987 on Build GRCh38
rs747859333 Chr.12: 112141112-112141112 on Build GRCh38
rs146944659 Chr.12: 112140953-112140953 on Build GRCh38
rs916051791 Chr.12: 112140898-112140898 on Build GRCh38
rs114591514 Chr.12: 112140920-112140920 on Build GRCh38
rs1017175071 Chr.12: 112141181-112141181 on Build GRCh38
rs149081968 Chr.12: 112140995-112140995 on Build GRCh38
rs910000455 Chr.12: 112140807-112140807 on Build GRCh38
rs775005660 Chr.12: 112140989-112140990 on Build GRCh38
rs756046542 Chr.12: 112141166-112141166 on Build GRCh38
rs769094645 Chr.12: 112141187-112141187 on Build GRCh38

View Full Product Details