Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112869617-112870113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
-
UniGene
Hs.21239
Gene Chromosome Location
Chr.12: 112791538-112898881 on Build GRCh38
Transcripts
NM_001143854.1 NM_014954.3 XM_011538048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1948792 c.617G>C p.S206T Chr.12: 112869765-112869765 on Build GRCh38
5525531 c.735C>T p.S245S Chr.12: 112869978-112869978 on Build GRCh38
7238347 c.737G>T p.R246L Chr.12: 112869980-112869980 on Build GRCh38
4722577 c.677G>A p.R226Q Chr.12: 112869920-112869920 on Build GRCh38
3456272 c.785G>A p.R262Q Chr.12: 112870028-112870028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747582874 Chr.12: 112870039-112870039 on Build GRCh38
rs1023009461 Chr.12: 112869687-112869687 on Build GRCh38
rs993655050 Chr.12: 112870009-112870009 on Build GRCh38
rs180930503 Chr.12: 112869885-112869885 on Build GRCh38
rs747671742 Chr.12: 112870020-112870020 on Build GRCh38
rs763311733 Chr.12: 112869809-112869809 on Build GRCh38
rs986628159 Chr.12: 112869974-112869974 on Build GRCh38
rs768230622 Chr.12: 112869901-112869901 on Build GRCh38
rs774717873 Chr.12: 112869915-112869915 on Build GRCh38
rs750921318 Chr.12: 112869728-112869728 on Build GRCh38
rs1023169526 Chr.12: 112869665-112869665 on Build GRCh38
rs374869588 Chr.12: 112869935-112869935 on Build GRCh38
rs147818920 Chr.12: 112870007-112870007 on Build GRCh38
rs746535322 Chr.12: 112869897-112869897 on Build GRCh38
rs145114678 Chr.12: 112869943-112869943 on Build GRCh38
rs757147809 Chr.12: 112869873-112869873 on Build GRCh38
rs746199452 Chr.12: 112870037-112870037 on Build GRCh38
rs576092129 Chr.12: 112869783-112869783 on Build GRCh38
rs368311022 Chr.12: 112870028-112870028 on Build GRCh38
rs140680955 Chr.12: 112869955-112869955 on Build GRCh38
rs376018956 Chr.12: 112869987-112869987 on Build GRCh38
rs201228672 Chr.12: 112869765-112869765 on Build GRCh38
rs760818180 Chr.12: 112869979-112869979 on Build GRCh38
rs1008168752 Chr.12: 112869703-112869703 on Build GRCh38
rs752762635 Chr.12: 112869919-112869919 on Build GRCh38
rs144758251 Chr.12: 112869928-112869928 on Build GRCh38
rs577513589 Chr.12: 112869876-112869876 on Build GRCh38
rs754482274 Chr.12: 112870008-112870008 on Build GRCh38
rs754983272 Chr.12: 112869834-112869834 on Build GRCh38
rs144656378 Chr.12: 112869965-112869965 on Build GRCh38
rs758157549 Chr.12: 112870003-112870003 on Build GRCh38
rs147898060 Chr.12: 112869980-112869980 on Build GRCh38
rs755114928 Chr.12: 112869737-112869737 on Build GRCh38
rs779668181 Chr.12: 112869894-112869894 on Build GRCh38
rs779991033 Chr.12: 112870006-112870006 on Build GRCh38
rs375588972 Chr.12: 112870000-112870000 on Build GRCh38
rs970014439 Chr.12: 112869825-112869825 on Build GRCh38
rs563388762 Chr.12: 112869999-112869999 on Build GRCh38
rs867127827 Chr.12: 112869898-112869898 on Build GRCh38
rs780740367 Chr.12: 112870013-112870013 on Build GRCh38
rs934676445 Chr.12: 112869798-112869798 on Build GRCh38
rs374904981 Chr.12: 112869800-112869800 on Build GRCh38
rs141246952 Chr.12: 112869781-112869781 on Build GRCh38
rs768175767 Chr.12: 112869950-112869950 on Build GRCh38
rs745314704 Chr.12: 112869791-112869791 on Build GRCh38
rs757143055 Chr.12: 112869859-112869859 on Build GRCh38
rs570689092 Chr.12: 112869672-112869672 on Build GRCh38
rs776294551 Chr.12: 112869906-112869906 on Build GRCh38
rs772596187 Chr.12: 112869962-112869962 on Build GRCh38
rs762149756 Chr.12: 112869984-112869984 on Build GRCh38
rs765626804 Chr.12: 112869714-112869714 on Build GRCh38
rs747865237 Chr.12: 112869954-112869954 on Build GRCh38
rs765569272 Chr.12: 112869801-112869801 on Build GRCh38
rs748443926 Chr.12: 112869753-112869753 on Build GRCh38
rs371895375 Chr.12: 112869784-112869784 on Build GRCh38
rs761799523 Chr.12: 112870067-112870067 on Build GRCh38
rs758294722 Chr.12: 112869940-112869940 on Build GRCh38
rs774971449 Chr.12: 112869797-112869797 on Build GRCh38
rs367990066 Chr.12: 112869734-112869734 on Build GRCh38
rs774721328 Chr.12: 112869957-112869957 on Build GRCh38
rs777769955 Chr.12: 112869775-112869775 on Build GRCh38
rs776790418 Chr.12: 112870053-112870053 on Build GRCh38
rs551182268 Chr.12: 112869736-112869736 on Build GRCh38
rs149945171 Chr.12: 112869939-112869939 on Build GRCh38
rs201634592 Chr.12: 112869874-112869874 on Build GRCh38
rs780578050 Chr.12: 112869920-112869920 on Build GRCh38
rs755969538 Chr.12: 112869772-112869772 on Build GRCh38
rs373196382 Chr.12: 112869964-112869964 on Build GRCh38
rs762785433 Chr.12: 112870087-112870087 on Build GRCh38
rs754165001 Chr.12: 112869929-112869929 on Build GRCh38
rs552941735 Chr.12: 112869702-112869702 on Build GRCh38
rs201082152 Chr.12: 112869983-112869983 on Build GRCh38
rs771368523 Chr.12: 112869913-112869913 on Build GRCh38
rs374612736 Chr.12: 112869729-112869729 on Build GRCh38
rs773328921 Chr.12: 112870083-112870083 on Build GRCh38
rs745320677 Chr.12: 112869961-112869961 on Build GRCh38
rs535256359 Chr.12: 112869751-112869751 on Build GRCh38
rs914617955 Chr.12: 112870022-112870022 on Build GRCh38
rs773507762 Chr.12: 112869803-112869803 on Build GRCh38
rs200826385 Chr.12: 112869754-112869754 on Build GRCh38
rs371775080 Chr.12: 112869933-112869933 on Build GRCh38
rs763252983 Chr.12: 112869912-112869912 on Build GRCh38
rs199945677 Chr.12: 112869852-112869852 on Build GRCh38
rs746281742 Chr.12: 112869853-112869853 on Build GRCh38
rs766830169 Chr.12: 112869816-112869816 on Build GRCh38
rs1003574380 Chr.12: 112869697-112869697 on Build GRCh38
rs924513696 Chr.12: 112869786-112869786 on Build GRCh38
rs771658689 Chr.12: 112869793-112869793 on Build GRCh38
rs1013106673 Chr.12: 112869660-112869660 on Build GRCh38
rs746482059 Chr.12: 112869944-112869944 on Build GRCh38
rs768164688 Chr.12: 112869909-112869909 on Build GRCh38
rs368654367 Chr.12: 112869835-112869835 on Build GRCh38
rs528240437 Chr.12: 112869701-112869701 on Build GRCh38
rs776112889 Chr.12: 112870038-112870038 on Build GRCh38
rs552230104 Chr.12: 112870027-112870027 on Build GRCh38
rs139152687 Chr.12: 112869916-112869916 on Build GRCh38
rs915844793 Chr.12: 112869815-112869815 on Build GRCh38
rs776166351 Chr.12: 112869972-112869972 on Build GRCh38
rs372578786 Chr.12: 112869854-112869854 on Build GRCh38
rs143757610 Chr.12: 112870040-112870040 on Build GRCh38
rs868304671 Chr.12: 112869812-112869812 on Build GRCh38
rs765458966 Chr.12: 112870074-112870074 on Build GRCh38
rs370484039 Chr.12: 112869717-112869717 on Build GRCh38

View Full Product Details