Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112908609-112909123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAS1
Entrez Gene ID
4938
Gene Name
2'-5'-oligoadenylate synthetase 1
Gene Aliases
E18/E16, IFI-4, OIAS, OIASI
UniGene
Hs.524760
Gene Chromosome Location
Chr.12: 112906777-112932190 on Build GRCh38
Transcripts
NM_001032409.2 NM_001320151.1 NM_002534.3 NM_016816.3 XM_006719434.1 XM_011538413.1 XM_017019361.1 XM_017019362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1948841 c.282C>T p.R94R Chr.12: 112908637-112908637 on Build GRCh38
3456309 c.402C>T p.F134F Chr.12: 112908757-112908757 on Build GRCh38
4038893 c.393G>A p.A131A Chr.12: 112908748-112908748 on Build GRCh38
1948849 c.436G>T p.E146* Chr.12: 112908791-112908791 on Build GRCh38
3456303 c.301G>A p.E101K Chr.12: 112908656-112908656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757842108 Chr.12: 112908712-112908712 on Build GRCh38
rs773737618 Chr.12: 112908866-112908866 on Build GRCh38
rs779611860 Chr.12: 112908779-112908779 on Build GRCh38
rs186974774 Chr.12: 112908955-112908955 on Build GRCh38
rs533960394 Chr.12: 112908986-112908986 on Build GRCh38
rs943909898 Chr.12: 112908810-112908810 on Build GRCh38
rs368268014 Chr.12: 112908647-112908647 on Build GRCh38
rs755148052 Chr.12: 112908926-112908926 on Build GRCh38
rs146859513 Chr.12: 112908741-112908741 on Build GRCh38
rs201406745 Chr.12: 112908735-112908735 on Build GRCh38
rs780116900 Chr.12: 112908916-112908916 on Build GRCh38
rs111902215 Chr.12: 112908758-112908758 on Build GRCh38
rs139804868 Chr.12: 112908665-112908665 on Build GRCh38
rs377100213 Chr.12: 112908695-112908695 on Build GRCh38
rs745982177 Chr.12: 112908728-112908728 on Build GRCh38
rs778421230 Chr.12: 112908705-112908705 on Build GRCh38
rs144476783 Chr.12: 112908769-112908769 on Build GRCh38
rs763983553 Chr.12: 112908639-112908639 on Build GRCh38
rs776426424 Chr.12: 112908743-112908743 on Build GRCh38
rs150082040 Chr.12: 112908971-112908971 on Build GRCh38
rs756930576 Chr.12: 112908851-112908851 on Build GRCh38
rs2079223 Chr.12: 112908974-112908974 on Build GRCh38
rs751672413 Chr.12: 112908918-112908918 on Build GRCh38
rs765654789 Chr.12: 112908904-112908904 on Build GRCh38
rs147639145 Chr.12: 112908748-112908748 on Build GRCh38
rs200127558 Chr.12: 112908704-112908704 on Build GRCh38
rs553546694 Chr.12: 112909088-112909088 on Build GRCh38
rs891934983 Chr.12: 112909098-112909098 on Build GRCh38
rs566701570 Chr.12: 112908984-112908984 on Build GRCh38
rs922028878 Chr.12: 112908954-112908954 on Build GRCh38
rs1031664033 Chr.12: 112908736-112908736 on Build GRCh38
rs761673884 Chr.12: 112908747-112908747 on Build GRCh38
rs1044679537 Chr.12: 112909037-112909037 on Build GRCh38
rs374246759 Chr.12: 112908850-112908850 on Build GRCh38
rs201869673 Chr.12: 112908638-112908638 on Build GRCh38
rs764672017 Chr.12: 112908666-112908666 on Build GRCh38
rs996358586 Chr.12: 112908641-112908641 on Build GRCh38
rs767300544 Chr.12: 112908887-112908887 on Build GRCh38
rs765920599 Chr.12: 112909027-112909027 on Build GRCh38
rs754232555 Chr.12: 112908893-112908893 on Build GRCh38
rs757731960 Chr.12: 112908898-112908898 on Build GRCh38
rs759223309 Chr.12: 112908883-112908883 on Build GRCh38
rs758840121 Chr.12: 112908908-112908908 on Build GRCh38
rs751294150 Chr.12: 112908768-112908768 on Build GRCh38
rs200307293 Chr.12: 112908784-112908784 on Build GRCh38
rs749753475 Chr.12: 112908711-112908711 on Build GRCh38
rs756540454 Chr.12: 112908783-112908783 on Build GRCh38
rs1051914858 Chr.12: 112908646-112908646 on Build GRCh38
rs906089477 Chr.12: 112909043-112909043 on Build GRCh38
rs377392276 Chr.12: 112908864-112908864 on Build GRCh38
rs747193487 Chr.12: 112908785-112908785 on Build GRCh38
rs779778631 Chr.12: 112908677-112908677 on Build GRCh38
rs760164326 Chr.12: 112908684-112908687 on Build GRCh38
rs1050995 Chr.12: 112908698-112908698 on Build GRCh38
rs753173709 Chr.12: 112908735-112908736 on Build GRCh38
rs142314177 Chr.12: 112908770-112908770 on Build GRCh38
rs891502627 Chr.12: 112908739-112908739 on Build GRCh38
rs376780624 Chr.12: 112908703-112908703 on Build GRCh38
rs975794978 Chr.12: 112909064-112909064 on Build GRCh38
rs868520100 Chr.12: 112908652-112908652 on Build GRCh38
rs776374570 Chr.12: 112908816-112908816 on Build GRCh38
rs747963981 Chr.12: 112908827-112908827 on Build GRCh38
rs143793109 Chr.12: 112908727-112908727 on Build GRCh38
rs901918534 Chr.12: 112908678-112908678 on Build GRCh38
rs749958330 Chr.12: 112908670-112908670 on Build GRCh38
rs550302030 Chr.12: 112908939-112908939 on Build GRCh38
rs1035851831 Chr.12: 112909084-112909084 on Build GRCh38
rs754283775 Chr.12: 112908772-112908772 on Build GRCh38
rs772158833 Chr.12: 112908729-112908729 on Build GRCh38
rs1035553696 Chr.12: 112908770-112908771 on Build GRCh38
rs1021340095 Chr.12: 112908744-112908744 on Build GRCh38
rs769697834 Chr.12: 112908838-112908838 on Build GRCh38
rs138408097 Chr.12: 112908972-112908972 on Build GRCh38
rs768118701 Chr.12: 112908686-112908687 on Build GRCh38
rs943161449 Chr.12: 112908960-112908960 on Build GRCh38
rs929349171 Chr.12: 112908679-112908679 on Build GRCh38
rs753837415 Chr.12: 112908650-112908650 on Build GRCh38
rs200753414 Chr.12: 112908783-112908783 on Build GRCh38
rs1003081424 Chr.12: 112909068-112909068 on Build GRCh38
rs983425355 Chr.12: 112908798-112908798 on Build GRCh38
rs750994615 Chr.12: 112908906-112908906 on Build GRCh38
rs765917849 Chr.12: 112908765-112908765 on Build GRCh38
rs4767022 Chr.12: 112908734-112908734 on Build GRCh38
rs1052267861 Chr.12: 112908964-112908964 on Build GRCh38
rs768873305 Chr.12: 112908797-112908797 on Build GRCh38
rs200029463 Chr.12: 112908709-112908709 on Build GRCh38
rs775098125 Chr.12: 112908635-112908635 on Build GRCh38
rs757951086 Chr.12: 112908676-112908676 on Build GRCh38
rs757789086 Chr.12: 112908778-112908778 on Build GRCh38
rs746773073 Chr.12: 112908740-112908740 on Build GRCh38

View Full Product Details