Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113427880-113428377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDSL
Entrez Gene ID
113675
Gene Name
serine dehydratase like
Gene Aliases
SDH 2, SDS-RS1, TDH, cSDH
UniGene
Hs.337594
Gene Chromosome Location
Chr.12: 113422265-113438277 on Build GRCh38
Transcripts
NM_001304993.1 NM_138432.3 XM_005253831.4 XM_011537846.2 XM_017018763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1746766 c.165C>G p.F55L Chr.12: 113428147-113428147 on Build GRCh38
3456430 c.81C>T p.S27S Chr.12: 113428063-113428063 on Build GRCh38
467835 c.99T>A p.P33P Chr.12: 113428081-113428081 on Build GRCh38
4457782 c.105C>T p.F35F Chr.12: 113428087-113428087 on Build GRCh38
6000615 c.169C>G p.Q57E Chr.12: 113428151-113428151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763916309 Chr.12: 113427958-113427958 on Build GRCh38
rs1051007000 Chr.12: 113428303-113428303 on Build GRCh38
rs141768354 Chr.12: 113428130-113428130 on Build GRCh38
rs765296857 Chr.12: 113428150-113428150 on Build GRCh38
rs375985606 Chr.12: 113428072-113428072 on Build GRCh38
rs779804620 Chr.12: 113427991-113427991 on Build GRCh38
rs952976897 Chr.12: 113427951-113427951 on Build GRCh38
rs921341266 Chr.12: 113428246-113428246 on Build GRCh38
rs144727433 Chr.12: 113428114-113428114 on Build GRCh38
rs932791710 Chr.12: 113428264-113428264 on Build GRCh38
rs147923357 Chr.12: 113428115-113428115 on Build GRCh38
rs778179308 Chr.12: 113428013-113428013 on Build GRCh38
rs1004059685 Chr.12: 113428321-113428321 on Build GRCh38
rs971597648 Chr.12: 113428126-113428126 on Build GRCh38
rs780114269 Chr.12: 113428189-113428189 on Build GRCh38
rs180761717 Chr.12: 113428239-113428239 on Build GRCh38
rs368001124 Chr.12: 113428171-113428171 on Build GRCh38
rs11542885 Chr.12: 113428084-113428084 on Build GRCh38
rs371591042 Chr.12: 113428179-113428179 on Build GRCh38
rs906707301 Chr.12: 113427922-113427922 on Build GRCh38
rs1029652735 Chr.12: 113427955-113427955 on Build GRCh38
rs994380900 Chr.12: 113427923-113427923 on Build GRCh38
rs367609319 Chr.12: 113428163-113428163 on Build GRCh38
rs148370808 Chr.12: 113428335-113428335 on Build GRCh38
rs747056349 Chr.12: 113428208-113428208 on Build GRCh38
rs771401005 Chr.12: 113428027-113428027 on Build GRCh38
rs957920539 Chr.12: 113428310-113428310 on Build GRCh38
rs764017308 Chr.12: 113428053-113428053 on Build GRCh38
rs751044078 Chr.12: 113428170-113428170 on Build GRCh38
rs149197891 Chr.12: 113428056-113428056 on Build GRCh38
rs747615464 Chr.12: 113428339-113428339 on Build GRCh38
rs765169161 Chr.12: 113428064-113428064 on Build GRCh38
rs746428994 Chr.12: 113428184-113428184 on Build GRCh38
rs142840268 Chr.12: 113428057-113428057 on Build GRCh38
rs977677452 Chr.12: 113428221-113428221 on Build GRCh38
rs772309440 Chr.12: 113428034-113428034 on Build GRCh38
rs947742226 Chr.12: 113428325-113428325 on Build GRCh38
rs1025896940 Chr.12: 113427928-113427928 on Build GRCh38
rs1046519177 Chr.12: 113428327-113428327 on Build GRCh38
rs774427340 Chr.12: 113428028-113428028 on Build GRCh38
rs749608367 Chr.12: 113428111-113428111 on Build GRCh38
rs200331108 Chr.12: 113427988-113427988 on Build GRCh38
rs113073504 Chr.12: 113428234-113428234 on Build GRCh38
rs768859227 Chr.12: 113428140-113428140 on Build GRCh38
rs923063545 Chr.12: 113428058-113428058 on Build GRCh38
rs749619445 Chr.12: 113428230-113428230 on Build GRCh38
rs757208911 Chr.12: 113427964-113427964 on Build GRCh38
rs11066560 Chr.12: 113427961-113427961 on Build GRCh38
rs967470399 Chr.12: 113428247-113428247 on Build GRCh38
rs776307371 Chr.12: 113428141-113428141 on Build GRCh38
rs144641930 Chr.12: 113427989-113427989 on Build GRCh38
rs761623253 Chr.12: 113428148-113428148 on Build GRCh38
rs542356199 Chr.12: 113428308-113428309 on Build GRCh38
rs767148045 Chr.12: 113428178-113428178 on Build GRCh38
rs151061253 Chr.12: 113428062-113428062 on Build GRCh38
rs770098381 Chr.12: 113428007-113428007 on Build GRCh38
rs777599109 Chr.12: 113428257-113428257 on Build GRCh38
rs773322567 Chr.12: 113428162-113428162 on Build GRCh38
rs775725261 Chr.12: 113428042-113428042 on Build GRCh38
rs748555581 Chr.12: 113428004-113428004 on Build GRCh38
rs1015349567 Chr.12: 113428003-113428003 on Build GRCh38
rs751307425 Chr.12: 113428103-113428103 on Build GRCh38
rs573333893 Chr.12: 113428071-113428071 on Build GRCh38
rs759562353 Chr.12: 113428030-113428030 on Build GRCh38
rs778356649 Chr.12: 113427973-113427973 on Build GRCh38
rs142390928 Chr.12: 113428068-113428068 on Build GRCh38
rs997242520 Chr.12: 113427902-113427902 on Build GRCh38
rs879704548 Chr.12: 113428284-113428284 on Build GRCh38
rs995733279 Chr.12: 113428350-113428350 on Build GRCh38
rs143376928 Chr.12: 113428322-113428322 on Build GRCh38
rs758899846 Chr.12: 113428185-113428185 on Build GRCh38
rs747265272 Chr.12: 113428131-113428131 on Build GRCh38
rs929736440 Chr.12: 113428351-113428351 on Build GRCh38
rs754653820 Chr.12: 113428109-113428109 on Build GRCh38
rs376773055 Chr.12: 113427971-113427971 on Build GRCh38
rs190700301 Chr.12: 113427912-113427912 on Build GRCh38
rs778002654 Chr.12: 113428110-113428110 on Build GRCh38
rs562156734 Chr.12: 113428331-113428331 on Build GRCh38
rs749826570 Chr.12: 113428019-113428019 on Build GRCh38
rs112655099 Chr.12: 113428304-113428304 on Build GRCh38
rs556243291 Chr.12: 113428233-113428233 on Build GRCh38
rs376277048 Chr.12: 113428352-113428352 on Build GRCh38
rs760418520 Chr.12: 113427948-113427948 on Build GRCh38
rs754599624 Chr.12: 113428177-113428177 on Build GRCh38
rs771847636 Chr.12: 113428180-113428180 on Build GRCh38

View Full Product Details