Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114677460-114677954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX3
Entrez Gene ID
6926
Gene Name
T-box 3
Gene Aliases
TBX3-ISO, UMS, XHL
UniGene
Hs.744016
Gene Chromosome Location
Chr.12: 114670254-114684164 on Build GRCh38
Transcripts
NM_005996.3 NM_016569.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
111572 c.898delG p.A300fs*23 Chr.12: 114677623-114677623 on Build GRCh38
5639656 c.934G>C p.E312Q Chr.12: 114677587-114677587 on Build GRCh38
6934310 c.904_905ins40 p.G302fs*38 Chr.12: 114677616-114677617 on Build GRCh38
5025042 c.890A>G p.N297S Chr.12: 114677631-114677631 on Build GRCh38
5731086 c.887delA p.N296fs*27 Chr.12: 114677634-114677634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748621209 Chr.12: 114677875-114677875 on Build GRCh38
rs770312024 Chr.12: 114677606-114677606 on Build GRCh38
rs781566311 Chr.12: 114677799-114677799 on Build GRCh38
rs139906364 Chr.12: 114677848-114677848 on Build GRCh38
rs969207709 Chr.12: 114677823-114677823 on Build GRCh38
rs749869983 Chr.12: 114677659-114677659 on Build GRCh38
rs553370331 Chr.12: 114677507-114677507 on Build GRCh38
rs759640660 Chr.12: 114677697-114677697 on Build GRCh38
rs779733514 Chr.12: 114677593-114677593 on Build GRCh38
rs373904412 Chr.12: 114677820-114677820 on Build GRCh38
rs778036694 Chr.12: 114677920-114677920 on Build GRCh38
rs771562847 Chr.12: 114677666-114677666 on Build GRCh38
rs965583891 Chr.12: 114677691-114677691 on Build GRCh38
rs199780926 Chr.12: 114677808-114677808 on Build GRCh38
rs755306919 Chr.12: 114677757-114677757 on Build GRCh38
rs745374468 Chr.12: 114677563-114677563 on Build GRCh38
rs920513254 Chr.12: 114677829-114677829 on Build GRCh38
rs756297400 Chr.12: 114677882-114677882 on Build GRCh38
rs752487479 Chr.12: 114677539-114677539 on Build GRCh38
rs767749049 Chr.12: 114677531-114677531 on Build GRCh38
rs895906689 Chr.12: 114677802-114677802 on Build GRCh38
rs1023894793 Chr.12: 114677810-114677810 on Build GRCh38
rs1020402675 Chr.12: 114677744-114677744 on Build GRCh38
rs995600539 Chr.12: 114677555-114677555 on Build GRCh38
rs910695260 Chr.12: 114677596-114677596 on Build GRCh38
rs780583112 Chr.12: 114677758-114677758 on Build GRCh38
rs778450208 Chr.12: 114677562-114677562 on Build GRCh38
rs1006462379 Chr.12: 114677825-114677825 on Build GRCh38
rs1050288047 Chr.12: 114677856-114677856 on Build GRCh38
rs376967378 Chr.12: 114677717-114677717 on Build GRCh38
rs774334787 Chr.12: 114677672-114677672 on Build GRCh38
rs376913198 Chr.12: 114677552-114677552 on Build GRCh38
rs757126073 Chr.12: 114677559-114677559 on Build GRCh38
rs373797289 Chr.12: 114677603-114677603 on Build GRCh38
rs1057041305 Chr.12: 114677784-114677784 on Build GRCh38
rs1007000811 Chr.12: 114677909-114677909 on Build GRCh38
rs184878027 Chr.12: 114677512-114677512 on Build GRCh38
rs139956231 Chr.12: 114677563-114677566 on Build GRCh38
rs763794799 Chr.12: 114677530-114677530 on Build GRCh38
rs889883077 Chr.12: 114677561-114677561 on Build GRCh38
rs751937475 Chr.12: 114677515-114677515 on Build GRCh38
rs868153604 Chr.12: 114677735-114677735 on Build GRCh38
rs762095058 Chr.12: 114677788-114677788 on Build GRCh38
rs778214854 Chr.12: 114677633-114677633 on Build GRCh38
rs192786215 Chr.12: 114677494-114677494 on Build GRCh38
rs369944278 Chr.12: 114677591-114677591 on Build GRCh38
rs104894376 Chr.12: 114677644-114677644 on Build GRCh38
rs930435973 Chr.12: 114677840-114677840 on Build GRCh38
rs202136964 Chr.12: 114677548-114677548 on Build GRCh38
rs866796237 Chr.12: 114677592-114677592 on Build GRCh38
rs372456075 Chr.12: 114677706-114677706 on Build GRCh38
rs978955872 Chr.12: 114677776-114677776 on Build GRCh38
rs992475782 Chr.12: 114677800-114677800 on Build GRCh38
rs763840075 Chr.12: 114677551-114677551 on Build GRCh38

View Full Product Details