Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19677623-19678165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSPC1
Entrez Gene ID
55269
Gene Name
paraspeckle component 1
Gene Aliases
PSP1
UniGene
Hs.213198
Gene Chromosome Location
Chr.13: 19674752-19783047 on Build GRCh38
Transcripts
NR_003272.2 NR_044998.1 XM_011535137.1 XM_011535139.1 XM_011535140.1 XM_017020650.1

SNP Detail

SNP ID Chromosome Location
rs368279712 Chr.13: 19677935-19677935 on Build GRCh38
rs772106952 Chr.13: 19677804-19677804 on Build GRCh38
rs767940183 Chr.13: 19678106-19678108 on Build GRCh38
rs562133815 Chr.13: 19677733-19677733 on Build GRCh38
rs912842804 Chr.13: 19677700-19677700 on Build GRCh38
rs752086329 Chr.13: 19678073-19678074 on Build GRCh38
rs779808315 Chr.13: 19677799-19677799 on Build GRCh38
rs74038314 Chr.13: 19678123-19678123 on Build GRCh38
rs756306641 Chr.13: 19677792-19677792 on Build GRCh38
rs1010024408 Chr.13: 19677959-19677959 on Build GRCh38
rs537238052 Chr.13: 19677881-19677881 on Build GRCh38
rs905537240 Chr.13: 19677915-19677915 on Build GRCh38
rs150444208 Chr.13: 19678023-19678023 on Build GRCh38
rs529334742 Chr.13: 19677748-19677748 on Build GRCh38
rs771568322 Chr.13: 19677937-19677938 on Build GRCh38
rs542631560 Chr.13: 19677761-19677761 on Build GRCh38
rs546638494 Chr.13: 19677730-19677730 on Build GRCh38
rs569400514 Chr.13: 19677815-19677815 on Build GRCh38
rs778803639 Chr.13: 19677800-19677800 on Build GRCh38
rs753444854 Chr.13: 19677861-19677861 on Build GRCh38
rs966787154 Chr.13: 19678021-19678021 on Build GRCh38
rs535141585 Chr.13: 19678113-19678113 on Build GRCh38
rs746033241 Chr.13: 19677782-19677782 on Build GRCh38
rs985728916 Chr.13: 19677671-19677671 on Build GRCh38
rs570716560 Chr.13: 19678007-19678007 on Build GRCh38
rs985488296 Chr.13: 19678082-19678082 on Build GRCh38
rs931058184 Chr.13: 19677902-19677902 on Build GRCh38
rs1054424927 Chr.13: 19677945-19677945 on Build GRCh38
rs764625503 Chr.13: 19677758-19677758 on Build GRCh38
rs192067516 Chr.13: 19678025-19678025 on Build GRCh38
rs747456100 Chr.13: 19678114-19678114 on Build GRCh38
rs1001382464 Chr.13: 19677931-19677931 on Build GRCh38
rs958585227 Chr.13: 19678027-19678027 on Build GRCh38
rs551038177 Chr.13: 19677777-19677777 on Build GRCh38
rs796225772 Chr.13: 19677891-19677892 on Build GRCh38
rs747991639 Chr.13: 19677803-19677803 on Build GRCh38
rs944174276 Chr.13: 19677774-19677774 on Build GRCh38
rs564437609 Chr.13: 19677680-19677680 on Build GRCh38
rs181803718 Chr.13: 19677715-19677715 on Build GRCh38
rs778488533 Chr.13: 19677969-19677969 on Build GRCh38
rs757411579 Chr.13: 19677765-19677765 on Build GRCh38
rs944118526 Chr.13: 19677795-19677795 on Build GRCh38
rs878856553 Chr.13: 19677676-19677676 on Build GRCh38
rs2290810 Chr.13: 19677651-19677651 on Build GRCh38
rs9508836 Chr.13: 19677953-19677953 on Build GRCh38
rs187274318 Chr.13: 19678024-19678024 on Build GRCh38
rs954250394 Chr.13: 19677667-19677667 on Build GRCh38
rs546016601 Chr.13: 19677652-19677652 on Build GRCh38
rs747335733 Chr.13: 19677998-19677999 on Build GRCh38
rs1041075866 Chr.13: 19677830-19677830 on Build GRCh38
rs533518577 Chr.13: 19678099-19678099 on Build GRCh38
rs1048023849 Chr.13: 19677912-19677912 on Build GRCh38

View Full Product Details