Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21566512-21567043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICU2
Entrez Gene ID
221154
Gene Name
mitochondrial calcium uptake 2
Gene Aliases
1110008L20Rik, EFHA1
UniGene
Hs.412103
Gene Chromosome Location
Chr.13: 21492689-21604216 on Build GRCh38
Transcripts
NM_152726.2 XM_017020433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695803 c.358+1G>C p.? Chr.13: 21566796-21566796 on Build GRCh38
6334067 c.335C>A p.S112* Chr.13: 21566820-21566820 on Build GRCh38
3931794 c.295G>A p.E99K Chr.13: 21566860-21566860 on Build GRCh38
5362247 c.319C>T p.R107* Chr.13: 21566836-21566836 on Build GRCh38
5616615 c.309T>C p.Y103Y Chr.13: 21566846-21566846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370202488 Chr.13: 21566796-21566796 on Build GRCh38
rs547146586 Chr.13: 21566762-21566762 on Build GRCh38
rs956451654 Chr.13: 21566763-21566763 on Build GRCh38
rs139051523 Chr.13: 21566933-21566933 on Build GRCh38
rs138030448 Chr.13: 21566889-21566889 on Build GRCh38
rs564547952 Chr.13: 21566664-21566664 on Build GRCh38
rs66467009 Chr.13: 21566773-21566773 on Build GRCh38
rs780017636 Chr.13: 21566772-21566773 on Build GRCh38
rs1031184655 Chr.13: 21566724-21566724 on Build GRCh38
rs200376390 Chr.13: 21566899-21566899 on Build GRCh38
rs536072102 Chr.13: 21566833-21566833 on Build GRCh38
rs953057717 Chr.13: 21566729-21566729 on Build GRCh38
rs1045961913 Chr.13: 21566689-21566689 on Build GRCh38
rs749678405 Chr.13: 21566775-21566775 on Build GRCh38
rs758822979 Chr.13: 21566912-21566912 on Build GRCh38
rs1015180200 Chr.13: 21566701-21566701 on Build GRCh38
rs764302608 Chr.13: 21566761-21566762 on Build GRCh38
rs555042099 Chr.13: 21566898-21566898 on Build GRCh38
rs144126023 Chr.13: 21566917-21566917 on Build GRCh38
rs765326422 Chr.13: 21566875-21566875 on Build GRCh38
rs766774688 Chr.13: 21566825-21566825 on Build GRCh38
rs745370212 Chr.13: 21566590-21566590 on Build GRCh38
rs760896116 Chr.13: 21566866-21566866 on Build GRCh38
rs568539904 Chr.13: 21566769-21566769 on Build GRCh38
rs757915263 Chr.13: 21566608-21566608 on Build GRCh38
rs894728721 Chr.13: 21566661-21566661 on Build GRCh38
rs1056364530 Chr.13: 21566565-21566571 on Build GRCh38
rs140343130 Chr.13: 21566819-21566819 on Build GRCh38
rs778474100 Chr.13: 21566916-21566916 on Build GRCh38
rs776458287 Chr.13: 21566836-21566836 on Build GRCh38
rs1021720104 Chr.13: 21566983-21566983 on Build GRCh38
rs556957719 Chr.13: 21566906-21566906 on Build GRCh38
rs907417223 Chr.13: 21566708-21566708 on Build GRCh38
rs982589643 Chr.13: 21566839-21566839 on Build GRCh38
rs985874610 Chr.13: 21566733-21566733 on Build GRCh38
rs1029984377 Chr.13: 21566725-21566725 on Build GRCh38
rs768629884 Chr.13: 21566799-21566799 on Build GRCh38
rs760631435 Chr.13: 21566990-21566990 on Build GRCh38
rs763064465 Chr.13: 21566982-21566982 on Build GRCh38
rs749936687 Chr.13: 21566835-21566835 on Build GRCh38
rs199551748 Chr.13: 21566967-21566967 on Build GRCh38
rs780792134 Chr.13: 21566958-21566958 on Build GRCh38
rs763918348 Chr.13: 21566831-21566831 on Build GRCh38
rs775566916 Chr.13: 21566961-21566961 on Build GRCh38
rs750226199 Chr.13: 21566773-21566774 on Build GRCh38
rs975244764 Chr.13: 21566541-21566541 on Build GRCh38
rs138492568 Chr.13: 21566834-21566834 on Build GRCh38
rs191525541 Chr.13: 21566941-21566941 on Build GRCh38
rs773441023 Chr.13: 21566797-21566797 on Build GRCh38
rs750343727 Chr.13: 21566892-21566892 on Build GRCh38
rs75781856 Chr.13: 21566773-21566773 on Build GRCh38
rs751381933 Chr.13: 21566880-21566880 on Build GRCh38
rs896512571 Chr.13: 21566577-21566577 on Build GRCh38
rs768156925 Chr.13: 21566855-21566855 on Build GRCh38
rs377461488 Chr.13: 21566991-21566991 on Build GRCh38
rs551386130 Chr.13: 21566547-21566547 on Build GRCh38
rs1056668496 Chr.13: 21566816-21566816 on Build GRCh38
rs1021920974 Chr.13: 21566792-21566792 on Build GRCh38
rs113126801 Chr.13: 21566923-21566923 on Build GRCh38
rs761526718 Chr.13: 21566882-21566882 on Build GRCh38
rs1038344843 Chr.13: 21566626-21566626 on Build GRCh38
rs769047337 Chr.13: 21566786-21566786 on Build GRCh38
rs150339447 Chr.13: 21566873-21566873 on Build GRCh38
rs887606190 Chr.13: 21566683-21566683 on Build GRCh38
rs376441377 Chr.13: 21566850-21566850 on Build GRCh38
rs778495852 Chr.13: 21566565-21566565 on Build GRCh38
rs753241593 Chr.13: 21566903-21566903 on Build GRCh38
rs771524116 Chr.13: 21566919-21566919 on Build GRCh38
rs373237105 Chr.13: 21566858-21566858 on Build GRCh38
rs754987931 Chr.13: 21566844-21566845 on Build GRCh38
rs754078154 Chr.13: 21566828-21566828 on Build GRCh38
rs1045993590 Chr.13: 21566931-21566931 on Build GRCh38
rs748406109 Chr.13: 21566862-21566862 on Build GRCh38
rs968879948 Chr.13: 21566762-21566765 on Build GRCh38
rs149549167 Chr.13: 21566907-21566907 on Build GRCh38
rs773853625 Chr.13: 21566864-21566864 on Build GRCh38
rs569454339 Chr.13: 21566851-21566851 on Build GRCh38
rs754674711 Chr.13: 21566905-21566905 on Build GRCh38
rs762706133 Chr.13: 21566829-21566829 on Build GRCh38
rs746972133 Chr.13: 21566715-21566715 on Build GRCh38
rs75238402 Chr.13: 21566684-21566684 on Build GRCh38
rs550404982 Chr.13: 21566582-21566582 on Build GRCh38
rs751491033 Chr.13: 21566820-21566822 on Build GRCh38
rs573366291 Chr.13: 21566971-21566971 on Build GRCh38
rs770046745 Chr.13: 21566938-21566938 on Build GRCh38
rs1047207227 Chr.13: 21566566-21566566 on Build GRCh38
rs771113308 Chr.13: 21566867-21566867 on Build GRCh38
rs760771373 Chr.13: 21566824-21566824 on Build GRCh38
rs779898162 Chr.13: 21566668-21566668 on Build GRCh38

View Full Product Details