Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12115485-12115980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF8
Entrez Gene ID
943
Gene Name
TNF receptor superfamily member 8
Gene Aliases
CD30, D1S166E, Ki-1
UniGene
Hs.1314
Gene Chromosome Location
Chr.1: 12063318-12144213 on Build GRCh38
Transcripts
NM_001243.4 NM_001281430.2 XM_011542441.2 XM_011542443.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328170 c.842G>A p.C281Y Chr.1: 12115625-12115625 on Build GRCh38
6805088 c.820G>T p.A274S Chr.1: 12115603-12115603 on Build GRCh38
5992170 c.939G>A p.K313K Chr.1: 12115722-12115722 on Build GRCh38
3472368 c.943C>T p.Q315* Chr.1: 12115726-12115726 on Build GRCh38
5993102 c.850C>T p.R284* Chr.1: 12115633-12115633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201412076 Chr.1: 12115637-12115637 on Build GRCh38
rs763878547 Chr.1: 12115561-12115561 on Build GRCh38
rs1023273581 Chr.1: 12115842-12115842 on Build GRCh38
rs530146043 Chr.1: 12115552-12115552 on Build GRCh38
rs777062822 Chr.1: 12115541-12115541 on Build GRCh38
rs1021691056 Chr.1: 12115543-12115543 on Build GRCh38
rs765891942 Chr.1: 12115665-12115665 on Build GRCh38
rs11569894 Chr.1: 12115830-12115831 on Build GRCh38
rs541107601 Chr.1: 12115512-12115512 on Build GRCh38
rs201661836 Chr.1: 12115678-12115678 on Build GRCh38
rs969554166 Chr.1: 12115868-12115868 on Build GRCh38
rs889880271 Chr.1: 12115689-12115689 on Build GRCh38
rs778203361 Chr.1: 12115595-12115595 on Build GRCh38
rs757850568 Chr.1: 12115771-12115771 on Build GRCh38
rs766952305 Chr.1: 12115761-12115761 on Build GRCh38
rs753518229 Chr.1: 12115670-12115670 on Build GRCh38
rs753474537 Chr.1: 12115756-12115756 on Build GRCh38
rs141890526 Chr.1: 12115594-12115594 on Build GRCh38
rs927693680 Chr.1: 12115946-12115946 on Build GRCh38
rs762712685 Chr.1: 12115619-12115619 on Build GRCh38
rs1043391417 Chr.1: 12115778-12115778 on Build GRCh38
rs1043014905 Chr.1: 12115680-12115680 on Build GRCh38
rs764390146 Chr.1: 12115763-12115763 on Build GRCh38
rs970003693 Chr.1: 12115947-12115948 on Build GRCh38
rs200643752 Chr.1: 12115610-12115610 on Build GRCh38
rs780155636 Chr.1: 12115777-12115777 on Build GRCh38
rs1763642 Chr.1: 12115672-12115672 on Build GRCh38
rs750121000 Chr.1: 12115574-12115574 on Build GRCh38
rs760164725 Chr.1: 12115744-12115744 on Build GRCh38
rs928787885 Chr.1: 12115629-12115629 on Build GRCh38
rs772723761 Chr.1: 12115741-12115741 on Build GRCh38
rs769902715 Chr.1: 12115700-12115700 on Build GRCh38
rs943278699 Chr.1: 12115790-12115790 on Build GRCh38
rs778171119 Chr.1: 12115676-12115676 on Build GRCh38
rs181861016 Chr.1: 12115773-12115773 on Build GRCh38
rs1006171705 Chr.1: 12115704-12115704 on Build GRCh38
rs200165570 Chr.1: 12115546-12115546 on Build GRCh38
rs763942820 Chr.1: 12115622-12115622 on Build GRCh38
rs1040269590 Chr.1: 12115837-12115837 on Build GRCh38
rs146527504 Chr.1: 12115712-12115712 on Build GRCh38
rs867554047 Chr.1: 12115764-12115764 on Build GRCh38
rs147298360 Chr.1: 12115643-12115643 on Build GRCh38
rs1048314385 Chr.1: 12115874-12115874 on Build GRCh38
rs751904523 Chr.1: 12115770-12115770 on Build GRCh38
rs371950593 Chr.1: 12115776-12115776 on Build GRCh38
rs141089207 Chr.1: 12115724-12115724 on Build GRCh38
rs140175552 Chr.1: 12115618-12115618 on Build GRCh38
rs533782066 Chr.1: 12115647-12115647 on Build GRCh38
rs373120260 Chr.1: 12115564-12115564 on Build GRCh38
rs983808591 Chr.1: 12115812-12115812 on Build GRCh38
rs762951877 Chr.1: 12115557-12115557 on Build GRCh38
rs761712429 Chr.1: 12115633-12115633 on Build GRCh38
rs768879885 Chr.1: 12115780-12115780 on Build GRCh38
rs768381369 Chr.1: 12115713-12115713 on Build GRCh38
rs371441635 Chr.1: 12115745-12115745 on Build GRCh38
rs752393553 Chr.1: 12115553-12115553 on Build GRCh38
rs749492353 Chr.1: 12115779-12115779 on Build GRCh38
rs544837580 Chr.1: 12115906-12115906 on Build GRCh38
rs561387407 Chr.1: 12115526-12115526 on Build GRCh38
rs148303398 Chr.1: 12115828-12115829 on Build GRCh38
rs759867786 Chr.1: 12115605-12115605 on Build GRCh38
rs769346386 Chr.1: 12115898-12115898 on Build GRCh38
rs2275170 Chr.1: 12115723-12115723 on Build GRCh38
rs901677640 Chr.1: 12115859-12115859 on Build GRCh38
rs114975232 Chr.1: 12115596-12115596 on Build GRCh38
rs1003538499 Chr.1: 12115948-12115948 on Build GRCh38
rs767032588 Chr.1: 12115634-12115634 on Build GRCh38
rs1019868198 Chr.1: 12115746-12115746 on Build GRCh38
rs781425743 Chr.1: 12115686-12115686 on Build GRCh38
rs1022909116 Chr.1: 12115921-12115921 on Build GRCh38
rs755711631 Chr.1: 12115578-12115578 on Build GRCh38
rs779370092 Chr.1: 12115579-12115579 on Build GRCh38
rs368632391 Chr.1: 12115733-12115733 on Build GRCh38
rs598661 Chr.1: 12115761-12115761 on Build GRCh38
rs550362745 Chr.1: 12115565-12115565 on Build GRCh38
rs751500558 Chr.1: 12115563-12115563 on Build GRCh38
rs928813047 Chr.1: 12115754-12115754 on Build GRCh38
rs745885297 Chr.1: 12115696-12115696 on Build GRCh38
rs901045810 Chr.1: 12115760-12115760 on Build GRCh38
rs200005472 Chr.1: 12115710-12115710 on Build GRCh38
rs771123475 Chr.1: 12115532-12115532 on Build GRCh38
rs748399721 Chr.1: 12115583-12115583 on Build GRCh38
rs199906292 Chr.1: 12115627-12115627 on Build GRCh38
rs765909853 Chr.1: 12115755-12115755 on Build GRCh38
rs771174816 Chr.1: 12115600-12115600 on Build GRCh38

View Full Product Details