Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 13749248-13749784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM2
Entrez Gene ID
7799
Gene Name
PR domain 2
Gene Aliases
HUMHOXY1, KMT8, MTB-ZF, RIZ, RIZ1, RIZ2
UniGene
Hs.371823
Gene Chromosome Location
Chr.1: 13698875-13825079 on Build GRCh38
Transcripts
NM_001007257.2 NM_001135610.1 NM_012231.4 NM_015866.4 XM_017002253.1 XM_017002254.1 XM_017002255.1 XM_017002256.1 XM_017002257.1 XM_017002258.1 XM_017002259.1 XM_017002262.1 XM_017002264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4431289 c.494C>A p.S165Y Chr.1: 13749470-13749470 on Build GRCh38
4399807 c.459G>A p.E153E Chr.1: 13749435-13749435 on Build GRCh38
7385453 c.475G>C p.A159P Chr.1: 13749451-13749451 on Build GRCh38
4399808 c.459G>A p.E153E Chr.1: 13749435-13749435 on Build GRCh38
5843371 c.479G>A p.R160Q Chr.1: 13749455-13749455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892494299 Chr.1: 13749503-13749503 on Build GRCh38
rs1022412889 Chr.1: 13749524-13749524 on Build GRCh38
rs868166917 Chr.1: 13749306-13749306 on Build GRCh38
rs760560043 Chr.1: 13749384-13749384 on Build GRCh38
rs934472723 Chr.1: 13749663-13749663 on Build GRCh38
rs748308675 Chr.1: 13749448-13749448 on Build GRCh38
rs1043749686 Chr.1: 13749681-13749681 on Build GRCh38
rs893035536 Chr.1: 13749675-13749675 on Build GRCh38
rs762917415 Chr.1: 13749351-13749351 on Build GRCh38
rs979449929 Chr.1: 13749286-13749287 on Build GRCh38
rs530360396 Chr.1: 13749285-13749285 on Build GRCh38
rs144029481 Chr.1: 13749452-13749452 on Build GRCh38
rs776077953 Chr.1: 13749454-13749454 on Build GRCh38
rs779322475 Chr.1: 13749499-13749499 on Build GRCh38
rs753256126 Chr.1: 13749320-13749320 on Build GRCh38
rs919498556 Chr.1: 13749599-13749599 on Build GRCh38
rs551419635 Chr.1: 13749406-13749406 on Build GRCh38
rs757747800 Chr.1: 13749419-13749419 on Build GRCh38
rs948437680 Chr.1: 13749470-13749470 on Build GRCh38
rs746196158 Chr.1: 13749431-13749431 on Build GRCh38
rs781549621 Chr.1: 13749336-13749336 on Build GRCh38
rs376400657 Chr.1: 13749718-13749718 on Build GRCh38
rs956895065 Chr.1: 13749287-13749287 on Build GRCh38
rs928502156 Chr.1: 13749652-13749652 on Build GRCh38
rs774367029 Chr.1: 13749357-13749357 on Build GRCh38
rs75417771 Chr.1: 13749385-13749385 on Build GRCh38
rs753552653 Chr.1: 13749390-13749390 on Build GRCh38
rs989233079 Chr.1: 13749302-13749302 on Build GRCh38
rs951777654 Chr.1: 13749600-13749600 on Build GRCh38
rs769899423 Chr.1: 13749342-13749342 on Build GRCh38
rs778095036 Chr.1: 13749324-13749324 on Build GRCh38
rs201537864 Chr.1: 13749446-13749446 on Build GRCh38
rs889090573 Chr.1: 13749686-13749686 on Build GRCh38
rs755227424 Chr.1: 13749496-13749496 on Build GRCh38
rs749676309 Chr.1: 13749491-13749491 on Build GRCh38
rs966186455 Chr.1: 13749571-13749571 on Build GRCh38
rs568964063 Chr.1: 13749353-13749353 on Build GRCh38
rs986175979 Chr.1: 13749540-13749540 on Build GRCh38
rs985035207 Chr.1: 13749752-13749752 on Build GRCh38
rs750242269 Chr.1: 13749379-13749379 on Build GRCh38
rs1045429464 Chr.1: 13749651-13749651 on Build GRCh38
rs1022783875 Chr.1: 13749319-13749319 on Build GRCh38
rs948492468 Chr.1: 13749646-13749646 on Build GRCh38
rs1022599824 Chr.1: 13749747-13749747 on Build GRCh38
rs775610644 Chr.1: 13749502-13749509 on Build GRCh38
rs373014373 Chr.1: 13749346-13749346 on Build GRCh38
rs1010591869 Chr.1: 13749734-13749734 on Build GRCh38
rs756390641 Chr.1: 13749475-13749475 on Build GRCh38
rs200292612 Chr.1: 13749473-13749473 on Build GRCh38
rs767625644 Chr.1: 13749375-13749375 on Build GRCh38
rs781710949 Chr.1: 13749429-13749429 on Build GRCh38
rs1011459466 Chr.1: 13749678-13749678 on Build GRCh38
rs139226961 Chr.1: 13749451-13749451 on Build GRCh38
rs866976875 Chr.1: 13749369-13749369 on Build GRCh38
rs907134399 Chr.1: 13749272-13749272 on Build GRCh38
rs576153398 Chr.1: 13749668-13749668 on Build GRCh38
rs754403875 Chr.1: 13749392-13749392 on Build GRCh38
rs956409674 Chr.1: 13749297-13749297 on Build GRCh38
rs556070285 Chr.1: 13749477-13749477 on Build GRCh38
rs756547427 Chr.1: 13749435-13749435 on Build GRCh38
rs201907593 Chr.1: 13749321-13749321 on Build GRCh38
rs972367140 Chr.1: 13749578-13749578 on Build GRCh38
rs994062380 Chr.1: 13749743-13749743 on Build GRCh38
rs772301096 Chr.1: 13749509-13749509 on Build GRCh38
rs915536399 Chr.1: 13749632-13749632 on Build GRCh38
rs1019135791 Chr.1: 13749551-13749551 on Build GRCh38
rs1007675802 Chr.1: 13749704-13749704 on Build GRCh38
rs750928702 Chr.1: 13749618-13749618 on Build GRCh38
rs748216562 Chr.1: 13749505-13749505 on Build GRCh38
rs553659427 Chr.1: 13749461-13749461 on Build GRCh38
rs749532757 Chr.1: 13749440-13749440 on Build GRCh38
rs989611351 Chr.1: 13749294-13749294 on Build GRCh38
rs761858927 Chr.1: 13749368-13749368 on Build GRCh38
rs918958663 Chr.1: 13749425-13749425 on Build GRCh38
rs774563180 Chr.1: 13749447-13749447 on Build GRCh38
rs771216227 Chr.1: 13749334-13749334 on Build GRCh38
rs1026446765 Chr.1: 13749746-13749746 on Build GRCh38
rs746288161 Chr.1: 13749337-13749337 on Build GRCh38
rs752005423 Chr.1: 13749414-13749414 on Build GRCh38
rs1011322828 Chr.1: 13749522-13749522 on Build GRCh38
rs984869917 Chr.1: 13749607-13749607 on Build GRCh38
rs998743602 Chr.1: 13749502-13749505 on Build GRCh38
rs747544897 Chr.1: 13749330-13749330 on Build GRCh38
rs536087579 Chr.1: 13749474-13749474 on Build GRCh38
rs765342855 Chr.1: 13749495-13749495 on Build GRCh38
rs571174750 Chr.1: 13749436-13749436 on Build GRCh38
rs183206714 Chr.1: 13749586-13749586 on Build GRCh38
rs953451168 Chr.1: 13749525-13749525 on Build GRCh38

View Full Product Details