Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26667410-26667955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WASF3
Entrez Gene ID
10810
Gene Name
WAS protein family member 3
Gene Aliases
Brush-1, SCAR3, WAVE3
UniGene
Hs.618732
Gene Chromosome Location
Chr.13: 26557703-26688945 on Build GRCh38
Transcripts
NM_001291965.1 NM_006646.5 XM_011534889.1 XM_011534890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2069830 c.304A>G p.K102E Chr.13: 26667552-26667552 on Build GRCh38
6821213 c.278A>G p.Q93R Chr.13: 26667526-26667526 on Build GRCh38
3936282 c.313A>G p.T105A Chr.13: 26667561-26667561 on Build GRCh38
2069836 c.397C>A p.P133T Chr.13: 26667645-26667645 on Build GRCh38
6821212 c.278A>G p.Q93R Chr.13: 26667526-26667526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765322091 Chr.13: 26667697-26667697 on Build GRCh38
rs553950827 Chr.13: 26667746-26667746 on Build GRCh38
rs640409 Chr.13: 26667867-26667867 on Build GRCh38
rs909331326 Chr.13: 26667765-26667765 on Build GRCh38
rs201409351 Chr.13: 26667645-26667645 on Build GRCh38
rs749123186 Chr.13: 26667638-26667638 on Build GRCh38
rs1020736816 Chr.13: 26667567-26667567 on Build GRCh38
rs185123572 Chr.13: 26667822-26667822 on Build GRCh38
rs918909960 Chr.13: 26667741-26667741 on Build GRCh38
rs758520406 Chr.13: 26667546-26667546 on Build GRCh38
rs755108011 Chr.13: 26667517-26667517 on Build GRCh38
rs768040180 Chr.13: 26667494-26667495 on Build GRCh38
rs779228823 Chr.13: 26667537-26667537 on Build GRCh38
rs142839529 Chr.13: 26667654-26667654 on Build GRCh38
rs748113527 Chr.13: 26667541-26667541 on Build GRCh38
rs903200299 Chr.13: 26667926-26667926 on Build GRCh38
rs767852120 Chr.13: 26667482-26667482 on Build GRCh38
rs777059387 Chr.13: 26667745-26667745 on Build GRCh38
rs373736425 Chr.13: 26667593-26667593 on Build GRCh38
rs899912290 Chr.13: 26667448-26667448 on Build GRCh38
rs572719077 Chr.13: 26667908-26667908 on Build GRCh38
rs148479716 Chr.13: 26667843-26667843 on Build GRCh38
rs987798173 Chr.13: 26667819-26667819 on Build GRCh38
rs766895693 Chr.13: 26667674-26667674 on Build GRCh38
rs200246981 Chr.13: 26667597-26667597 on Build GRCh38
rs369038974 Chr.13: 26667483-26667483 on Build GRCh38
rs775863891 Chr.13: 26667890-26667890 on Build GRCh38
rs868446113 Chr.13: 26667914-26667914 on Build GRCh38
rs768739566 Chr.13: 26667450-26667450 on Build GRCh38
rs774692814 Chr.13: 26667721-26667721 on Build GRCh38
rs796898567 Chr.13: 26667440-26667440 on Build GRCh38
rs768125939 Chr.13: 26667643-26667643 on Build GRCh38
rs758407418 Chr.13: 26667716-26667716 on Build GRCh38
rs763826664 Chr.13: 26667466-26667466 on Build GRCh38
rs370615100 Chr.13: 26667681-26667681 on Build GRCh38
rs569715756 Chr.13: 26667493-26667493 on Build GRCh38
rs942112749 Chr.13: 26667821-26667821 on Build GRCh38
rs992827950 Chr.13: 26667665-26667665 on Build GRCh38
rs181363569 Chr.13: 26667905-26667905 on Build GRCh38
rs753025530 Chr.13: 26667710-26667710 on Build GRCh38
rs754037003 Chr.13: 26667494-26667494 on Build GRCh38
rs367882739 Chr.13: 26667601-26667601 on Build GRCh38
rs761219941 Chr.13: 26667669-26667669 on Build GRCh38
rs767121352 Chr.13: 26667647-26667647 on Build GRCh38
rs537742982 Chr.13: 26667644-26667644 on Build GRCh38
rs545701506 Chr.13: 26667889-26667889 on Build GRCh38
rs189591935 Chr.13: 26667881-26667881 on Build GRCh38
rs942058729 Chr.13: 26667462-26667462 on Build GRCh38
rs890964343 Chr.13: 26667498-26667498 on Build GRCh38
rs7329295 Chr.13: 26667438-26667438 on Build GRCh38
rs778045071 Chr.13: 26667718-26667718 on Build GRCh38
rs903642532 Chr.13: 26667920-26667920 on Build GRCh38
rs1051468663 Chr.13: 26667860-26667860 on Build GRCh38
rs371716239 Chr.13: 26667485-26667485 on Build GRCh38
rs781266699 Chr.13: 26667551-26667551 on Build GRCh38
rs749391813 Chr.13: 26667563-26667563 on Build GRCh38
rs891023694 Chr.13: 26667861-26667861 on Build GRCh38
rs533722401 Chr.13: 26667739-26667739 on Build GRCh38
rs1022701303 Chr.13: 26667666-26667666 on Build GRCh38
rs549866682 Chr.13: 26667452-26667452 on Build GRCh38
rs745870998 Chr.13: 26667555-26667555 on Build GRCh38
rs761389915 Chr.13: 26667481-26667481 on Build GRCh38
rs773690969 Chr.13: 26667467-26667467 on Build GRCh38
rs776269464 Chr.13: 26667720-26667720 on Build GRCh38

View Full Product Details