Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27074904-27075444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP12
Entrez Gene ID
219333
Gene Name
ubiquitin specific peptidase 12
Gene Aliases
UBH1, USP12L1
UniGene
Hs.42400
Gene Chromosome Location
Chr.13: 27066150-27171896 on Build GRCh38
Transcripts
NM_182488.3 XM_005266282.2 XM_011534956.2 XM_017020416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6190842 c.922C>T p.H308Y Chr.13: 27075201-27075201 on Build GRCh38
6230734 c.829C>T p.R277W Chr.13: 27075294-27075294 on Build GRCh38
4839761 c.800A>T p.Q267L Chr.13: 27075323-27075323 on Build GRCh38
4973634 c.855T>C p.R285R Chr.13: 27075268-27075268 on Build GRCh38
6374373 c.830G>T p.R277L Chr.13: 27075293-27075293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781633488 Chr.13: 27075388-27075388 on Build GRCh38
rs910393267 Chr.13: 27075378-27075378 on Build GRCh38
rs532685727 Chr.13: 27075096-27075096 on Build GRCh38
rs368434115 Chr.13: 27075318-27075318 on Build GRCh38
rs776417947 Chr.13: 27075269-27075269 on Build GRCh38
rs781550624 Chr.13: 27075143-27075143 on Build GRCh38
rs962030861 Chr.13: 27075119-27075119 on Build GRCh38
rs936462508 Chr.13: 27075382-27075382 on Build GRCh38
rs531487421 Chr.13: 27074941-27074941 on Build GRCh38
rs1006484392 Chr.13: 27075107-27075107 on Build GRCh38
rs954813789 Chr.13: 27075260-27075260 on Build GRCh38
rs769554767 Chr.13: 27075222-27075222 on Build GRCh38
rs1003477885 Chr.13: 27074981-27074981 on Build GRCh38
rs751416921 Chr.13: 27075411-27075411 on Build GRCh38
rs772185400 Chr.13: 27075414-27075414 on Build GRCh38
rs771701264 Chr.13: 27075013-27075013 on Build GRCh38
rs770058125 Chr.13: 27075170-27075170 on Build GRCh38
rs374759487 Chr.13: 27074943-27074943 on Build GRCh38
rs911236493 Chr.13: 27074953-27074953 on Build GRCh38
rs1023693649 Chr.13: 27075174-27075174 on Build GRCh38
rs756269699 Chr.13: 27075416-27075416 on Build GRCh38
rs11839835 Chr.13: 27075092-27075092 on Build GRCh38
rs569734106 Chr.13: 27075004-27075004 on Build GRCh38
rs375248356 Chr.13: 27075252-27075252 on Build GRCh38
rs906217757 Chr.13: 27075176-27075176 on Build GRCh38
rs746258512 Chr.13: 27075407-27075407 on Build GRCh38
rs542999112 Chr.13: 27074961-27074961 on Build GRCh38
rs370714845 Chr.13: 27075177-27075177 on Build GRCh38
rs777450034 Chr.13: 27075274-27075274 on Build GRCh38
rs892139754 Chr.13: 27074935-27074935 on Build GRCh38
rs764962303 Chr.13: 27075286-27075286 on Build GRCh38
rs1014785113 Chr.13: 27075206-27075206 on Build GRCh38
rs779742764 Chr.13: 27075270-27075270 on Build GRCh38
rs34937145 Chr.13: 27074939-27074940 on Build GRCh38
rs757961837 Chr.13: 27075301-27075301 on Build GRCh38
rs766109391 Chr.13: 27075256-27075256 on Build GRCh38
rs746061873 Chr.13: 27075292-27075292 on Build GRCh38
rs113419409 Chr.13: 27074970-27074970 on Build GRCh38
rs559705244 Chr.13: 27075173-27075173 on Build GRCh38
rs757762594 Chr.13: 27075141-27075141 on Build GRCh38
rs893544097 Chr.13: 27075128-27075128 on Build GRCh38
rs766243884 Chr.13: 27075409-27075411 on Build GRCh38
rs764504408 Chr.13: 27075310-27075310 on Build GRCh38
rs1016444721 Chr.13: 27075113-27075113 on Build GRCh38
rs747341850 Chr.13: 27075140-27075140 on Build GRCh38
rs371701380 Chr.13: 27075124-27075124 on Build GRCh38
rs906270393 Chr.13: 27074973-27074973 on Build GRCh38
rs752169828 Chr.13: 27075313-27075313 on Build GRCh38
rs757850604 Chr.13: 27075348-27075348 on Build GRCh38
rs771874624 Chr.13: 27075183-27075183 on Build GRCh38
rs1034523021 Chr.13: 27075002-27075002 on Build GRCh38
rs746028379 Chr.13: 27075167-27075167 on Build GRCh38
rs868488155 Chr.13: 27075090-27075090 on Build GRCh38

View Full Product Details