Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30751974-30752477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5AP
Entrez Gene ID
241
Gene Name
arachidonate 5-lipoxygenase activating protein
Gene Aliases
FLAP
UniGene
Hs.507658
Gene Chromosome Location
Chr.13: 30713478-30764428 on Build GRCh38
Transcripts
NM_001204406.1 NM_001629.3 XM_017020522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
946638 c.222G>A p.A74A Chr.13: 30752103-30752103 on Build GRCh38
2070852 c.192C>A p.Y64* Chr.13: 30752073-30752073 on Build GRCh38
5554310 c.201C>A p.F67L Chr.13: 30752082-30752082 on Build GRCh38
4046972 c.221C>T p.A74V Chr.13: 30752102-30752102 on Build GRCh38
3726630 c.241+2T>G p.? Chr.13: 30752124-30752124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530927922 Chr.13: 30752184-30752184 on Build GRCh38
rs975579977 Chr.13: 30752158-30752158 on Build GRCh38
rs561279124 Chr.13: 30752189-30752189 on Build GRCh38
rs144243124 Chr.13: 30752070-30752070 on Build GRCh38
rs750930358 Chr.13: 30752003-30752003 on Build GRCh38
rs546802161 Chr.13: 30752224-30752224 on Build GRCh38
rs948767572 Chr.13: 30752218-30752218 on Build GRCh38
rs12721459 Chr.13: 30752186-30752186 on Build GRCh38
rs1018580549 Chr.13: 30752293-30752293 on Build GRCh38
rs199505040 Chr.13: 30752159-30752159 on Build GRCh38
rs376956587 Chr.13: 30752104-30752104 on Build GRCh38
rs373870868 Chr.13: 30752301-30752301 on Build GRCh38
rs1007070969 Chr.13: 30752213-30752213 on Build GRCh38
rs897270030 Chr.13: 30752185-30752185 on Build GRCh38
rs1041280254 Chr.13: 30752441-30752441 on Build GRCh38
rs758448218 Chr.13: 30752046-30752046 on Build GRCh38
rs186646149 Chr.13: 30752382-30752382 on Build GRCh38
rs990701324 Chr.13: 30752292-30752292 on Build GRCh38
rs201824468 Chr.13: 30752086-30752086 on Build GRCh38
rs908806572 Chr.13: 30752440-30752440 on Build GRCh38
rs757321893 Chr.13: 30752212-30752212 on Build GRCh38
rs571778895 Chr.13: 30752250-30752250 on Build GRCh38
rs964431072 Chr.13: 30752127-30752128 on Build GRCh38
rs779898044 Chr.13: 30752069-30752069 on Build GRCh38
rs766941046 Chr.13: 30752012-30752012 on Build GRCh38
rs1020686750 Chr.13: 30752265-30752265 on Build GRCh38
rs530940896 Chr.13: 30752042-30752042 on Build GRCh38
rs921480610 Chr.13: 30752177-30752177 on Build GRCh38
rs567170819 Chr.13: 30752376-30752376 on Build GRCh38
rs563599872 Chr.13: 30752060-30752060 on Build GRCh38
rs761292818 Chr.13: 30752004-30752004 on Build GRCh38
rs764123878 Chr.13: 30752018-30752021 on Build GRCh38
rs17239172 Chr.13: 30752197-30752197 on Build GRCh38
rs201237838 Chr.13: 30752157-30752157 on Build GRCh38
rs908768512 Chr.13: 30752187-30752187 on Build GRCh38
rs570802966 Chr.13: 30752227-30752227 on Build GRCh38
rs577742408 Chr.13: 30752453-30752453 on Build GRCh38
rs17239186 Chr.13: 30752408-30752408 on Build GRCh38
rs918665034 Chr.13: 30752314-30752314 on Build GRCh38
rs758670042 Chr.13: 30752164-30752164 on Build GRCh38
rs925211658 Chr.13: 30752010-30752010 on Build GRCh38
rs756061691 Chr.13: 30752068-30752068 on Build GRCh38
rs532631164 Chr.13: 30752260-30752260 on Build GRCh38
rs768483394 Chr.13: 30752075-30752075 on Build GRCh38
rs200393677 Chr.13: 30752103-30752103 on Build GRCh38
rs952931349 Chr.13: 30752401-30752401 on Build GRCh38
rs569250026 Chr.13: 30752269-30752269 on Build GRCh38
rs771922376 Chr.13: 30752315-30752315 on Build GRCh38
rs1044873386 Chr.13: 30752239-30752239 on Build GRCh38
rs545270644 Chr.13: 30752035-30752035 on Build GRCh38
rs777940375 Chr.13: 30752058-30752058 on Build GRCh38
rs201136470 Chr.13: 30752062-30752062 on Build GRCh38
rs940241738 Chr.13: 30752442-30752442 on Build GRCh38
rs1032153779 Chr.13: 30752280-30752280 on Build GRCh38
rs142118435 Chr.13: 30752277-30752277 on Build GRCh38
rs777267497 Chr.13: 30752132-30752132 on Build GRCh38
rs753221284 Chr.13: 30752033-30752036 on Build GRCh38
rs771580492 Chr.13: 30752090-30752090 on Build GRCh38
rs973167307 Chr.13: 30752308-30752308 on Build GRCh38
rs779674447 Chr.13: 30752409-30752409 on Build GRCh38
rs959196546 Chr.13: 30752290-30752290 on Build GRCh38
rs202144118 Chr.13: 30752013-30752013 on Build GRCh38
rs759870460 Chr.13: 30752152-30752152 on Build GRCh38
rs550841128 Chr.13: 30752261-30752261 on Build GRCh38
rs756806011 Chr.13: 30752168-30752168 on Build GRCh38
rs547271298 Chr.13: 30752326-30752326 on Build GRCh38
rs752945827 Chr.13: 30752040-30752040 on Build GRCh38
rs974522655 Chr.13: 30752333-30752333 on Build GRCh38
rs148308449 Chr.13: 30752102-30752102 on Build GRCh38
rs966891374 Chr.13: 30752268-30752268 on Build GRCh38
rs773923945 Chr.13: 30752085-30752085 on Build GRCh38
rs4769874 Chr.13: 30752304-30752304 on Build GRCh38
rs949372273 Chr.13: 30752162-30752162 on Build GRCh38
rs200245621 Chr.13: 30752018-30752018 on Build GRCh38
rs940043127 Chr.13: 30752191-30752191 on Build GRCh38

View Full Product Details