Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32326129-32326661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6986698 c.587G>A p.S196N Chr.13: 32326569-32326569 on Build GRCh38
6957532 c.586A>C p.S196R Chr.13: 32326568-32326568 on Build GRCh38
3688676 c.472T>C p.S158P Chr.13: 32326147-32326147 on Build GRCh38
696753 c.559G>T p.E187* Chr.13: 32326541-32326541 on Build GRCh38
469351 c.565G>C p.D189H Chr.13: 32326547-32326547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs81002834 Chr.13: 32326300-32326300 on Build GRCh38
rs778866949 Chr.13: 32326176-32326176 on Build GRCh38
rs762445103 Chr.13: 32326211-32326211 on Build GRCh38
rs863224308 Chr.13: 32326166-32326166 on Build GRCh38
rs398122787 Chr.13: 32326255-32326255 on Build GRCh38
rs28897704 Chr.13: 32326609-32326609 on Build GRCh38
rs80358780 Chr.13: 32326541-32326541 on Build GRCh38
rs886040603 Chr.13: 32326548-32326549 on Build GRCh38
rs886038051 Chr.13: 32326252-32326252 on Build GRCh38
rs754219537 Chr.13: 32326383-32326383 on Build GRCh38
rs587782600 Chr.13: 32326252-32326254 on Build GRCh38
rs748625734 Chr.13: 32326220-32326220 on Build GRCh38
rs80358818 Chr.13: 32326569-32326569 on Build GRCh38
rs750385844 Chr.13: 32326505-32326505 on Build GRCh38
rs80358747 Chr.13: 32326503-32326503 on Build GRCh38
rs80358723 Chr.13: 32326262-32326262 on Build GRCh38
rs770207940 Chr.13: 32326635-32326635 on Build GRCh38
rs41293471 Chr.13: 32326602-32326602 on Build GRCh38
rs864622238 Chr.13: 32326551-32326551 on Build GRCh38
rs730881504 Chr.13: 32326155-32326155 on Build GRCh38
rs368799999 Chr.13: 32326336-32326336 on Build GRCh38
rs552306042 Chr.13: 32326394-32326394 on Build GRCh38
rs81002804 Chr.13: 32326495-32326495 on Build GRCh38
rs1024574221 Chr.13: 32326545-32326545 on Build GRCh38
rs397507340 Chr.13: 32326241-32326241 on Build GRCh38
rs765435155 Chr.13: 32326299-32326299 on Build GRCh38
rs397507768 Chr.13: 32326499-32326499 on Build GRCh38
rs80358805 Chr.13: 32326557-32326557 on Build GRCh38
rs767985386 Chr.13: 32326253-32326253 on Build GRCh38
rs983380958 Chr.13: 32326558-32326558 on Build GRCh38
rs1031272655 Chr.13: 32326365-32326365 on Build GRCh38
rs778528034 Chr.13: 32326575-32326575 on Build GRCh38
rs768171277 Chr.13: 32326218-32326218 on Build GRCh38
rs1056440334 Chr.13: 32326186-32326186 on Build GRCh38
rs398122793 Chr.13: 32326278-32326279 on Build GRCh38
rs878853593 Chr.13: 32326578-32326578 on Build GRCh38
rs773767096 Chr.13: 32326469-32326469 on Build GRCh38
rs786202156 Chr.13: 32326575-32326578 on Build GRCh38
rs80358730 Chr.13: 32326272-32326272 on Build GRCh38
rs772629465 Chr.13: 32326468-32326468 on Build GRCh38
rs397507747 Chr.13: 32326254-32326255 on Build GRCh38
rs81002876 Chr.13: 32326289-32326289 on Build GRCh38
rs897368704 Chr.13: 32326498-32326498 on Build GRCh38
rs773802339 Chr.13: 32326237-32326237 on Build GRCh38
rs80359560 Chr.13: 32326592-32326592 on Build GRCh38
rs11571622 Chr.13: 32326303-32326303 on Build GRCh38
rs886040926 Chr.13: 32326238-32326241 on Build GRCh38
rs397507763 Chr.13: 32326283-32326284 on Build GRCh38
rs200458964 Chr.13: 32326227-32326227 on Build GRCh38
rs431825331 Chr.13: 32326536-32326536 on Build GRCh38
rs371431745 Chr.13: 32326239-32326239 on Build GRCh38
rs587782398 Chr.13: 32326554-32326555 on Build GRCh38
rs773238258 Chr.13: 32326364-32326364 on Build GRCh38
rs80359510 Chr.13: 32326520-32326521 on Build GRCh38
rs864622305 Chr.13: 32326560-32326560 on Build GRCh38
rs397507359 Chr.13: 32326548-32326548 on Build GRCh38
rs56251699 Chr.13: 32326384-32326384 on Build GRCh38
rs886040620 Chr.13: 32326580-32326581 on Build GRCh38
rs80358810 Chr.13: 32326564-32326564 on Build GRCh38
rs979900660 Chr.13: 32326433-32326433 on Build GRCh38
rs80358761 Chr.13: 32326521-32326521 on Build GRCh38
rs397507832 Chr.13: 32326599-32326599 on Build GRCh38
rs397507766 Chr.13: 32326282-32326283 on Build GRCh38
rs81002897 Chr.13: 32326614-32326614 on Build GRCh38
rs276174857 Chr.13: 32326521-32326521 on Build GRCh38
rs755939798 Chr.13: 32326325-32326325 on Build GRCh38
rs759861270 Chr.13: 32326427-32326427 on Build GRCh38
rs397507840 Chr.13: 32326616-32326616 on Build GRCh38
rs886040606 Chr.13: 32326551-32326552 on Build GRCh38
rs276174848 Chr.13: 32326154-32326154 on Build GRCh38
rs933685346 Chr.13: 32326432-32326432 on Build GRCh38
rs781175758 Chr.13: 32326302-32326302 on Build GRCh38
rs779658258 Chr.13: 32326452-32326452 on Build GRCh38
rs876660394 Chr.13: 32326526-32326526 on Build GRCh38
rs80358717 Chr.13: 32326257-32326257 on Build GRCh38
rs527717043 Chr.13: 32326393-32326393 on Build GRCh38
rs781168884 Chr.13: 32326182-32326182 on Build GRCh38
rs747638859 Chr.13: 32326465-32326465 on Build GRCh38
rs397507830 Chr.13: 32326595-32326596 on Build GRCh38
rs786202001 Chr.13: 32326586-32326586 on Build GRCh38
rs397507841 Chr.13: 32326617-32326617 on Build GRCh38
rs886038052 Chr.13: 32326523-32326523 on Build GRCh38
rs80358775 Chr.13: 32326533-32326533 on Build GRCh38
rs80358871 Chr.13: 32326613-32326613 on Build GRCh38
rs863224593 Chr.13: 32326530-32326530 on Build GRCh38
rs876658364 Chr.13: 32326263-32326263 on Build GRCh38
rs81002884 Chr.13: 32326242-32326242 on Build GRCh38
rs779931795 Chr.13: 32326518-32326518 on Build GRCh38
rs876660795 Chr.13: 32326570-32326570 on Build GRCh38
rs276174849 Chr.13: 32326233-32326234 on Build GRCh38
rs11571623 Chr.13: 32326480-32326480 on Build GRCh38
rs779951024 Chr.13: 32326494-32326494 on Build GRCh38
rs1028102988 Chr.13: 32326167-32326167 on Build GRCh38
rs80358726 Chr.13: 32326268-32326268 on Build GRCh38
rs750805758 Chr.13: 32326267-32326267 on Build GRCh38
rs775202093 Chr.13: 32326205-32326205 on Build GRCh38
rs746899590 Chr.13: 32326375-32326375 on Build GRCh38
rs398122543 Chr.13: 32326580-32326580 on Build GRCh38
rs570584580 Chr.13: 32326410-32326410 on Build GRCh38
rs28897702 Chr.13: 32326500-32326500 on Build GRCh38
rs80359492 Chr.13: 32326500-32326500 on Build GRCh38
rs746963311 Chr.13: 32326154-32326154 on Build GRCh38
rs80358865 Chr.13: 32326605-32326605 on Build GRCh38
rs80359533 Chr.13: 32326556-32326557 on Build GRCh38
rs397507798 Chr.13: 32326554-32326554 on Build GRCh38
rs397507822 Chr.13: 32326584-32326584 on Build GRCh38
rs81002828 Chr.13: 32326488-32326488 on Build GRCh38
rs398122547 Chr.13: 32326590-32326590 on Build GRCh38
rs182828913 Chr.13: 32326296-32326296 on Build GRCh38
rs759965817 Chr.13: 32326631-32326631 on Build GRCh38
rs754678843 Chr.13: 32326543-32326543 on Build GRCh38
rs769389363 Chr.13: 32326199-32326199 on Build GRCh38
rs376582345 Chr.13: 32326577-32326577 on Build GRCh38
rs80359790 Chr.13: 32326282-32326282 on Build GRCh38
rs80358858 Chr.13: 32326601-32326601 on Build GRCh38
rs431825327 Chr.13: 32326281-32326281 on Build GRCh38
rs886040622 Chr.13: 32326577-32326580 on Build GRCh38
rs752881400 Chr.13: 32326496-32326496 on Build GRCh38
rs81002896 Chr.13: 32326292-32326292 on Build GRCh38
rs748993891 Chr.13: 32326457-32326457 on Build GRCh38
rs786204272 Chr.13: 32326223-32326223 on Build GRCh38
rs587782880 Chr.13: 32326238-32326241 on Build GRCh38
rs80359511 Chr.13: 32326521-32326522 on Build GRCh38
rs200930221 Chr.13: 32326461-32326461 on Build GRCh38
rs81002839 Chr.13: 32326483-32326483 on Build GRCh38
rs81002795 Chr.13: 32326153-32326153 on Build GRCh38
rs397507799 Chr.13: 32326554-32326554 on Build GRCh38
rs81002899 Chr.13: 32326615-32326615 on Build GRCh38
rs276174850 Chr.13: 32326233-32326233 on Build GRCh38
rs587781402 Chr.13: 32326581-32326581 on Build GRCh38
rs398122785 Chr.13: 32326238-32326238 on Build GRCh38
rs864622312 Chr.13: 32326550-32326550 on Build GRCh38
rs886040615 Chr.13: 32326565-32326565 on Build GRCh38
rs745653804 Chr.13: 32326190-32326190 on Build GRCh38
rs876659740 Chr.13: 32326248-32326248 on Build GRCh38
rs397507745 Chr.13: 32326249-32326249 on Build GRCh38
rs431825339 Chr.13: 32326620-32326620 on Build GRCh38
rs587780653 Chr.13: 32326250-32326250 on Build GRCh38
rs566374241 Chr.13: 32326440-32326440 on Build GRCh38
rs730881505 Chr.13: 32326571-32326571 on Build GRCh38
rs80359792 Chr.13: 32326531-32326531 on Build GRCh38
rs551045508 Chr.13: 32326353-32326353 on Build GRCh38
rs397507816 Chr.13: 32326581-32326582 on Build GRCh38
rs397507764 Chr.13: 32326284-32326284 on Build GRCh38
rs55698822 Chr.13: 32326164-32326164 on Build GRCh38
rs398122540 Chr.13: 32326555-32326555 on Build GRCh38
rs749053313 Chr.13: 32326532-32326532 on Build GRCh38
rs80358809 Chr.13: 32326563-32326563 on Build GRCh38
rs545312299 Chr.13: 32326340-32326340 on Build GRCh38
rs886040927 Chr.13: 32326283-32326283 on Build GRCh38
rs786201450 Chr.13: 32326259-32326259 on Build GRCh38
rs886040605 Chr.13: 32326550-32326557 on Build GRCh38
rs81002853 Chr.13: 32326240-32326240 on Build GRCh38
rs147022828 Chr.13: 32326244-32326244 on Build GRCh38
rs41293469 Chr.13: 32326502-32326502 on Build GRCh38
rs397507802 Chr.13: 32326556-32326557 on Build GRCh38
rs876659940 Chr.13: 32326572-32326572 on Build GRCh38
rs41293467 Chr.13: 32326271-32326271 on Build GRCh38
rs81002858 Chr.13: 32326497-32326497 on Build GRCh38
rs398122795 Chr.13: 32326487-32326487 on Build GRCh38
rs120074205 Chr.13: 32326255-32326256 on Build GRCh38
rs886038053 Chr.13: 32326566-32326566 on Build GRCh38
rs886040563 Chr.13: 32326272-32326272 on Build GRCh38
rs28897703 Chr.13: 32326516-32326516 on Build GRCh38
rs397507762 Chr.13: 32326283-32326283 on Build GRCh38
rs81002849 Chr.13: 32326498-32326498 on Build GRCh38
rs786202685 Chr.13: 32326527-32326527 on Build GRCh38
rs768257523 Chr.13: 32326459-32326459 on Build GRCh38
rs747726394 Chr.13: 32326588-32326588 on Build GRCh38

View Full Product Details