Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40933715-40934224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF1
Entrez Gene ID
1997
Gene Name
E74 like ETS transcription factor 1
Gene Aliases
-
UniGene
Hs.135646
Gene Chromosome Location
Chr.13: 40931919-41061414 on Build GRCh38
Transcripts
NM_001145353.1 NM_172373.3 XM_005266276.1 XM_005266277.3 XM_011534950.1 XM_011534951.2 XM_017020409.1 XM_017020410.1 XM_017020411.1 XM_017020412.1 XM_017020413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
306124 c.1561A>G p.S521G Chr.13: 40933724-40933724 on Build GRCh38
4416227 c.1372T>C p.S458P Chr.13: 40933913-40933913 on Build GRCh38
6490093 c.1284A>T p.P428P Chr.13: 40934001-40934001 on Build GRCh38
1940686 c.1514A>T p.Q505L Chr.13: 40933771-40933771 on Build GRCh38
6074681 c.1460A>T p.Q487L Chr.13: 40933825-40933825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745958024 Chr.13: 40933882-40933882 on Build GRCh38
rs149160961 Chr.13: 40933807-40933807 on Build GRCh38
rs544414935 Chr.13: 40933759-40933759 on Build GRCh38
rs375157134 Chr.13: 40934171-40934171 on Build GRCh38
rs752471658 Chr.13: 40933961-40933961 on Build GRCh38
rs747475380 Chr.13: 40934032-40934032 on Build GRCh38
rs760208850 Chr.13: 40933925-40933925 on Build GRCh38
rs140720559 Chr.13: 40933884-40933884 on Build GRCh38
rs987371423 Chr.13: 40933800-40933800 on Build GRCh38
rs146846076 Chr.13: 40933874-40933874 on Build GRCh38
rs777083535 Chr.13: 40933796-40933796 on Build GRCh38
rs1028894276 Chr.13: 40934174-40934174 on Build GRCh38
rs759194622 Chr.13: 40934062-40934062 on Build GRCh38
rs376481585 Chr.13: 40933810-40933810 on Build GRCh38
rs917326027 Chr.13: 40934117-40934117 on Build GRCh38
rs540284464 Chr.13: 40933930-40933930 on Build GRCh38
rs771601333 Chr.13: 40934056-40934058 on Build GRCh38
rs769160866 Chr.13: 40933891-40933891 on Build GRCh38
rs371815642 Chr.13: 40933918-40933918 on Build GRCh38
rs199564946 Chr.13: 40933931-40933931 on Build GRCh38
rs1047811070 Chr.13: 40934098-40934098 on Build GRCh38
rs200709823 Chr.13: 40933735-40933735 on Build GRCh38
rs755181758 Chr.13: 40933995-40933995 on Build GRCh38
rs766060383 Chr.13: 40933927-40933927 on Build GRCh38
rs750520786 Chr.13: 40933987-40933987 on Build GRCh38
rs41306656 Chr.13: 40933913-40933913 on Build GRCh38
rs369423492 Chr.13: 40933830-40933830 on Build GRCh38
rs375140290 Chr.13: 40933967-40933967 on Build GRCh38
rs1033120802 Chr.13: 40933935-40933935 on Build GRCh38
rs899027469 Chr.13: 40933833-40933833 on Build GRCh38
rs372346993 Chr.13: 40933871-40933871 on Build GRCh38
rs1015171884 Chr.13: 40934087-40934087 on Build GRCh38
rs748531706 Chr.13: 40934012-40934012 on Build GRCh38
rs368042351 Chr.13: 40933990-40933990 on Build GRCh38
rs746092397 Chr.13: 40933790-40933790 on Build GRCh38
rs778799774 Chr.13: 40933753-40933753 on Build GRCh38
rs976068572 Chr.13: 40933767-40933767 on Build GRCh38
rs779209874 Chr.13: 40934006-40934006 on Build GRCh38
rs965963878 Chr.13: 40934096-40934096 on Build GRCh38
rs745845554 Chr.13: 40933797-40933798 on Build GRCh38
rs759955495 Chr.13: 40933851-40933851 on Build GRCh38
rs776064185 Chr.13: 40934054-40934054 on Build GRCh38
rs776066204 Chr.13: 40933803-40933803 on Build GRCh38
rs754738116 Chr.13: 40933856-40933856 on Build GRCh38
rs781286344 Chr.13: 40933787-40933787 on Build GRCh38
rs1020667557 Chr.13: 40933965-40933965 on Build GRCh38
rs764868700 Chr.13: 40934070-40934070 on Build GRCh38
rs770947022 Chr.13: 40933786-40933786 on Build GRCh38
rs749531568 Chr.13: 40933994-40933994 on Build GRCh38
rs762228481 Chr.13: 40933908-40933908 on Build GRCh38
rs79846574 Chr.13: 40933880-40933880 on Build GRCh38
rs1001223538 Chr.13: 40933921-40933921 on Build GRCh38
rs953378607 Chr.13: 40934184-40934185 on Build GRCh38
rs765000740 Chr.13: 40933940-40933940 on Build GRCh38
rs766946264 Chr.13: 40933813-40933813 on Build GRCh38
rs921953794 Chr.13: 40933805-40933805 on Build GRCh38
rs138161971 Chr.13: 40933812-40933812 on Build GRCh38
rs145131167 Chr.13: 40933857-40933857 on Build GRCh38
rs552484372 Chr.13: 40933941-40933941 on Build GRCh38
rs770451571 Chr.13: 40934036-40934036 on Build GRCh38
rs765031385 Chr.13: 40933952-40933952 on Build GRCh38
rs764082039 Chr.13: 40933975-40933975 on Build GRCh38
rs145860913 Chr.13: 40933898-40933898 on Build GRCh38
rs766003101 Chr.13: 40933852-40933852 on Build GRCh38
rs987840910 Chr.13: 40933944-40933944 on Build GRCh38
rs753681912 Chr.13: 40933853-40933853 on Build GRCh38
rs772503411 Chr.13: 40934024-40934024 on Build GRCh38
rs780405648 Chr.13: 40933886-40933886 on Build GRCh38
rs756235921 Chr.13: 40933988-40933988 on Build GRCh38
rs749478443 Chr.13: 40933890-40933890 on Build GRCh38
rs545755055 Chr.13: 40934170-40934170 on Build GRCh38
rs749880705 Chr.13: 40933820-40933820 on Build GRCh38
rs1023178685 Chr.13: 40933897-40933897 on Build GRCh38

View Full Product Details