Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49308494-49308998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAB39L
Entrez Gene ID
81617
Gene Name
calcium binding protein 39 like
Gene Aliases
MO25-BETA, MO2L, bA103J18.3
UniGene
Hs.742971
Gene Chromosome Location
Chr.13: 49308628-49444346 on Build GRCh38
Transcripts
NM_001079670.2 NM_001287337.1 NM_001287338.1 NM_001287339.1 NM_030925.3 XM_011535254.2 XM_011535255.2 XM_011535256.2 XM_017020785.1 XM_017020786.1 XM_017020787.1 XM_017020788.1

SNP Detail

SNP ID Chromosome Location
rs542111112 Chr.13: 49308692-49308692 on Build GRCh38
rs747958032 Chr.13: 49308590-49308590 on Build GRCh38
rs962517295 Chr.13: 49308768-49308768 on Build GRCh38
rs754731781 Chr.13: 49308875-49308875 on Build GRCh38
rs772328484 Chr.13: 49308589-49308589 on Build GRCh38
rs1035072745 Chr.13: 49308594-49308594 on Build GRCh38
rs894132472 Chr.13: 49308655-49308655 on Build GRCh38
rs957542423 Chr.13: 49308749-49308749 on Build GRCh38
rs971200292 Chr.13: 49308511-49308511 on Build GRCh38
rs905470445 Chr.13: 49308549-49308549 on Build GRCh38
rs1022754259 Chr.13: 49308771-49308771 on Build GRCh38
rs770454111 Chr.13: 49308695-49308695 on Build GRCh38
rs953617503 Chr.13: 49308565-49308565 on Build GRCh38
rs184116939 Chr.13: 49308804-49308804 on Build GRCh38
rs905377318 Chr.13: 49308934-49308934 on Build GRCh38
rs998656810 Chr.13: 49308720-49308720 on Build GRCh38
rs972245244 Chr.13: 49308800-49308800 on Build GRCh38
rs916107932 Chr.13: 49308886-49308886 on Build GRCh38
rs189436281 Chr.13: 49308909-49308909 on Build GRCh38
rs367595697 Chr.13: 49308792-49308792 on Build GRCh38
rs986092337 Chr.13: 49308781-49308781 on Build GRCh38
rs934394613 Chr.13: 49308947-49308947 on Build GRCh38
rs527663119 Chr.13: 49308923-49308923 on Build GRCh38
rs748662204 Chr.13: 49308513-49308513 on Build GRCh38
rs955443131 Chr.13: 49308780-49308780 on Build GRCh38
rs202041140 Chr.13: 49308605-49308605 on Build GRCh38
rs577931394 Chr.13: 49308586-49308586 on Build GRCh38
rs567566030 Chr.13: 49308971-49308971 on Build GRCh38
rs986208614 Chr.13: 49308566-49308566 on Build GRCh38
rs557867079 Chr.13: 49308536-49308536 on Build GRCh38
rs930890756 Chr.13: 49308599-49308599 on Build GRCh38
rs760829840 Chr.13: 49308713-49308713 on Build GRCh38
rs766359424 Chr.13: 49308816-49308816 on Build GRCh38
rs915351242 Chr.13: 49308854-49308854 on Build GRCh38
rs1052915233 Chr.13: 49308965-49308965 on Build GRCh38
rs537382483 Chr.13: 49308772-49308772 on Build GRCh38
rs369660027 Chr.13: 49308803-49308803 on Build GRCh38
rs1049233087 Chr.13: 49308654-49308654 on Build GRCh38
rs751303129 Chr.13: 49308831-49308831 on Build GRCh38
rs549390848 Chr.13: 49308966-49308966 on Build GRCh38
rs41284774 Chr.13: 49308777-49308777 on Build GRCh38
rs919522914 Chr.13: 49308593-49308593 on Build GRCh38
rs1011948997 Chr.13: 49308968-49308968 on Build GRCh38
rs573150592 Chr.13: 49308547-49308547 on Build GRCh38
rs1031441071 Chr.13: 49308733-49308733 on Build GRCh38
rs188626843 Chr.13: 49308519-49308519 on Build GRCh38
rs140433453 Chr.13: 49308539-49308539 on Build GRCh38
rs560457613 Chr.13: 49308892-49308892 on Build GRCh38
rs181014819 Chr.13: 49308918-49308918 on Build GRCh38
rs116798358 Chr.13: 49308690-49308690 on Build GRCh38
rs762852427 Chr.13: 49308801-49308801 on Build GRCh38
rs372395128 Chr.13: 49308911-49308911 on Build GRCh38
rs574311314 Chr.13: 49308915-49308915 on Build GRCh38
rs530775183 Chr.13: 49308791-49308791 on Build GRCh38
rs193260175 Chr.13: 49308769-49308769 on Build GRCh38
rs1031555637 Chr.13: 49308531-49308531 on Build GRCh38
rs1004161881 Chr.13: 49308952-49308952 on Build GRCh38

View Full Product Details