Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51362151-51362681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS6
Entrez Gene ID
26512
Gene Name
integrator complex subunit 6
Gene Aliases
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2
UniGene
Hs.439440
Gene Chromosome Location
Chr.13: 51353171-51453236 on Build GRCh38
Transcripts
NM_001039937.1 NM_001306091.1 NM_012141.2 XM_011535040.2 XM_011535041.1 XM_017020540.1 XM_017020707.1

Gene Symbol
SERPINE3
Entrez Gene ID
647174
Gene Name
serpin family E member 3
Gene Aliases
-
UniGene
Hs.685891
Gene Chromosome Location
Chr.13: 51334976-51362391 on Build GRCh38
Transcripts
NM_001039937.1 NM_001306091.1 NM_012141.2 XM_011535040.2 XM_011535041.1 XM_017020540.1 XM_017020707.1

SNP Detail

SNP ID Chromosome Location
rs1001728196 Chr.13: 51362226-51362226 on Build GRCh38
rs905391335 Chr.13: 51362475-51362475 on Build GRCh38
rs569660247 Chr.13: 51362435-51362435 on Build GRCh38
rs562782299 Chr.13: 51362354-51362354 on Build GRCh38
rs140059943 Chr.13: 51362491-51362491 on Build GRCh38
rs1001255695 Chr.13: 51362483-51362483 on Build GRCh38
rs143695462 Chr.13: 51362592-51362592 on Build GRCh38
rs1027456051 Chr.13: 51362602-51362602 on Build GRCh38
rs1055738754 Chr.13: 51362578-51362578 on Build GRCh38
rs968477376 Chr.13: 51362191-51362191 on Build GRCh38
rs752375930 Chr.13: 51362569-51362569 on Build GRCh38
rs9596507 Chr.13: 51362287-51362287 on Build GRCh38
rs544576648 Chr.13: 51362352-51362352 on Build GRCh38
rs17837216 Chr.13: 51362499-51362499 on Build GRCh38
rs752596711 Chr.13: 51362372-51362372 on Build GRCh38
rs532835643 Chr.13: 51362480-51362480 on Build GRCh38
rs959675663 Chr.13: 51362497-51362497 on Build GRCh38
rs953339610 Chr.13: 51362604-51362604 on Build GRCh38
rs1034009830 Chr.13: 51362256-51362256 on Build GRCh38
rs112585939 Chr.13: 51362375-51362375 on Build GRCh38
rs779339681 Chr.13: 51362538-51362538 on Build GRCh38
rs1033837527 Chr.13: 51362309-51362309 on Build GRCh38
rs17837215 Chr.13: 51362268-51362268 on Build GRCh38
rs186014183 Chr.13: 51362338-51362338 on Build GRCh38
rs1014567940 Chr.13: 51362557-51362557 on Build GRCh38
rs752038083 Chr.13: 51362422-51362422 on Build GRCh38
rs146493327 Chr.13: 51362429-51362429 on Build GRCh38
rs958732761 Chr.13: 51362321-51362321 on Build GRCh38
rs538773684 Chr.13: 51362470-51362470 on Build GRCh38
rs1026611512 Chr.13: 51362335-51362335 on Build GRCh38
rs913916064 Chr.13: 51362430-51362430 on Build GRCh38
rs1043895844 Chr.13: 51362472-51362472 on Build GRCh38
rs554603715 Chr.13: 51362193-51362193 on Build GRCh38
rs750867762 Chr.13: 51362374-51362374 on Build GRCh38
rs936791934 Chr.13: 51362398-51362398 on Build GRCh38
rs144113161 Chr.13: 51362403-51362403 on Build GRCh38
rs777378780 Chr.13: 51362626-51362626 on Build GRCh38
rs907814744 Chr.13: 51362315-51362315 on Build GRCh38
rs921099493 Chr.13: 51362392-51362392 on Build GRCh38
rs944710262 Chr.13: 51362387-51362387 on Build GRCh38
rs961934214 Chr.13: 51362576-51362576 on Build GRCh38
rs548394607 Chr.13: 51362464-51362464 on Build GRCh38
rs1011184370 Chr.13: 51362444-51362444 on Build GRCh38
rs892711294 Chr.13: 51362436-51362436 on Build GRCh38
rs368224765 Chr.13: 51362322-51362322 on Build GRCh38
rs906720295 Chr.13: 51362281-51362281 on Build GRCh38
rs568698099 Chr.13: 51362341-51362341 on Build GRCh38
rs912447660 Chr.13: 51362369-51362369 on Build GRCh38
rs1026665225 Chr.13: 51362642-51362642 on Build GRCh38
rs554569721 Chr.13: 51362636-51362636 on Build GRCh38
rs995226050 Chr.13: 51362609-51362609 on Build GRCh38

View Full Product Details