Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77698346-77698864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLAIN1
Entrez Gene ID
122060
Gene Name
SLAIN motif family member 1
Gene Aliases
C13orf32
UniGene
Hs.349955
Gene Chromosome Location
Chr.13: 77697854-77764242 on Build GRCh38
Transcripts
NM_001242868.1 XM_011534929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7258796 c.388C>T p.P130S Chr.13: 77698367-77698367 on Build GRCh38
5747495 c.401T>C p.F134S Chr.13: 77698380-77698380 on Build GRCh38
6287680 c.539G>T p.R180L Chr.13: 77698518-77698518 on Build GRCh38
5761753 c.477G>T p.S159S Chr.13: 77698456-77698456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772395032 Chr.13: 77698454-77698455 on Build GRCh38
rs956672792 Chr.13: 77698702-77698702 on Build GRCh38
rs183900611 Chr.13: 77698771-77698771 on Build GRCh38
rs892665912 Chr.13: 77698566-77698566 on Build GRCh38
rs777190816 Chr.13: 77698825-77698825 on Build GRCh38
rs879541378 Chr.13: 77698551-77698551 on Build GRCh38
rs919115016 Chr.13: 77698431-77698431 on Build GRCh38
rs556471320 Chr.13: 77698786-77698786 on Build GRCh38
rs904831431 Chr.13: 77698606-77698606 on Build GRCh38
rs964642456 Chr.13: 77698831-77698831 on Build GRCh38
rs942719032 Chr.13: 77698460-77698460 on Build GRCh38
rs951878489 Chr.13: 77698619-77698619 on Build GRCh38
rs935354765 Chr.13: 77698397-77698397 on Build GRCh38
rs577981200 Chr.13: 77698835-77698835 on Build GRCh38
rs914030946 Chr.13: 77698756-77698756 on Build GRCh38
rs913150057 Chr.13: 77698716-77698716 on Build GRCh38
rs762046528 Chr.13: 77698547-77698547 on Build GRCh38
rs965021649 Chr.13: 77698651-77698651 on Build GRCh38
rs1052930435 Chr.13: 77698417-77698417 on Build GRCh38
rs1045556554 Chr.13: 77698719-77698719 on Build GRCh38
rs1049252450 Chr.13: 77698774-77698774 on Build GRCh38
rs768940437 Chr.13: 77698446-77698446 on Build GRCh38
rs905595444 Chr.13: 77698571-77698571 on Build GRCh38
rs1004557794 Chr.13: 77698609-77698609 on Build GRCh38
rs1002320305 Chr.13: 77698409-77698409 on Build GRCh38
rs762291430 Chr.13: 77698496-77698496 on Build GRCh38
rs774847974 Chr.13: 77698455-77698455 on Build GRCh38
rs951928774 Chr.13: 77698440-77698440 on Build GRCh38
rs922705222 Chr.13: 77698518-77698518 on Build GRCh38
rs566994968 Chr.13: 77698390-77698390 on Build GRCh38
rs763254872 Chr.13: 77698428-77698428 on Build GRCh38
rs887567705 Chr.13: 77698785-77698785 on Build GRCh38
rs925249460 Chr.13: 77698388-77698388 on Build GRCh38
rs969043748 Chr.13: 77698400-77698400 on Build GRCh38
rs573914258 Chr.13: 77698747-77698747 on Build GRCh38
rs975439147 Chr.13: 77698681-77698681 on Build GRCh38
rs1034782204 Chr.13: 77698410-77698410 on Build GRCh38
rs1029601646 Chr.13: 77698628-77698628 on Build GRCh38
rs997664410 Chr.13: 77698620-77698620 on Build GRCh38
rs946891175 Chr.13: 77698770-77698770 on Build GRCh38
rs538417359 Chr.13: 77698395-77698395 on Build GRCh38
rs902081049 Chr.13: 77698823-77698823 on Build GRCh38
rs376955917 Chr.13: 77698817-77698817 on Build GRCh38
rs953587355 Chr.13: 77698653-77698653 on Build GRCh38
rs979904184 Chr.13: 77698666-77698666 on Build GRCh38
rs988137469 Chr.13: 77698711-77698711 on Build GRCh38
rs949994734 Chr.13: 77698718-77698718 on Build GRCh38
rs937192148 Chr.13: 77698772-77698772 on Build GRCh38
rs1017540629 Chr.13: 77698648-77698648 on Build GRCh38
rs934111393 Chr.13: 77698544-77698544 on Build GRCh38
rs993224466 Chr.13: 77698778-77698778 on Build GRCh38
rs867992630 Chr.13: 77698444-77698444 on Build GRCh38
rs1011138884 Chr.13: 77698383-77698383 on Build GRCh38
rs925395622 Chr.13: 77698692-77698692 on Build GRCh38
rs534032274 Chr.13: 77698602-77698602 on Build GRCh38
rs555738841 Chr.13: 77698705-77698705 on Build GRCh38
rs935773 Chr.13: 77698550-77698550 on Build GRCh38
rs896219090 Chr.13: 77698612-77698612 on Build GRCh38
rs750439184 Chr.13: 77698517-77698517 on Build GRCh38
rs761003905 Chr.13: 77698536-77698536 on Build GRCh38
rs1015175634 Chr.13: 77698614-77698614 on Build GRCh38
rs887400417 Chr.13: 77698607-77698607 on Build GRCh38
rs1001720034 Chr.13: 77698579-77698579 on Build GRCh38
rs1036050789 Chr.13: 77698797-77698797 on Build GRCh38
rs1040160089 Chr.13: 77698463-77698463 on Build GRCh38
rs984668325 Chr.13: 77698620-77698620 on Build GRCh38
rs573087056 Chr.13: 77698782-77698782 on Build GRCh38
rs966321674 Chr.13: 77698615-77698615 on Build GRCh38
rs1035138368 Chr.13: 77698611-77698611 on Build GRCh38
rs938286556 Chr.13: 77698773-77698773 on Build GRCh38
rs1056118409 Chr.13: 77698773-77698773 on Build GRCh38
rs754616290 Chr.13: 77698782-77698784 on Build GRCh38
rs767649882 Chr.13: 77698504-77698504 on Build GRCh38

View Full Product Details