Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108307126-108307628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF13B
Entrez Gene ID
10673
Gene Name
tumor necrosis factor superfamily member 13b
Gene Aliases
BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, TNLG7A, ZTNF4
UniGene
Hs.525157
Gene Chromosome Location
Chr.13: 108268240-108308484 on Build GRCh38
Transcripts
NM_001145645.2 NM_006573.4

SNP Detail

SNP ID Chromosome Location
rs1015490738 Chr.13: 108307346-108307346 on Build GRCh38
rs533385212 Chr.13: 108307235-108307235 on Build GRCh38
rs903880416 Chr.13: 108307549-108307549 on Build GRCh38
rs1004553614 Chr.13: 108307339-108307339 on Build GRCh38
rs989194091 Chr.13: 108307482-108307482 on Build GRCh38
rs141131834 Chr.13: 108307597-108307598 on Build GRCh38
rs980395389 Chr.13: 108307310-108307310 on Build GRCh38
rs868765580 Chr.13: 108307277-108307277 on Build GRCh38
rs560774398 Chr.13: 108307328-108307328 on Build GRCh38
rs777841374 Chr.13: 108307174-108307174 on Build GRCh38
rs942801483 Chr.13: 108307447-108307447 on Build GRCh38
rs1030820984 Chr.13: 108307175-108307175 on Build GRCh38
rs1049055837 Chr.13: 108307318-108307318 on Build GRCh38
rs966380658 Chr.13: 108307524-108307524 on Build GRCh38
rs777031089 Chr.13: 108307451-108307451 on Build GRCh38
rs906385440 Chr.13: 108307376-108307376 on Build GRCh38
rs759864756 Chr.13: 108307507-108307507 on Build GRCh38
rs962981323 Chr.13: 108307333-108307333 on Build GRCh38
rs892384876 Chr.13: 108307577-108307577 on Build GRCh38
rs912065990 Chr.13: 108307246-108307246 on Build GRCh38
rs371923168 Chr.13: 108307280-108307280 on Build GRCh38
rs529461431 Chr.13: 108307344-108307344 on Build GRCh38
rs1039124647 Chr.13: 108307294-108307294 on Build GRCh38
rs1015919803 Chr.13: 108307320-108307320 on Build GRCh38
rs936698417 Chr.13: 108307560-108307560 on Build GRCh38
rs978340171 Chr.13: 108307196-108307196 on Build GRCh38
rs758126391 Chr.13: 108307317-108307317 on Build GRCh38
rs1033467498 Chr.13: 108307401-108307401 on Build GRCh38
rs185198828 Chr.13: 108307392-108307392 on Build GRCh38
rs766122602 Chr.13: 108307563-108307563 on Build GRCh38
rs944874741 Chr.13: 108307282-108307282 on Build GRCh38
rs1048176073 Chr.13: 108307155-108307155 on Build GRCh38
rs747226662 Chr.13: 108307342-108307342 on Build GRCh38
rs115788212 Chr.13: 108307523-108307523 on Build GRCh38
rs549357865 Chr.13: 108307383-108307383 on Build GRCh38
rs892264626 Chr.13: 108307181-108307181 on Build GRCh38
rs1024639317 Chr.13: 108307254-108307254 on Build GRCh38
rs193216832 Chr.13: 108307223-108307223 on Build GRCh38
rs912645013 Chr.13: 108307424-108307424 on Build GRCh38
rs978774023 Chr.13: 108307559-108307559 on Build GRCh38
rs930687293 Chr.13: 108307308-108307308 on Build GRCh38
rs969115876 Chr.13: 108307301-108307301 on Build GRCh38
rs749892447 Chr.13: 108307316-108307316 on Build GRCh38
rs367978634 Chr.13: 108307493-108307493 on Build GRCh38
rs547277438 Chr.13: 108307259-108307259 on Build GRCh38
rs986975265 Chr.13: 108307413-108307413 on Build GRCh38

View Full Product Details