Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23048824-23049321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH24
Entrez Gene ID
64403
Gene Name
cadherin 24
Gene Aliases
CDH11L
UniGene
Hs.155912
Gene Chromosome Location
Chr.14: 23047061-23057619 on Build GRCh38
Transcripts
NM_022478.3 NM_144985.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025458 c.1817C>T p.A606V Chr.14: 23049056-23049056 on Build GRCh38
158693 c.1656C>A p.Y552* Chr.14: 23049217-23049217 on Build GRCh38
6025457 c.1931C>T p.A644V Chr.14: 23049056-23049056 on Build GRCh38
3495087 c.1826C>T p.T609I Chr.14: 23049047-23049047 on Build GRCh38
3495086 c.1940C>T p.T647I Chr.14: 23049047-23049047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780584284 Chr.14: 23049169-23049169 on Build GRCh38
rs200375242 Chr.14: 23048979-23048979 on Build GRCh38
rs766791521 Chr.14: 23049040-23049040 on Build GRCh38
rs776898472 Chr.14: 23049236-23049236 on Build GRCh38
rs778683269 Chr.14: 23048995-23048995 on Build GRCh38
rs377668553 Chr.14: 23049199-23049199 on Build GRCh38
rs755731670 Chr.14: 23049268-23049268 on Build GRCh38
rs569750230 Chr.14: 23049004-23049004 on Build GRCh38
rs944403523 Chr.14: 23048969-23048969 on Build GRCh38
rs150669817 Chr.14: 23048868-23048868 on Build GRCh38
rs756479067 Chr.14: 23048987-23048987 on Build GRCh38
rs549916614 Chr.14: 23048931-23048931 on Build GRCh38
rs752646571 Chr.14: 23049267-23049267 on Build GRCh38
rs78717454 Chr.14: 23048858-23048858 on Build GRCh38
rs144187499 Chr.14: 23048927-23048927 on Build GRCh38
rs748288705 Chr.14: 23049291-23049291 on Build GRCh38
rs754898378 Chr.14: 23049246-23049246 on Build GRCh38
rs751042082 Chr.14: 23049227-23049227 on Build GRCh38
rs374416201 Chr.14: 23049042-23049042 on Build GRCh38
rs767559098 Chr.14: 23049084-23049084 on Build GRCh38
rs762021060 Chr.14: 23049239-23049239 on Build GRCh38
rs8006527 Chr.14: 23048859-23048859 on Build GRCh38
rs754028783 Chr.14: 23049126-23049126 on Build GRCh38
rs767610187 Chr.14: 23048981-23048981 on Build GRCh38
rs200046002 Chr.14: 23049067-23049067 on Build GRCh38
rs754190038 Chr.14: 23049294-23049295 on Build GRCh38
rs375821742 Chr.14: 23049179-23049179 on Build GRCh38
rs764139008 Chr.14: 23049231-23049231 on Build GRCh38
rs750170156 Chr.14: 23049001-23049001 on Build GRCh38
rs374708717 Chr.14: 23049140-23049140 on Build GRCh38
rs547673515 Chr.14: 23048900-23048900 on Build GRCh38
rs760073680 Chr.14: 23049079-23049079 on Build GRCh38
rs760900809 Chr.14: 23049102-23049102 on Build GRCh38
rs570462960 Chr.14: 23048922-23048922 on Build GRCh38
rs764261647 Chr.14: 23049124-23049124 on Build GRCh38
rs775572481 Chr.14: 23049101-23049101 on Build GRCh38
rs756323654 Chr.14: 23048994-23048994 on Build GRCh38
rs566103988 Chr.14: 23049229-23049229 on Build GRCh38
rs779897619 Chr.14: 23049021-23049021 on Build GRCh38
rs200471169 Chr.14: 23049066-23049066 on Build GRCh38
rs186404113 Chr.14: 23049279-23049279 on Build GRCh38
rs555921275 Chr.14: 23049178-23049178 on Build GRCh38
rs780752199 Chr.14: 23049062-23049062 on Build GRCh38
rs981367238 Chr.14: 23048851-23048851 on Build GRCh38
rs778969842 Chr.14: 23049070-23049070 on Build GRCh38
rs907643960 Chr.14: 23049294-23049294 on Build GRCh38
rs1046828800 Chr.14: 23048855-23048855 on Build GRCh38
rs767444570 Chr.14: 23049261-23049261 on Build GRCh38
rs777604682 Chr.14: 23049182-23049182 on Build GRCh38
rs558001433 Chr.14: 23049297-23049297 on Build GRCh38
rs201573362 Chr.14: 23049141-23049141 on Build GRCh38
rs754507800 Chr.14: 23049052-23049052 on Build GRCh38
rs373998017 Chr.14: 23049245-23049245 on Build GRCh38
rs141827332 Chr.14: 23049063-23049063 on Build GRCh38
rs748923700 Chr.14: 23049187-23049187 on Build GRCh38
rs900442061 Chr.14: 23049049-23049049 on Build GRCh38
rs746365911 Chr.14: 23049222-23049222 on Build GRCh38
rs1021492649 Chr.14: 23049135-23049135 on Build GRCh38
rs973842177 Chr.14: 23049181-23049181 on Build GRCh38
rs374252336 Chr.14: 23049127-23049127 on Build GRCh38
rs774772546 Chr.14: 23049206-23049206 on Build GRCh38
rs1041917435 Chr.14: 23049036-23049036 on Build GRCh38
rs760784695 Chr.14: 23049230-23049230 on Build GRCh38
rs754880907 Chr.14: 23049165-23049165 on Build GRCh38
rs772573589 Chr.14: 23049226-23049226 on Build GRCh38
rs867134254 Chr.14: 23049225-23049225 on Build GRCh38
rs753610405 Chr.14: 23049285-23049285 on Build GRCh38
rs746943790 Chr.14: 23049037-23049037 on Build GRCh38
rs771389147 Chr.14: 23049069-23049069 on Build GRCh38

View Full Product Details