Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23351660-23352151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A17
Entrez Gene ID
51310
Gene Name
solute carrier family 22 member 17
Gene Aliases
24p3R, BOCT, BOIT, NGALR, NGALR2, NGALR3, hBOIT
UniGene
Hs.373498
Gene Chromosome Location
Chr.14: 23346304-23354991 on Build GRCh38
Transcripts
NM_001289050.1 NM_016609.4 NM_020372.3 NR_110290.1 XM_005267747.4 XM_005267748.4 XM_017021361.1 XM_017021362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4797717 c.251C>T p.T84I Chr.14: 23351964-23351964 on Build GRCh38
6360194 c.82G>A p.E28K Chr.14: 23352133-23352133 on Build GRCh38
6258499 c.285C>A p.D95E Chr.14: 23351838-23351838 on Build GRCh38
7390225 c.256G>A p.A86T Chr.14: 23351959-23351959 on Build GRCh38
2152406 c.363C>T p.P121P Chr.14: 23351760-23351760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778604088 Chr.14: 23351910-23351910 on Build GRCh38
rs750305340 Chr.14: 23351778-23351778 on Build GRCh38
rs773457957 Chr.14: 23351930-23351930 on Build GRCh38
rs375707398 Chr.14: 23352077-23352077 on Build GRCh38
rs752733577 Chr.14: 23351955-23351955 on Build GRCh38
rs777150704 Chr.14: 23351718-23351718 on Build GRCh38
rs755648828 Chr.14: 23352109-23352109 on Build GRCh38
rs759660181 Chr.14: 23351942-23351942 on Build GRCh38
rs775943931 Chr.14: 23352131-23352131 on Build GRCh38
rs746585584 Chr.14: 23351919-23351919 on Build GRCh38
rs747617414 Chr.14: 23352023-23352023 on Build GRCh38
rs777063932 Chr.14: 23351745-23351745 on Build GRCh38
rs777390383 Chr.14: 23351984-23351986 on Build GRCh38
rs986296633 Chr.14: 23351807-23351807 on Build GRCh38
rs766207590 Chr.14: 23352079-23352079 on Build GRCh38
rs752893070 Chr.14: 23351902-23351902 on Build GRCh38
rs376977274 Chr.14: 23351784-23351784 on Build GRCh38
rs1036146411 Chr.14: 23351924-23351924 on Build GRCh38
rs765678245 Chr.14: 23351846-23351846 on Build GRCh38
rs367840234 Chr.14: 23351985-23351985 on Build GRCh38
rs996121282 Chr.14: 23352049-23352049 on Build GRCh38
rs1046288857 Chr.14: 23352003-23352003 on Build GRCh38
rs756278749 Chr.14: 23351907-23351907 on Build GRCh38
rs376360400 Chr.14: 23352106-23352106 on Build GRCh38
rs1053719694 Chr.14: 23351923-23351923 on Build GRCh38
rs754993643 Chr.14: 23352088-23352088 on Build GRCh38
rs143160984 Chr.14: 23351840-23351840 on Build GRCh38
rs148898539 Chr.14: 23352042-23352042 on Build GRCh38
rs774265715 Chr.14: 23351705-23351705 on Build GRCh38
rs750166775 Chr.14: 23351913-23351913 on Build GRCh38
rs760785974 Chr.14: 23352070-23352070 on Build GRCh38
rs1027278203 Chr.14: 23352094-23352094 on Build GRCh38
rs546469474 Chr.14: 23351796-23351796 on Build GRCh38
rs758209318 Chr.14: 23351991-23351991 on Build GRCh38
rs563074566 Chr.14: 23351841-23351841 on Build GRCh38
rs533654599 Chr.14: 23351685-23351685 on Build GRCh38
rs374337000 Chr.14: 23351838-23351838 on Build GRCh38
rs922911402 Chr.14: 23351766-23351766 on Build GRCh38
rs1009698618 Chr.14: 23351728-23351728 on Build GRCh38
rs750867112 Chr.14: 23351737-23351737 on Build GRCh38
rs943838172 Chr.14: 23351843-23351843 on Build GRCh38
rs762257651 Chr.14: 23351734-23351734 on Build GRCh38
rs751329853 Chr.14: 23352086-23352086 on Build GRCh38
rs751644969 Chr.14: 23351889-23351889 on Build GRCh38
rs1053874000 Chr.14: 23352101-23352101 on Build GRCh38
rs779282529 Chr.14: 23352117-23352117 on Build GRCh38
rs958199364 Chr.14: 23351703-23351703 on Build GRCh38
rs781375939 Chr.14: 23351760-23351760 on Build GRCh38
rs749146365 Chr.14: 23351777-23351777 on Build GRCh38
rs746461323 Chr.14: 23351831-23351831 on Build GRCh38
rs776906049 Chr.14: 23352073-23352073 on Build GRCh38
rs760890969 Chr.14: 23351966-23351966 on Build GRCh38
rs370223410 Chr.14: 23352035-23352035 on Build GRCh38
rs780884240 Chr.14: 23352020-23352020 on Build GRCh38
rs772006019 Chr.14: 23351814-23351814 on Build GRCh38
rs766395004 Chr.14: 23351739-23351739 on Build GRCh38
rs1039579420 Chr.14: 23351852-23351852 on Build GRCh38
rs775569647 Chr.14: 23352051-23352051 on Build GRCh38
rs775015962 Chr.14: 23351715-23351715 on Build GRCh38
rs753612649 Chr.14: 23352058-23352096 on Build GRCh38
rs144107183 Chr.14: 23351933-23351933 on Build GRCh38
rs200344917 Chr.14: 23352103-23352103 on Build GRCh38
rs748985510 Chr.14: 23352111-23352111 on Build GRCh38
rs751587001 Chr.14: 23351743-23351743 on Build GRCh38
rs200761849 Chr.14: 23351891-23351891 on Build GRCh38
rs375667142 Chr.14: 23351736-23351736 on Build GRCh38
rs562284046 Chr.14: 23352008-23352008 on Build GRCh38
rs765594626 Chr.14: 23351873-23351873 on Build GRCh38
rs146673692 Chr.14: 23351954-23351954 on Build GRCh38
rs764261485 Chr.14: 23352071-23352071 on Build GRCh38
rs771315337 Chr.14: 23352046-23352046 on Build GRCh38
rs372265182 Chr.14: 23352010-23352010 on Build GRCh38
rs779899269 Chr.14: 23352004-23352004 on Build GRCh38
rs932210156 Chr.14: 23351858-23351858 on Build GRCh38
rs574164261 Chr.14: 23351683-23351683 on Build GRCh38
rs750244904 Chr.14: 23351990-23351990 on Build GRCh38
rs766847637 Chr.14: 23351883-23351883 on Build GRCh38
rs759915561 Chr.14: 23351868-23351868 on Build GRCh38
rs772615374 Chr.14: 23352129-23352129 on Build GRCh38
rs763359500 Chr.14: 23351931-23351931 on Build GRCh38
rs747829437 Chr.14: 23351926-23351926 on Build GRCh38
rs771459884 Chr.14: 23351713-23351713 on Build GRCh38
rs756794632 Chr.14: 23352112-23352112 on Build GRCh38
rs774888933 Chr.14: 23351941-23351941 on Build GRCh38
rs367910877 Chr.14: 23351881-23351881 on Build GRCh38
rs373630736 Chr.14: 23351892-23351892 on Build GRCh38
rs780029661 Chr.14: 23351918-23351918 on Build GRCh38
rs772529185 Chr.14: 23352050-23352050 on Build GRCh38
rs774867319 Chr.14: 23351820-23351820 on Build GRCh38
rs371094181 Chr.14: 23351716-23351716 on Build GRCh38
rs547915381 Chr.14: 23352110-23352110 on Build GRCh38
rs562079314 Chr.14: 23351717-23351717 on Build GRCh38
rs745423097 Chr.14: 23351711-23351711 on Build GRCh38
rs768251970 Chr.14: 23351920-23351920 on Build GRCh38
rs769452086 Chr.14: 23351929-23351929 on Build GRCh38
rs370600233 Chr.14: 23351798-23351798 on Build GRCh38
rs752298906 Chr.14: 23352107-23352107 on Build GRCh38
rs746181868 Chr.14: 23352120-23352120 on Build GRCh38
rs774641507 Chr.14: 23352047-23352047 on Build GRCh38

View Full Product Details