Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23415926-23416428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MHRT
Entrez Gene ID
104564225
Gene Name
myosin heavy chain-associated RNA transcript
Gene Aliases
Myheart
UniGene
Gene Chromosome Location
Chr.14: 23415450-23417595 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075528 c.4895C>T p.A1632V Chr.14: 23416062-23416062 on Build GRCh38
3983193 c.4767C>A p.N1589K Chr.14: 23416190-23416190 on Build GRCh38
6075527 c.4772T>A p.L1591Q Chr.14: 23416185-23416185 on Build GRCh38
3885898 c.4738G>A p.D1580N Chr.14: 23416219-23416219 on Build GRCh38
416235 c.4741G>A p.E1581K Chr.14: 23416216-23416216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200593263 Chr.14: 23416284-23416284 on Build GRCh38
rs727505176 Chr.14: 23416294-23416294 on Build GRCh38
rs762272480 Chr.14: 23416060-23416060 on Build GRCh38
rs369699564 Chr.14: 23415977-23415977 on Build GRCh38
rs150241539 Chr.14: 23416049-23416049 on Build GRCh38
rs775613023 Chr.14: 23416350-23416350 on Build GRCh38
rs730880806 Chr.14: 23416278-23416278 on Build GRCh38
rs750987717 Chr.14: 23416240-23416240 on Build GRCh38
rs757569128 Chr.14: 23416364-23416364 on Build GRCh38
rs143471552 Chr.14: 23416301-23416301 on Build GRCh38
rs146858930 Chr.14: 23416169-23416169 on Build GRCh38
rs757517169 Chr.14: 23416272-23416287 on Build GRCh38
rs151113658 Chr.14: 23416046-23416046 on Build GRCh38
rs755294127 Chr.14: 23416366-23416366 on Build GRCh38
rs397516227 Chr.14: 23416237-23416237 on Build GRCh38
rs730880808 Chr.14: 23416185-23416185 on Build GRCh38
rs779213919 Chr.14: 23416337-23416337 on Build GRCh38
rs730880912 Chr.14: 23416308-23416308 on Build GRCh38
rs772521188 Chr.14: 23415964-23415964 on Build GRCh38
rs746331276 Chr.14: 23416347-23416350 on Build GRCh38
rs995158635 Chr.14: 23416396-23416396 on Build GRCh38
rs397516232 Chr.14: 23416057-23416057 on Build GRCh38
rs45476496 Chr.14: 23416232-23416232 on Build GRCh38
rs914575620 Chr.14: 23415950-23415950 on Build GRCh38
rs201326686 Chr.14: 23416285-23416285 on Build GRCh38
rs1051971790 Chr.14: 23416307-23416307 on Build GRCh38
rs565663412 Chr.14: 23416063-23416063 on Build GRCh38
rs527948764 Chr.14: 23416274-23416274 on Build GRCh38
rs369164804 Chr.14: 23416064-23416064 on Build GRCh38
rs764365579 Chr.14: 23415969-23415969 on Build GRCh38
rs587779393 Chr.14: 23416020-23416020 on Build GRCh38
rs34479915 Chr.14: 23415963-23415964 on Build GRCh38
rs773149848 Chr.14: 23416362-23416362 on Build GRCh38
rs587779390 Chr.14: 23416162-23416162 on Build GRCh38
rs757090529 Chr.14: 23416125-23416125 on Build GRCh38
rs777116428 Chr.14: 23415972-23415972 on Build GRCh38
rs141122361 Chr.14: 23416048-23416048 on Build GRCh38
rs755981142 Chr.14: 23416109-23416109 on Build GRCh38
rs775491087 Chr.14: 23416280-23416280 on Build GRCh38
rs797045730 Chr.14: 23416071-23416071 on Build GRCh38
rs775990796 Chr.14: 23415965-23415965 on Build GRCh38
rs797044596 Chr.14: 23416105-23416107 on Build GRCh38
rs769259796 Chr.14: 23416054-23416054 on Build GRCh38
rs752682962 Chr.14: 23415993-23415993 on Build GRCh38
rs397516228 Chr.14: 23416154-23416154 on Build GRCh38
rs570079347 Chr.14: 23416298-23416298 on Build GRCh38
rs397516230 Chr.14: 23416119-23416119 on Build GRCh38
rs535961150 Chr.14: 23416338-23416338 on Build GRCh38
rs926251807 Chr.14: 23416371-23416371 on Build GRCh38
rs751747383 Chr.14: 23416096-23416096 on Build GRCh38
rs745415989 Chr.14: 23415956-23415956 on Build GRCh38
rs45442096 Chr.14: 23416102-23416102 on Build GRCh38
rs765004831 Chr.14: 23416206-23416206 on Build GRCh38
rs769006978 Chr.14: 23416136-23416136 on Build GRCh38
rs766561921 Chr.14: 23416085-23416085 on Build GRCh38
rs373509029 Chr.14: 23415991-23415991 on Build GRCh38
rs397516229 Chr.14: 23416123-23416123 on Build GRCh38
rs142694139 Chr.14: 23416297-23416297 on Build GRCh38
rs587781085 Chr.14: 23416130-23416130 on Build GRCh38
rs373514686 Chr.14: 23416140-23416140 on Build GRCh38
rs761921843 Chr.14: 23415975-23415975 on Build GRCh38
rs777500254 Chr.14: 23415993-23415993 on Build GRCh38
rs933520031 Chr.14: 23416000-23416000 on Build GRCh38
rs371949574 Chr.14: 23415971-23415971 on Build GRCh38
rs949337517 Chr.14: 23416315-23416315 on Build GRCh38
rs761723774 Chr.14: 23416187-23416187 on Build GRCh38
rs202026847 Chr.14: 23416142-23416142 on Build GRCh38
rs1037858653 Chr.14: 23415974-23415974 on Build GRCh38
rs750487401 Chr.14: 23415989-23415989 on Build GRCh38
rs730880913 Chr.14: 23416126-23416126 on Build GRCh38
rs587779391 Chr.14: 23416134-23416134 on Build GRCh38
rs979509908 Chr.14: 23416058-23416058 on Build GRCh38
rs121913648 Chr.14: 23416106-23416108 on Build GRCh38
rs757702936 Chr.14: 23416336-23416336 on Build GRCh38
rs771236158 Chr.14: 23416277-23416277 on Build GRCh38
rs779715863 Chr.14: 23416236-23416236 on Build GRCh38
rs550244337 Chr.14: 23416295-23416295 on Build GRCh38
rs730880805 Chr.14: 23416293-23416293 on Build GRCh38
rs941699326 Chr.14: 23416392-23416392 on Build GRCh38
rs145822086 Chr.14: 23416053-23416053 on Build GRCh38
rs770477774 Chr.14: 23416268-23416268 on Build GRCh38
rs372362265 Chr.14: 23416334-23416334 on Build GRCh38
rs777416671 Chr.14: 23416014-23416014 on Build GRCh38
rs899401418 Chr.14: 23416369-23416369 on Build GRCh38
rs376604168 Chr.14: 23415987-23415987 on Build GRCh38
rs527491211 Chr.14: 23416395-23416395 on Build GRCh38
rs730880809 Chr.14: 23416150-23416150 on Build GRCh38
rs746571601 Chr.14: 23416135-23416135 on Build GRCh38
rs762762532 Chr.14: 23416289-23416289 on Build GRCh38
rs759691931 Chr.14: 23416171-23416171 on Build GRCh38
rs201469051 Chr.14: 23416217-23416217 on Build GRCh38
rs730880810 Chr.14: 23416129-23416129 on Build GRCh38
rs756827282 Chr.14: 23416038-23416038 on Build GRCh38
rs587779415 Chr.14: 23416051-23416051 on Build GRCh38
rs587779392 Chr.14: 23416122-23416122 on Build GRCh38
rs749154313 Chr.14: 23416016-23416016 on Build GRCh38
rs774540446 Chr.14: 23416170-23416170 on Build GRCh38
rs991408385 Chr.14: 23416160-23416160 on Build GRCh38
rs7140196 Chr.14: 23416241-23416241 on Build GRCh38
rs988401698 Chr.14: 23416260-23416260 on Build GRCh38
rs1008225259 Chr.14: 23415999-23415999 on Build GRCh38
rs796681603 Chr.14: 23416319-23416319 on Build GRCh38
rs769673559 Chr.14: 23416361-23416361 on Build GRCh38
rs756142583 Chr.14: 23416008-23416008 on Build GRCh38
rs752197136 Chr.14: 23416310-23416310 on Build GRCh38
rs797044600 Chr.14: 23416194-23416194 on Build GRCh38
rs200530211 Chr.14: 23416141-23416141 on Build GRCh38
rs545875689 Chr.14: 23416056-23416056 on Build GRCh38
rs758571263 Chr.14: 23415990-23415990 on Build GRCh38
rs781731991 Chr.14: 23416139-23416139 on Build GRCh38
rs368722536 Chr.14: 23416279-23416279 on Build GRCh38
rs764130237 Chr.14: 23416118-23416118 on Build GRCh38
rs756657320 Chr.14: 23416327-23416327 on Build GRCh38
rs955638282 Chr.14: 23415996-23415996 on Build GRCh38
rs200601164 Chr.14: 23416286-23416286 on Build GRCh38
rs772232610 Chr.14: 23416339-23416339 on Build GRCh38
rs376843222 Chr.14: 23416270-23416270 on Build GRCh38
rs199557822 Chr.14: 23416250-23416250 on Build GRCh38
rs397516231 Chr.14: 23416093-23416093 on Build GRCh38
rs864309553 Chr.14: 23416042-23416042 on Build GRCh38
rs951187471 Chr.14: 23416137-23416137 on Build GRCh38
rs763882473 Chr.14: 23416177-23416177 on Build GRCh38
rs778453839 Chr.14: 23416328-23416328 on Build GRCh38
rs779524255 Chr.14: 23416320-23416320 on Build GRCh38
rs765324390 Chr.14: 23416061-23416061 on Build GRCh38
rs730880807 Chr.14: 23416248-23416248 on Build GRCh38
rs142034311 Chr.14: 23416151-23416151 on Build GRCh38
rs749695436 Chr.14: 23416100-23416101 on Build GRCh38
rs747125455 Chr.14: 23416358-23416358 on Build GRCh38
rs778651539 Chr.14: 23416043-23416043 on Build GRCh38

View Full Product Details