Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24102671-24103166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRL
Entrez Gene ID
4901
Gene Name
neural retina leucine zipper
Gene Aliases
D14S46E, NRL-MAF, RP27
UniGene
Hs.652297
Gene Chromosome Location
Chr.14: 24078693-24114924 on Build GRCh38
Transcripts
XM_005267708.4 XM_005267710.3 XM_017021350.1 NM_001291556.1 NM_001308054.1 NM_004563.3 XM_006720158.2

Gene Symbol
PCK2
Entrez Gene ID
5106
Gene Name
phosphoenolpyruvate carboxykinase 2, mitochondrial
Gene Aliases
PEPCK, PEPCK-M, PEPCK2
UniGene
Hs.75812
Gene Chromosome Location
Chr.14: 24094131-24104132 on Build GRCh38
Transcripts
XM_005267708.4 XM_005267710.3 XM_017021350.1 NM_001291556.1 NM_001308054.1 NM_004563.3 XM_006720158.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138047 c.1372+1G>A p.? Chr.14: 24102891-24102891 on Build GRCh38
4050160 c.1342G>A p.A448T Chr.14: 24102860-24102860 on Build GRCh38
954880 c.1343C>T p.A448V Chr.14: 24102861-24102861 on Build GRCh38
1477496 c.1334C>A p.P445H Chr.14: 24102852-24102852 on Build GRCh38
1477495 c.1270T>C p.F424L Chr.14: 24102788-24102788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999633059 Chr.14: 24102857-24102857 on Build GRCh38
rs762854352 Chr.14: 24102842-24102842 on Build GRCh38
rs185687744 Chr.14: 24103078-24103078 on Build GRCh38
rs374717926 Chr.14: 24102757-24102757 on Build GRCh38
rs868742031 Chr.14: 24103039-24103039 on Build GRCh38
rs371591685 Chr.14: 24102722-24102722 on Build GRCh38
rs959198675 Chr.14: 24103088-24103088 on Build GRCh38
rs755240223 Chr.14: 24103115-24103115 on Build GRCh38
rs376631075 Chr.14: 24102799-24102799 on Build GRCh38
rs755717442 Chr.14: 24102732-24102732 on Build GRCh38
rs760474897 Chr.14: 24102879-24102879 on Build GRCh38
rs1012470681 Chr.14: 24102909-24102909 on Build GRCh38
rs370631813 Chr.14: 24102804-24102804 on Build GRCh38
rs200437491 Chr.14: 24102785-24102785 on Build GRCh38
rs747603045 Chr.14: 24102850-24102850 on Build GRCh38
rs748188739 Chr.14: 24103131-24103131 on Build GRCh38
rs538683533 Chr.14: 24103116-24103116 on Build GRCh38
rs778357924 Chr.14: 24102737-24102737 on Build GRCh38
rs779493794 Chr.14: 24102809-24102809 on Build GRCh38
rs770011316 Chr.14: 24102891-24102895 on Build GRCh38
rs367927444 Chr.14: 24102709-24102709 on Build GRCh38
rs761942693 Chr.14: 24102757-24102758 on Build GRCh38
rs933709180 Chr.14: 24102929-24102929 on Build GRCh38
rs775809596 Chr.14: 24102773-24102773 on Build GRCh38
rs530998057 Chr.14: 24102859-24102859 on Build GRCh38
rs773578521 Chr.14: 24102877-24102877 on Build GRCh38
rs1016686430 Chr.14: 24102931-24102931 on Build GRCh38
rs751362111 Chr.14: 24102791-24102791 on Build GRCh38
rs559309673 Chr.14: 24102803-24102803 on Build GRCh38
rs760364621 Chr.14: 24103137-24103137 on Build GRCh38
rs759399802 Chr.14: 24102795-24102795 on Build GRCh38
rs945299391 Chr.14: 24102886-24102886 on Build GRCh38
rs770241923 Chr.14: 24102716-24102716 on Build GRCh38
rs916154 Chr.14: 24102702-24102702 on Build GRCh38
rs759298772 Chr.14: 24102905-24102905 on Build GRCh38
rs889938950 Chr.14: 24102957-24102957 on Build GRCh38
rs747288937 Chr.14: 24102838-24102838 on Build GRCh38
rs547327387 Chr.14: 24103060-24103060 on Build GRCh38
rs774378735 Chr.14: 24102848-24102848 on Build GRCh38
rs1049516059 Chr.14: 24102949-24102949 on Build GRCh38
rs752400018 Chr.14: 24102798-24102798 on Build GRCh38
rs200768361 Chr.14: 24102818-24102818 on Build GRCh38
rs770726015 Chr.14: 24102844-24102844 on Build GRCh38
rs369939667 Chr.14: 24102821-24102821 on Build GRCh38
rs180748619 Chr.14: 24102729-24102729 on Build GRCh38
rs780078243 Chr.14: 24102824-24102824 on Build GRCh38
rs143639714 Chr.14: 24102840-24102840 on Build GRCh38
rs375633420 Chr.14: 24102901-24102901 on Build GRCh38
rs371724693 Chr.14: 24102743-24102743 on Build GRCh38
rs780533661 Chr.14: 24102937-24102937 on Build GRCh38
rs771412007 Chr.14: 24102753-24102753 on Build GRCh38
rs544405700 Chr.14: 24102710-24102710 on Build GRCh38
rs747201054 Chr.14: 24102938-24102938 on Build GRCh38
rs897371703 Chr.14: 24103010-24103010 on Build GRCh38
rs766166255 Chr.14: 24102789-24102789 on Build GRCh38
rs565080985 Chr.14: 24102846-24102846 on Build GRCh38
rs771651008 Chr.14: 24103006-24103006 on Build GRCh38
rs774165729 Chr.14: 24102897-24102898 on Build GRCh38
rs200079405 Chr.14: 24102878-24102878 on Build GRCh38
rs750360289 Chr.14: 24102704-24102704 on Build GRCh38
rs907225145 Chr.14: 24103065-24103065 on Build GRCh38
rs746074433 Chr.14: 24102764-24102764 on Build GRCh38
rs769923011 Chr.14: 24103142-24103142 on Build GRCh38
rs530386104 Chr.14: 24102928-24102928 on Build GRCh38
rs139687683 Chr.14: 24102768-24102768 on Build GRCh38
rs747563941 Chr.14: 24102731-24102731 on Build GRCh38
rs751218676 Chr.14: 24102723-24102723 on Build GRCh38
rs371329992 Chr.14: 24102786-24102786 on Build GRCh38
rs768717840 Chr.14: 24102839-24102839 on Build GRCh38
rs201015155 Chr.14: 24102921-24102921 on Build GRCh38
rs753451077 Chr.14: 24102906-24102906 on Build GRCh38
rs760676357 Chr.14: 24102783-24102783 on Build GRCh38
rs747231614 Chr.14: 24103087-24103087 on Build GRCh38
rs558675327 Chr.14: 24103143-24103143 on Build GRCh38
rs781416617 Chr.14: 24103119-24103119 on Build GRCh38
rs941543416 Chr.14: 24102966-24102966 on Build GRCh38
rs551184706 Chr.14: 24102860-24102860 on Build GRCh38

View Full Product Details